Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Primary Antibodies ›
  • GSK3 alpha Antibodies

Proteintech

Phospho-GSK3A (Ser21) Polyclonal Antibody

View all (24) GSK3 alpha antibodies

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Protocols
Questions & Answers
Datasheet
Protocols
Questions & Answers

Cite Phospho-GSK3A (Ser21) Polyclonal Antibody

  • Antibody Testing Data (1)
Phospho-GSK3A (Ser21) Antibody in Western Blot (WB)
Group 53 Created with Sketch.
Phospho-GSK3A (Ser21) Antibody in Western Blot (WB)
Group 53 Created with Sketch.

FIGURE: 1 / 1

{{ $ctrl.currentElement.advancedVerification.fullName }}
{{ $ctrl.currentElement.advancedVerification.text }}

Phospho-GSK3A (Ser21) Antibody (28875-1-AP) in WB

Non-treated PC-3 and Calyculin A treated PC-3 cells were subjected to SDS PAGE followed by western blot with 28875-1-AP (Phospho-GSK3A (S21) antibody) at dilution of 1:3000 incubated at 48451 overnight. The membrane was stripped and re-blotted with GAPDH antibody as loading control. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
Published figure supplied by benchsci-logo
PMID: {{$ctrl.currentElement.benchSciPubmedId}}
View Product

{{$ctrl.videoDesc}}

{{$ctrl.videoLongDesc}}

Phospho-GSK3A (Ser21) Antibody in Western Blot (WB)

Product Details

28875-1-AP

Applications
Tested Dilution
Publications

Western Blot (WB)

1:3,000
-
Product Specifications

Species Reactivity

Human

Host/Isotype

Rabbit / IgG

Class

Polyclonal

Type

Antibody

Immunogen

Peptide

Conjugate

Unconjugated Unconjugated Unconjugated

Form

Liquid

Concentration

400 µg/mL

Purification

Antigen affinity chromatography

Storage buffer

PBS, pH 7.3, with 50% glycerol

Contains

0.02% sodium azide

Storage conditions

-20°C

Shipping conditions

Wet ice

Product Specific Information

Aliquoting is unnecessary for -20°C storage.

Target Information

Glycogen synthase kinase 3 alpha (GSK3A) is a multifunctional protein serine/threonine kinase implicated in the control of several regulatory proteins and transcription factors. (GSK3A Alias: DKFZp686D0638). Human GSTs can be divided into five main classes: alpha, mu, pi, theta, and zeta. The theta class includes GSTT1 and GSTT2. The GSTT1 and GSTT2 share 55% amino acid sequence identity and both of them were claimed to have an important role in human carcinogenesis. The GSTT1 gene is located approximately 50kb away from the GSTT2 gene. The GSTT1 and GSTT2 genes have a similar structure, being composed of five exons with identical exon/intron boundaries.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

References (0)

Have you cited this product in a publication?
Let us know so we can reference it here.
Cite this product

Bioinformatics

Protein Aliases: DKFZp686D0638; Glycogen synthase kinase-3 alpha; GSK-3 alpha; gsk-3-alpha; OTTHUMP00000198368; Serine/threonine-protein kinase GSK3A

View more View less

Gene Aliases: GSK3A

View more View less

UniProt ID: (Human) P49840

View more View less

Entrez Gene ID: (Human) 2931

View more View less

Function(s)
protein serine/threonine kinase activity protein binding ATP binding protein kinase A catalytic subunit binding tau-protein kinase activity
Process(es)
regulation of systemic arterial blood pressure cardiac left ventricle morphogenesis glycogen metabolic process protein phosphorylation dopamine receptor signaling pathway nervous system development insulin receptor signaling pathway negative regulation of UDP-glucose catabolic process Wnt signaling pathway positive regulation of cAMP biosynthetic process negative regulation of TOR signaling cellular response to insulin stimulus cellular response to interleukin-3 IRE1-mediated unfolded protein response proteasome-mediated ubiquitin-dependent protein catabolic process negative regulation of glycogen biosynthetic process positive regulation of protein catabolic process positive regulation of heart contraction negative regulation of glucose import negative regulation of insulin receptor signaling pathway negative regulation of cell growth involved in cardiac muscle cell development positive regulation of adrenergic receptor signaling pathway negative regulation of canonical Wnt signaling pathway extrinsic apoptotic signaling pathway in absence of ligand chemical synaptic transmission, postsynaptic positive regulation of mitochondrial outer membrane permeabilization involved in apoptotic signaling pathway regulation of mitophagy positive regulation of protein targeting to mitochondrion negative regulation of glycogen synthase activity, transferring glucose-1-phosphate negative regulation of type B pancreatic cell development negative regulation of glycogen (starch) synthase activity positive regulation of glycogen (starch) synthase activity
It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-9dd846b7f-s9zn4:80/100.66.128.142:80.
git-commit: 17e8f6c92ed407c129a469238c057b56fdf8abbb
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0