Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Primary Antibodies ›
  • DNMT3A Antibodies

Bioss

Phospho-Dnmt3a (Ser390, Ser393) Polyclonal Antibody, HRP

View all (31) DNMT3A antibodies

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Protocols
Questions & Answers
Datasheet
Protocols
Questions & Answers

Cite Phospho-Dnmt3a (Ser390, Ser393) Polyclonal Antibody, HRP

Phospho-Dnmt3a (Ser390, Ser393) Polyclonal Antibody, HRP

Product Details

BS-14399R-HRP

Applications
Tested Dilution
Publications

Western Blot (WB)

1:300-1:5,000
-

ELISA (ELISA)

1:500-1,000
-
Product Specifications

Species Reactivity

Human

Host/Isotype

Rabbit / IgG

Class

Polyclonal

Type

Antibody

Immunogen

KLH conjugated Synthesised phosphopeptide derived from human Dnmt3a around the phosphorylation site of Ser390 & Ser393.
View immunogen

Conjugate

HRP HRP HRP

Form

Liquid

Concentration

1 mg/mL

Purification

Protein A

Storage buffer

TBS, pH 7.4, with 50% glycerol, 1% BSA

Contains

0.03% ProClin 300

Storage conditions

-20°C or -80°C if preferred

Shipping conditions

Ambient (domestic); Wet ice (international)

Target Information

Methylation of DNA at cytosine residues plays an important role in regulation of gene expression, genomic imprinting and is essential for mammalian development. Hypermethylation of CpG islands in tumor suppressor genes or hypomethylation of bulk genomic DNA may be linked with development of cancer. To date, 3 families of mammalian DNA methyltransferase genes have been identified which include Dnmt1, Dnmt2 and Dnmt3. Dnmt1 is constitutively expressed in proliferating cells and inactivation of this gene causes global demethylation of genomic DNA and embryonic lethality. Dnmt2 is expressed at low levels in adult tissues and its inactivation does not affect DNA methylation or maintenance of methylation. The Dnmt3 family members, Dnmt3a and Dnmt3b, are strongly expressed in ES cells but their expression is down regulated in differentiating ES cells and is low in adult somatic tissue. Recently, it has been shown that naturally occurring mutations of Dnmt3b gene occurs in patients with a rare autosomal recessive disorder, termed ICF (immunodeficiency, centromeric instability, and facial anomalies) syndrome.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

References (0)

Have you cited this product in a publication?
Let us know so we can reference it here.
Cite this product

Bioinformatics

Protein Aliases: AU015806; Cysteine methyltransferase DNMT3A; DNA (cytosine-5)-methyltransferase 3A; DNA (cytosine-5-)-methyltransferase 3 alpha; DNA cytosine methyltransferase 3A2; DNA methyltransferase HsaIIIA; DNA MTase HsaIIIA; Dnmt3a; W91664

View more View less

Gene Aliases: DNMT3A; DNMT3A2; M.HsaIIIA; TBRS

View more View less

UniProt ID: (Human) Q9Y6K1

View more View less

Entrez Gene ID: (Human) 1788

View more View less

Function(s)
DNA binding chromatin binding DNA (cytosine-5-)-methyltransferase activity protein binding DNA-methyltransferase activity identical protein binding unmethylated CpG binding metal ion binding DNA (cytosine-5-)-methyltransferase activity, acting on CpG substrates
Process(es)
negative regulation of transcription from RNA polymerase II promoter mitotic cell cycle DNA methylation methylation-dependent chromatin silencing regulation of gene expression by genetic imprinting spermatogenesis aging response to toxic substance response to ionizing radiation response to lead ion DNA methylation on cytosine within a CG sequence positive regulation of cell death neuron differentiation response to estradiol response to vitamin A response to antidepressant response to cocaine DNA methylation involved in embryo development DNA methylation involved in gamete generation hypermethylation of CpG island negative regulation of gene expression, epigenetic S-adenosylhomocysteine metabolic process S-adenosylmethioninamine metabolic process cellular response to amino acid stimulus cellular response to ethanol cellular response to hypoxia C-5 methylation of cytosine hepatocyte apoptotic process
It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-9dd846b7f-s9zn4:80/100.66.128.142:80.
git-commit: 17e8f6c92ed407c129a469238c057b56fdf8abbb
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0