Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Primary Antibodies ›
  • DNA Ligase IV Antibodies

Invitrogen

Phospho-DNA ligase IV (Thr650) Polyclonal Antibody

View all (19) DNA Ligase IV antibodies

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Protocols
Questions & Answers
Datasheet
Protocols
Questions & Answers

Cite Phospho-DNA ligase IV (Thr650) Polyclonal Antibody

  • Antibody Testing Data (3)
Phospho-DNA ligase IV (Thr650) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
Group 53 Created with Sketch.
Phospho-DNA ligase IV (Thr650) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
Group 53 Created with Sketch.

FIGURE: 1 / 3

{{ $ctrl.currentElement.advancedVerification.fullName }}
{{ $ctrl.currentElement.advancedVerification.text }}

Phospho-DNA ligase IV (Thr650) Antibody (PA5-105827) in IHC (P)

Immunohistochemistry analysis of paraffin-embedded Phospho-DNA ligase IV (Thr650) in mouse kidney tissue sections. Antigen retrieval was performed using citrate buffer. Samples were blocked with blocking buffer (1.5 hr, 22°C), incubated with Phospho-DNA ligase IV (Thr650) polyclonal antibody (Product # PA5-105827) using a dilution of 1:200 (1.5 hr, 22°C), followed by HRP conjugated goat anti-rabbit. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
Published figure supplied by benchsci-logo
PMID: {{$ctrl.currentElement.benchSciPubmedId}}
View Product

{{$ctrl.videoDesc}}

{{$ctrl.videoLongDesc}}

Phospho-DNA ligase IV (Thr650) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
Phospho-DNA ligase IV (Thr650) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
Phospho-DNA ligase IV (Thr650) Antibody in Western Blot (WB)
Phospho-DNA ligase IV (Thr650) Polyclonal Antibody

Product Details

PA5-105827

Applications
Tested Dilution
Publications

Western Blot (WB)

1:1,000-1:3,000
-

Immunohistochemistry (Paraffin) (IHC (P))

1:50-1:200
-
Product Specifications

Species Reactivity

Human, Mouse

Host/Isotype

Rabbit / IgG

Class

Polyclonal

Type

Antibody

Immunogen

A synthesized peptide derived from human LIG4(Accession P49917), corresponding to amino acid residues around phosphorylated Thr650.
View immunogen

Conjugate

Unconjugated Unconjugated Unconjugated

Form

Liquid

Concentration

1 mg/mL

Purification

sequential chromatography

Storage buffer

PBS, pH 7.4, with 50% glycerol

Contains

0.02% sodium azide

Storage conditions

-20°C

Shipping conditions

Ambient (domestic); Wet ice (international)

RRID

AB_2817226

Product Specific Information

Antibody detects endogenous levels of DNA Ligase IV only when phosphorylated at Thr650.

Target Information

The protein encoded by this gene is a DNA ligase that joins single-strand breaks in a double-stranded polydeoxynucleotide in an ATP-dependent reaction. This protein is essential for V(D)J recombination and DNA double-strand break (DSB) repair through nonhomologous end joining (NHEJ). This protein forms a complex with the X-ray repair cross complementing protein 4 (XRCC4), and further interacts with the DNA-dependent protein kinase (DNA-PK). Both XRCC4 and DNA-PK are known to be required for NHEJ. The crystal structure of the complex formed by this protein and XRCC4 has been resolved. Defects in this gene are the cause of LIG4 syndrome. Alternatively spliced transcript variants encoding the same protein have been observed.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

References (0)

Have you cited this product in a publication?
Let us know so we can reference it here.
Cite this product

Bioinformatics

Protein Aliases: DNA joinase; DNA ligase 4; DNA ligase IV; DNA repair enzyme; ligase IV, DNA, ATP-dependent; Polydeoxyribonucleotide synthase [ATP] 4; polynucleotide ligase; sealase

View more View less

Gene Aliases: 5830471N16Rik; LIG4; LIG4S; tiny

View more View less

UniProt ID: (Human) P49917, (Mouse) Q8BTF7

View more View less

Entrez Gene ID: (Human) 3981, (Mouse) 319583

View more View less

Function(s)
DNA binding DNA ligase activity DNA ligase (ATP) activity protein binding ATP binding protein C-terminus binding ligase activity metal ion binding nucleotide binding
Process(es)
single strand break repair in utero embryonic development pro-B cell differentiation DNA replication DNA ligation DNA repair nucleotide-excision repair, DNA gap filling double-strand break repair double-strand break repair via nonhomologous end joining DNA recombination cellular response to DNA damage stimulus cell cycle central nervous system development cell proliferation response to X-ray response to ionizing radiation response to gamma radiation T cell differentiation in thymus V(D)J recombination immunoglobulin V(D)J recombination T cell receptor V(D)J recombination somatic stem cell population maintenance negative regulation of neuron apoptotic process isotype switching positive regulation of fibroblast proliferation positive regulation of neurogenesis DNA ligation involved in DNA recombination DNA ligation involved in DNA repair chromosome organization cell division neuron apoptotic process DNA biosynthetic process double-strand break repair via classical nonhomologous end joining lagging strand elongation cellular response to lithium ion establishment of integrated proviral latency positive regulation of chromosome organization
It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-c7d89468c-sbmwt:80/100.66.128.142:80.
git-commit: 4db89d68e0ee702f860d53b64def1eb94897fade
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0