Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Primary Antibodies ›
  • PAX7 Antibodies

Rockland

Pax7 Polyclonal Antibody

View all (34) PAX7 antibodies

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Protocols
Questions & Answers
Datasheet
Protocols
Questions & Answers

Cite Pax7 Polyclonal Antibody

  • Antibody Testing Data (1)
Pax7 Antibody in Western Blot (WB)
Group 53 Created with Sketch.
Pax7 Antibody in Western Blot (WB)
Group 53 Created with Sketch.

FIGURE: 1 / 1

{{ $ctrl.currentElement.advancedVerification.fullName }}
{{ $ctrl.currentElement.advancedVerification.text }}

Pax7 Antibody (600-401-GR7) in WB

Western Blot of Rabbit anti-Pax7 antibody. Lane 1: Opal Pre-stained ladder (p/n MB-210-0500). Lane 2: Mouse muscle protein lysate. Lane 3: C2C12 Lysate (p/n W09-001-GL7). Lane 4: Ms spleen lysate (p/n W10-000-T029). Lane 5: Ms brain lysate (p/n W10-000-T004). Lane 6: HEK293T Lysate (W09-001-GX5). Lane 7: HeLa WCL (p/n W09-000-364). Lane 8: MCF7 Lysate (p/n W09-000-360). Lane 9: Jurkat Lysate (p/n W09-001-370). Lane 10: A431 Lysate (p/n W09-000-361). Lane 11: A549 Lysate (p/n W09-001-372). Lane 12: LNCap Lysate (p/n W09-001-GJ9). Lane 13... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
Published figure supplied by benchsci-logo
PMID: {{$ctrl.currentElement.benchSciPubmedId}}
View Product

{{$ctrl.videoDesc}}

{{$ctrl.videoLongDesc}}

Pax7 Antibody in Western Blot (WB)

Product Details

600-401-GR7

Applications
Tested Dilution
Publications

Western Blot (WB)

1.0 µg/mL
-

ELISA (ELISA)

1:10,000-1:40,000
-
Product Specifications

Species Reactivity

Mouse

Host/Isotype

Rabbit / IgG

Class

Polyclonal

Type

Antibody

Immunogen

Pax7 affinity purified antibody was prepared from whole rabbit serum produced by repeated immunizations with a synthetic peptide corresponding to an internal region of Pax7 protein.

Conjugate

Unconjugated Unconjugated Unconjugated

Form

Liquid

Concentration

1 mg/mL

Purification

Affinity chromatography

Storage buffer

0.02M potassium phosphate, pH 7.2, with 0.15M NaCl

Contains

0.01% sodium azide

Storage conditions

-20°C, Avoid Freeze/Thaw Cycles

Shipping conditions

Wet ice

Product Specific Information

Store vial at -20° C prior to opening. Aliquot contents and freeze at -20° C or below for extended storage. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4° C as an undiluted liquid. Dilute only prior to immediate use.

A BLAST analysis was used to suggest cross reactivity with Pax7 protein from mouse (100% homology) and human (66.7% homology) for the immunizing sequence. Cross-reactivity with Pax7 from other sources has not been determined.

Target Information

This gene is a member of the paired box (PAX) family of transcription factors. Members of this gene family typically contain a paired box domain, an octapeptide, and a paired-type homeodomain. These genes play critical roles during fetal development and cancer growth. The specific function of the paired box 7 gene is unknown but speculated to involve tumor suppression since fusion of this gene with a forkhead domain family member has been associated with alveolar rhabdomyosarcoma. Three transcript variants encoding different isoforms have been found for this gene.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

References (0)

Have you cited this product in a publication?
Let us know so we can reference it here.
Cite this product

Bioinformatics

Protein Aliases: FLJ37460; paired box gene 7; Paired box protein Pax-7; paired box transcription factor PAX7; PAX7A

View more View less

Gene Aliases: Pax-7; Pax7

View more View less

UniProt ID: (Mouse) P47239

View more View less

Entrez Gene ID: (Mouse) 18509

View more View less

Function(s)
transcription factor activity, RNA polymerase II core promoter sequence-specific DNA binding transcription factor activity, sequence-specific DNA binding protein binding sequence-specific DNA binding
Process(es)
chromatin remodeling transcription, DNA-templated regulation of transcription, DNA-templated regulation of transcription from RNA polymerase II promoter transcription from RNA polymerase II promoter multicellular organism development muscle organ development skeletal muscle tissue development organ morphogenesis regulation of cell fate commitment regulation of gene expression skeletal muscle satellite cell commitment spinal cord association neuron differentiation dorsal/ventral neural tube patterning positive regulation of histone methylation regulation of protein binding skeletal muscle tissue regeneration positive regulation of transcription from RNA polymerase II promoter neuron fate commitment embryonic skeletal system development cartilage development muscle tissue morphogenesis positive regulation of myoblast proliferation
It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-6d4c5dbbff-7nlnn:80/100.66.128.142:80.
git-commit: 3992c1c7134a3a3ff95f78ba9fe94218727d3638
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0