Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Primary Antibodies ›
  • PP5 Antibodies

OriGene

PPP5C Monoclonal Antibody (OTI5A8), TrueMAB™

View all (39) PP5 antibodies

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Protocols
Questions & Answers
Datasheet
Protocols
Questions & Answers

Cite PPP5C Monoclonal Antibody (OTI5A8), TrueMAB™

  • Antibody Testing Data (6)
PPP5C Antibody in Immunohistochemistry (Paraffin) (IHC (P))
Group 53 Created with Sketch.
PPP5C Antibody in Immunohistochemistry (Paraffin) (IHC (P))
Group 53 Created with Sketch.

FIGURE: 1 / 6

{{ $ctrl.currentElement.advancedVerification.fullName }}
{{ $ctrl.currentElement.advancedVerification.text }}

PPP5C Antibody (TA500619) in IHC (P)

Immunohistochemical staining of paraffin-embedded endometrium tissue within the normal limits using anti-PPP5Cmouse monoclonal antibody. (Heat-induced epitope retrieval by 10mM citric buffer, pH6.0, 100°C for 10min, TA500619, Dilution 1:50) {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
Published figure supplied by benchsci-logo
PMID: {{$ctrl.currentElement.benchSciPubmedId}}
View Product

{{$ctrl.videoDesc}}

{{$ctrl.videoLongDesc}}

PPP5C Antibody in Immunohistochemistry (Paraffin) (IHC (P))
PPP5C Antibody in Immunohistochemistry (Paraffin) (IHC (P))
PPP5C Antibody in Immunohistochemistry (Paraffin) (IHC (P))
PPP5C Antibody in Immunohistochemistry (Paraffin) (IHC (P))
PPP5C Antibody in Western Blot (WB)
PPP5C Antibody in Western Blot (WB)
PPP5C Monoclonal Antibody (OTI5A8), TrueMAB™

Product Details

TA500619

Applications
Tested Dilution
Publications

Western Blot (WB)

1:1,000
-

Immunohistochemistry (Paraffin) (IHC (P))

1:50
-
Product Specifications

Species Reactivity

Dog, Human, Rat

Host/Isotype

Mouse / IgG1

Class

Monoclonal

Type

Antibody

Clone

OTI5A8

Immunogen

Full-length protein expressed in 293T cell transfected with human PPP5C expression vector
View immunogen

Conjugate

Unconjugated Unconjugated Unconjugated

Form

Liquid

Concentration

1.4 mg/mL

Purification

Affinity chromatography

Storage buffer

PBS with 1% BSA, 50% glycerol

Contains

0.02% sodium azide

Storage conditions

-20°C, Avoid Freeze/Thaw Cycles

Shipping conditions

Ambient (domestic); Wet ice (international)

Target Information

This gene encodes a serine/threonine phosphatase which is a member of the protein phosphatase catalytic subunit family. Proteins in this family participate in pathways regulated by reversible phosphorylation at serine and threonine residues; many of these pathways are involved in the regulation of cell growth and differentiation. The product of this gene has been shown to participate in signaling pathways in response to hormones or cellular stress, and elevated levels of this protein may be associated with breast cancer development. Alternative splicing results in multiple transcript variants.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

References (0)

Have you cited this product in a publication?
Let us know so we can reference it here.
Cite this product

Bioinformatics

Protein Aliases: FLJ36922; FLJ55954; OTTHUMP00000165834; PP-T; PP5; PPT; protein phosphatase 5, catalytic subunit; Protein phosphatase T; Serine/threonine-protein phosphatase 5

View more View less

Gene Aliases: PP5; PPP5; PPP5C; PPT

View more View less

UniProt ID: (Human) P53041, (Rat) P53042

View more View less

Entrez Gene ID: (Dog) 612199, (Human) 5536, (Rat) 65179

View more View less

Function(s)
G-protein alpha-subunit binding RNA binding phosphoprotein phosphatase activity protein serine/threonine phosphatase activity signal transducer activity protein binding ATP binding microtubule binding lipid binding heat shock protein binding identical protein binding ADP binding metal ion binding
Process(es)
negative regulation of protein phosphorylation protein dephosphorylation signal transduction response to lead ion dephosphorylation histone dephosphorylation positive regulation of I-kappaB kinase/NF-kappaB signaling response to morphine protein oligomerization protein heterooligomerization negative regulation of cell death cellular response to hydrogen peroxide cellular response to cadmium ion negative regulation of neuron death positive regulation of glucocorticoid receptor signaling pathway MAPK cascade DNA repair transcription, DNA-templated mitotic nuclear division
It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0