Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Primary Antibodies ›
  • PPP2R1B Antibodies

Proteintech

PPP2R1B Polyclonal Antibody

2 References
View all (18) PPP2R1B antibodies

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Protocols
Questions & Answers
Datasheet
Protocols
Questions & Answers

Cite PPP2R1B Polyclonal Antibody

  • Antibody Testing Data (5)
PPP2R1B Antibody in Immunohistochemistry (Paraffin) (IHC (P))
Group 53 Created with Sketch.
PPP2R1B Antibody in Immunohistochemistry (Paraffin) (IHC (P))
Group 53 Created with Sketch.

FIGURE: 1 / 5

{{ $ctrl.currentElement.advancedVerification.fullName }}
{{ $ctrl.currentElement.advancedVerification.text }}

PPP2R1B Antibody (12621-1-AP) in IHC (P)

Immunohistochemical analysis of paraffin-embedded human liver tissue slide using (Product # 12621-1-AP) (PPP2R1B antibody) at dilution of 1:200 (under 40x lens). Heat mediated antigen retrieval with Tris-EDTA buffer (pH 9.0). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
Published figure supplied by benchsci-logo
PMID: {{$ctrl.currentElement.benchSciPubmedId}}
View Product

{{$ctrl.videoDesc}}

{{$ctrl.videoLongDesc}}

PPP2R1B Antibody in Immunohistochemistry (Paraffin) (IHC (P))
PPP2R1B Antibody in Western Blot (WB)
PPP2R1B Antibody in Western Blot (WB)
PPP2R1B Antibody in Western Blot (WB)
PPP2R1B Antibody in Western Blot (WB)

Product Details

12621-1-AP

Applications
Tested Dilution
Publications

Western Blot (WB)

1:500-1:2,000
View 2 publications 2 publications

Immunohistochemistry (Paraffin) (IHC (P))

1:200
-
Product Specifications

Species Reactivity

Human, Mouse, Rat

Host/Isotype

Rabbit / IgG

Class

Polyclonal

Type

Antibody

Immunogen

PPP2R1B Fusion Protein Ag3307 (320-667 aa encoded by BC027596)

Conjugate

Unconjugated Unconjugated Unconjugated

Form

Liquid

Concentration

0.13 mg/mL

Purification

Antigen affinity chromatography

Storage buffer

PBS, pH 7.3, with 50% glycerol

Contains

0.02% sodium azide

Storage conditions

-20°C

Shipping conditions

Wet ice (domestic); Dry ice (international)

Product Specific Information

Immunogen sequence: ELGENLPIE DRETIIMNQI LPYIKELVSD TNQHVKSALA SVIMGLSTIL GKENTIEHLL PLFLAQLKDE CPDVRLNIIS NLDCVNEVIG IRQLSQSLLP AIVELAEDAK WRVRLAIIEY MPLLAGQLGV EFFDEKLNSL CMAWLVDHVY AIREAATNNL MKLVQKFGTE WAQNTIVPKV LVMANDPNYL HRMTTLFCIN ALSEACGQEI TTKQMLPIVL KMAGDQVANV RFNVAKSLQK IGPILDTNAL QGEVKPVLQK LGQDEDMDVK YFAQEAISVV AQRLRKLEFP VKDSGEPSVP RADKNHFPRP TVPGEDMGKG PVYQLRGDTR DTLAQLGIAE LVHFSQSTD (320-667 aa encoded by BC027596)

Target Information

In eukaryotes, the phosphorylation and dephosphorylation of proteins on serine and threonine residues is an essential means of regulating a broad range of cellular functions, including division, homeostasis and apoptosis. A group of proteins that are intimately involved in this process are the protein phosphatases. In general, the protein phosphatase (PP) holoenzyme is a trimeric complex composed of a regulatory subunit, a variable subunit and a catalytic subunit. Four major families of protein phosphatase catalytic subunits have been identified, designated PP1, PP2A, PP2B (calcineurin) and PP2C. An additional protein phosphatase catalytic subunit, PPX (also known as PP4), is a putative member of a novel PP family. The PP2A family comprises subfamily members PP2A alpha and PP2A beta. The PP2A catalytic subunit associates with a variety of regulatory subunits. Regulatory subunits include PP2A-A alpha and -A beta, PP2A-B alpha and -B beta, PP2A-C alpha and -C beta, and PP2A-B56 alpha and -B56 beta.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

Bioinformatics

Protein Aliases: PP2A -AR1 beta; PP2A A beta; PP2A subunit A isoform PR65-beta; PP2A subunit A isoform R1-beta; PP2A, subunit A, PR65-beta isoform; PP2A, subunit A, R1-beta isoform; PP2A-A beta; PP2A-APR65 beta; PP2AA beta; protein phosphatase 2 (formerly 2A), regulatory subunit A (PR 65), beta isoform; protein phosphatase 2 (formerly 2A), regulatory subunit A, beta isoform; protein phosphatase 2 regulatory subunit A, beta; protein phosphatase 2, regulatory subunit A, beta; protein phosphatase 2, structural/regulatory subunit A, beta; Serine/threonine-protein phosphatase 2A 65 kDa regulatory subunit A beta isoform

View more View less

Gene Aliases: 2410091N08Rik; AI790395; PP2A-Abeta; PPP2R1B; PR65B

View more View less

UniProt ID: (Human) P30154, (Mouse) Q7TNP2, (Rat) Q4QQT4

View more View less

Entrez Gene ID: (Human) 5519, (Mouse) 73699, (Rat) 315648

View more View less

Function(s)
protein binding protein phosphatase type 2A regulator activity molecular_function
Process(es)
protein complex assembly regulation of protein phosphatase type 2A activity apoptotic process involved in morphogenesis positive regulation of extrinsic apoptotic signaling pathway in absence of ligand biological_process
It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0