Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Primary Antibodies ›
  • Cyclophilin F Antibodies

OriGene

PPIF Monoclonal Antibody (OTI1C4), TrueMAB™

Advanced Verification
View all (24) Cyclophilin F antibodies

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Protocols
Questions & Answers
Datasheet
Protocols
Questions & Answers

Cite PPIF Monoclonal Antibody (OTI1C4), TrueMAB™

  • Antibody Testing Data (6)
  • Advanced Verification (1)
PPIF Antibody in Immunohistochemistry (Paraffin) (IHC (P))
Group 53 Created with Sketch.
PPIF Antibody in Immunohistochemistry (Paraffin) (IHC (P))
Group 53 Created with Sketch.

FIGURE: 1 / 7

{{ $ctrl.currentElement.advancedVerification.fullName }}
{{ $ctrl.currentElement.advancedVerification.text }}

PPIF Antibody (TA809029) in IHC (P)

Immunohistochemical staining of paraffin-embedded human lymphoma tissue using anti-PPIF mouse monoclonal antibody. (Heat-induced epitope retrieval by 1mM EDTA in 10mM Tris buffer (pH8.5) at 120°C for 3 min, TA809029)(1:150). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
Published figure supplied by benchsci-logo
PMID: {{$ctrl.currentElement.benchSciPubmedId}}
View Product

{{$ctrl.videoDesc}}

{{$ctrl.videoLongDesc}}

PPIF Antibody in Immunohistochemistry (Paraffin) (IHC (P))
PPIF Antibody in Immunohistochemistry (Paraffin) (IHC (P))
PPIF Antibody in Immunohistochemistry (Paraffin) (IHC (P))
PPIF Antibody in Immunohistochemistry (Paraffin) (IHC (P))
PPIF Antibody in Immunohistochemistry (Paraffin) (IHC (P))
PPIF Antibody in Western Blot (WB)
PPIF Antibody
PPIF Monoclonal Antibody (OTI1C4), TrueMAB™

Product Details

TA809029

Applications
Tested Dilution
Publications

Western Blot (WB)

Assay-dependent
-

Immunohistochemistry (Paraffin) (IHC (P))

1:150
-
Product Specifications

Species Reactivity

Human

Host/Isotype

Mouse / IgG2a

Class

Monoclonal

Type

Antibody

Clone

OTI1C4

Immunogen

Full length human recombinant protein of human PPIF produced in E.coli.
View immunogen

Conjugate

Unconjugated Unconjugated Unconjugated

Form

Liquid

Concentration

1 mg/mL

Purification

Affinity chromatography

Storage buffer

PBS with 1% BSA, 50% glycerol

Contains

0.02% sodium azide

Storage conditions

-20°C, Avoid Freeze/Thaw Cycles

Shipping conditions

Ambient (domestic); Wet ice (international)

Target Information

The protein encoded by this gene is a member of the peptidyl-prolyl cis-trans isomerase (PPIase) family. PPIases catalyze the cis-trans isomerization of proline imidic peptide bonds in oligopeptides and accelerate the folding of proteins. This protein is part of the mitochondrial permeability transition pore in the inner mitochondrial membrane. Activation of this pore is thought to be involved in the induction of apoptotic and necrotic cell death.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

References (0)

Have you cited this product in a publication?
Let us know so we can reference it here.
Cite this product

Bioinformatics

Protein Aliases: cyclophilin 3; Cyclophilin D; Cyclophilin F; CyP-D; CyP-M; Mitochondrial cyclophilin; mitochondrial peptidyl-prolyl cis-trans isomerase; Peptidyl-prolyl cis-trans isomerase F; Peptidyl-prolyl cis-trans isomerase F, mitochondrial; peptidyl-prolyl cis-trans isomerase, mitochondrial; PPIase F; Rotamase F

View more View less

Gene Aliases: Cyp-D; CyP-M; CYP3; CypD; PPIF

View more View less

UniProt ID: (Human) P30405

View more View less

Entrez Gene ID: (Human) 10105

View more View less

Function(s)
peptidyl-prolyl cis-trans isomerase activity protein binding cyclosporin A binding
Process(es)
protein peptidyl-prolyl isomerization response to ischemia protein folding apoptotic mitochondrial changes regulation of proton-transporting ATPase activity, rotational mechanism regulation of necrotic cell death negative regulation of ATPase activity negative regulation of apoptotic process regulation of mitochondrial membrane permeability necroptotic process cellular response to hydrogen peroxide cellular response to arsenic-containing substance cellular response to calcium ion positive regulation of release of cytochrome c from mitochondria negative regulation of release of cytochrome c from mitochondria negative regulation of oxidative phosphorylation regulation of mitochondrial membrane permeability involved in programmed necrotic cell death negative regulation of oxidative phosphorylation uncoupler activity negative regulation of intrinsic apoptotic signaling pathway
It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-55c564d944-rwwh4:80/100.66.128.142:80.
git-commit: 3092084cb0e494f0e07f07633350183de7fa10a8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0