Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: FFRSVFYRSY KEQESKKGFD QEEVFEKPTR ETCQGMRHAI YDKLDDDGLI APGVRVSGDD VIIGKTVTLP ENEDELESTN RRYTKRDCST FLRTSE
Highest antigen sequence identity to the following orthologs: Mouse - 99%, Rat - 100%.
This gene encodes the second largest subunit of RNA polymerase II, the polymerase responsible for synthesizing messenger RNA in eukaryotes. This subunit, in combination with at least two other polymerase subunits, forms a structure within the polymerase that maintains contact in the active site of the enzyme between the DNA template and the newly synthesized RNA.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 3'-5' exoribonuclease; DNA-directed RNA polymerase II 140 kDa polypeptide; DNA-directed RNA polymerase II subunit B; DNA-directed RNA polymerase II subunit RPB2; hs hRPB140; hsRPB2; polymerase (RNA) II (DNA directed) polypeptide B, 140kDa; polymerase (RNA) II subunit B; RNA polymerase II second largest subunit; RNA polymerase II subunit 2; RNA polymerase II subunit B2; RNA-directed RNA polymerase II subunit RPB2; rpb 2
Gene Aliases: hRPB140; POL2RB; POLR2B; RPB2
UniProt ID: (Human) P30876
Entrez Gene ID: (Human) 5431
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support