Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Primary Antibodies ›
  • PNPase Antibodies

Proteintech

PNPT1 Monoclonal Antibody (3F2A1)

View all (15) PNPase antibodies

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Protocols
Questions & Answers
Datasheet
Protocols
Questions & Answers

Cite PNPT1 Monoclonal Antibody (3F2A1)

  • Antibody Testing Data (1)
PNPT1 Antibody in Western Blot (WB)
Group 53 Created with Sketch.
PNPT1 Antibody in Western Blot (WB)
Group 53 Created with Sketch.

FIGURE: 1 / 1

{{ $ctrl.currentElement.advancedVerification.fullName }}
{{ $ctrl.currentElement.advancedVerification.text }}

PNPT1 Antibody (68309-1-IG) in WB

Various lysates were subjected to SDS PAGE followed by western blot with 68309-1-Ig (PNPT1 antibody) at dilution of 1:10000 and incubated at room temperature for 1.5 hours. The membrane was stripped and reblotted with HRP-conjugated Beta Actin Monoclonal antibody (HRP-66009) as loading control. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
Published figure supplied by benchsci-logo
PMID: {{$ctrl.currentElement.benchSciPubmedId}}
View Product

{{$ctrl.videoDesc}}

{{$ctrl.videoLongDesc}}

PNPT1 Antibody in Western Blot (WB)
PNPT1 Monoclonal Antibody (3F2A1)

Product Details

68309-1-IG

Applications
Tested Dilution
Publications

Western Blot (WB)

1:5,000-1:50,000
-

ELISA (ELISA)

Assay-dependent
-
Product Specifications

Species Reactivity

Human, Mouse, Rat

Host/Isotype

Mouse / IgG1

Class

Monoclonal

Type

Antibody

Clone

3F2A1

Immunogen

Recombinant protein
View immunogen

Conjugate

Unconjugated Unconjugated Unconjugated
  • CoraLite Plus 488
  • Request custom conjugation

Form

Liquid

Purification

Protein G

Storage buffer

PBS with 50% glycerol

Contains

0.02% sodium azide

Storage conditions

-20°C

Shipping conditions

Wet ice

Product Specific Information

Immunogen sequence: YEFPPYATNE IGKVTGLNRR ELGHGALAEK ALYPVIPRDF PFTIRVTSEV LESNGSSSMA SACGGSLALM DSGVPISSAV AGVAIGLVTK TDPEKGEIED YRLLTDILGI EDYNGDMDFK IAGTNKGITA LQADIKLPGI PIKIVMEAIQ QASVAKKEIL QIMNKTISKP RASRKENGPV VETVQVPLSK RAKFVGPGGY NLKKLQAETG VTISQVDEET FSVFAPTPSA MHEARDFITE ICKDDQEQQL EFGAVYTATI TEIRDTGVMV KLYPNMTAVL LHNTQLDQRK IKHPTALGLE VGQEIQVKYF GRDPADGRMR LSRKVLQSPA TTVVRTLNDR SSIVMGEPIS QSSSNSQ

Target Information

PNPT1 (polyribonucleotide nucleotidyltransferase 1, mitochondirla) is a RNA-binding protein implicated in numerous RNA metabolic processes. It catalyzes the phosphorolysis of single-stranded polyribonucleotides processively in the 3'-to-5' direction. PNPT1 is a component of the mitochondrial degradosome (mtEXO) complex that degrades 3' overhang double-stranded RNA with a 3'-to-5' directionality in an ATP-dependent manner. It is required for correct processing and polyadenylation of mitochondrial mRNAs. It plays a role as a cytoplasmic RNA import factor, in mitochondrial morphogenesis and respiration, regulation of expression of the electron transport chain, regulation of the stability of specific mature miRNAs in melanoma cells, and RNA cell surveilance. Mutations of the gene can result in combined oxidative phosphorylation deficiency 13 (COXPD13) and deafness, autosomal recessive, 70 (DFNB70).

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

References (0)

Have you cited this product in a publication?
Let us know so we can reference it here.
Cite this product

Bioinformatics

Protein Aliases: 3'-5' RNA exonuclease OLD35; DKFZp762K1914; old 35; PNPase 1; PNPase old-35; Polynucleotide phosphorylase 1; Polynucleotide phosphorylase-like protein; Polyribonucleotide nucleotidyltransferase 1, mitochondrial

View more View less

Gene Aliases: 1200003F12Rik; COXPD13; DFNB70; old-35; OLD35; PNPASE; PNPT1; Pnptl1

View more View less

UniProt ID: (Human) Q8TCS8, (Mouse) Q8K1R3

View more View less

Entrez Gene ID: (Human) 87178, (Rat) 360992, (Mouse) 71701

View more View less

Function(s)
3'-5'-exoribonuclease activity polyribonucleotide nucleotidyltransferase activity protein binding poly(U) RNA binding poly(G) binding miRNA binding poly(A) RNA binding nucleic acid binding RNA binding nuclease activity exonuclease activity transferase activity nucleotidyltransferase activity hydrolase activity
Process(es)
mitochondrial RNA catabolic process mitochondrial mRNA catabolic process positive regulation of mitochondrial RNA catabolic process mitochondrial RNA 5'-end processing mitochondrial RNA 3'-end processing RNA catabolic process mRNA catabolic process cellular response to oxidative stress cellular response to interferon-beta RNA import into mitochondrion rRNA import into mitochondrion regulation of cellular respiration RNA polyadenylation negative regulation of growth protein homooligomerization positive regulation of mRNA catabolic process protein homotrimerization mitochondrion morphogenesis nuclear polyadenylation-dependent mRNA catabolic process mitotic cell cycle arrest RNA phosphodiester bond hydrolysis, exonucleolytic mitochondrial mRNA polyadenylation positive regulation of miRNA catabolic process regulation of cellular senescence RNA processing mRNA processing transport
It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-9dd846b7f-s9zn4:80/100.66.128.142:80.
git-commit: 17e8f6c92ed407c129a469238c057b56fdf8abbb
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0