Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: MRPGGERPV EGGACNGRSE LELLKLRSAE CIDEAAERLG ALSRAIWSQP ELAYEEHHAH RVLTHFFERE PPAASWAVQP HYQLPTAFRA EWEPPEARAP SATPRPLHLG FLCEYDALPG IGHACGHNLI AEVGAAAALG VRGALEGLPR PPPPVKVVVL GTPAEEDGGG KIDLIEAGAF TNLDVVFMAH PSQENAAYLP DMAEHDVTVK YYGKASHSAS YPWEGLNALD AAVLAYNNLS VFRQQMKPTW RVHGIIKNGG VKPNIIPSYS ELIYYFRAPS MKELQVLTKK AEDCFRAAAL ASGCTVEIKG GAHDYYNVLP NKSLWKAYME NGRKLGIEFI SEDTMLNGPS GSTDFGNVSF VVPGIHPYFH IGSNALNHTE QYTEAAGSQE AQFYTLRTAK ALAMTALDVI FKPELLEGIR EDFKLKLQEE QFVNAVE (1-436 aa encoded by BC035036)
PM20D2 belongs to the peptidase M20A family. The function of PM20D2 remains unknown.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: aminoacylase 1-like 2; aminoacylase 1-like protein 2; aminoacylase-1-like protein 2; Beta-Ala-Lys dipeptidase; peptidase M20 domain-containing protein 2; Xaa-Arg dipeptidase; β-alanyl-lysine dipeptidase
Gene Aliases: ACY1L2; bA63L7.3; Gm424; PM20D2
UniProt ID: (Human) Q8IYS1, (Mouse) A3KG59
Entrez Gene ID: (Human) 135293, (Rat) 313130, (Mouse) 242377
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support