Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Primary Antibodies ›
  • CD31 (PECAM-1) Antibodies

Abnova

PECAM1 Monoclonal Antibody (1D2-1A5)

View all (297) CD31 (PECAM-1) antibodies

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Protocols
Questions & Answers
Datasheet
Protocols
Questions & Answers

Cite PECAM1 Monoclonal Antibody (1D2-1A5)

  • Antibody Testing Data (1)
PECAM1 Antibody in Western Blot (WB)
Group 53 Created with Sketch.
PECAM1 Antibody in Western Blot (WB)
Group 53 Created with Sketch.

FIGURE: 1 / 1

{{ $ctrl.currentElement.advancedVerification.fullName }}
{{ $ctrl.currentElement.advancedVerification.text }}

PECAM1 Antibody (H00005175-M01) in WB

Western Blot analysis of PECAM1 expression in transfected 293T cell line by PECAM1 monoclonal antibody (M01), clone 1D2-1A5. Lane 1: PECAM1 transfected lysate(82.4 KDa). Lane 2: Non-transfected lysate. . {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
Published figure supplied by benchsci-logo
PMID: {{$ctrl.currentElement.benchSciPubmedId}}
View Product

{{$ctrl.videoDesc}}

{{$ctrl.videoLongDesc}}

PECAM1 Antibody in Western Blot (WB)

Product Details

H00005175-M01

Applications
Tested Dilution
Publications

Western Blot (WB)

1-5 µg/mL
-

Immunohistochemistry (Paraffin) (IHC (P))

3 µg/mL
-

ELISA (ELISA)

10 µg/mL
-

Immunoprecipitation (IP)

1-5 µg/mL
-
Product Specifications

Species Reactivity

Human

Host/Isotype

Mouse / IgG1, kappa

Class

Monoclonal

Type

Antibody

Clone

1D2-1A5

Immunogen

PECAM1 (AAH22512, 29 a.a. approximately 738 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Conjugate

Unconjugated Unconjugated Unconjugated

Form

Liquid

Concentration

0.5-1 mg/mL

Purification

Protein A

Storage buffer

PBS, pH 7.4

Contains

no preservative

Storage conditions

-20°C, Avoid Freeze/Thaw Cycles

Shipping conditions

Wet ice

Product Specific Information

Product may be used with Western Blot (Transfected lysate).

Immunogen sequence: ENSFTINSVD MKSLPDWTVQ NGKNLTLQCF ADVSTTSHVK PQHQMLFYKD DVLFYNISSM KSTESYFIPE VRIYDSGTYK CTVIVNNKEK TTAEYQVLVE GVPSPRVTLD KKEAIQGGIV RVNCSVPEEK APIHFTIEKL ELNEKMVKLK REKNSRDQNF VILEFPVEEQ DRVLSFRCQA RIISGIHMQT SESTKSELVT VTESFSTPKF HISPTGMIME GAQLHIKCTI QVTHLAQEFP EIIIQKDKAI VAHNRHGNKA VYSVMAMVEH SGNYTCKVES SRISKVSSIV VNITELFSKP ELESSFTHLD QGERLNLSCS IPGAPPANFT IQKEDTIVSQ TQDFTKIASK SDSGTYICTA GIDKVVKKSN TVQIVVCEML SQPRISYDAQ FEVIKGQTIE VRCESISGTL PISYQLLKTS KVLENSTKNS NDPAVFKDNP TEDVEYQCVA DNCHSHAKML SEVLRVKVIA PVDEVQISIL SSKVVESGED IVLQCAVNEG SGPITYKFYR EKEGKPFYQM TSNATQAFWT KQKANKEQEG EYYCTAFNRA NHASSVPRSK ILTVRVILAP WKKGLIAVVI IGVIIALLII AAKCYFLRKA KAKQMPVEMS RPAVPLLNSN NEKMSDPNME ANSHYGHNDD VGNHAMKPIN DNKEPLNSDV QYTEVQVSSA ESHKDLGKKD TETVYSEVRK AVPDAVESRY SRTEGSLDGT

Target Information

CD31 (platelet endothelial cell adhesion molecule-1, PECAM-1) is an inhibitory coreceptor involved in regulation of T cell and B cell signaling by a dual immunoreceptor tyrosine-based inhibitory motif (ITIM) that upon associated kinases-mediated phosphorylation provide docking sites for protein-tyrosine phosphatases. CD31 is expressed ubiquitously within the vascular compartment and is located mainly at junctions between adjacent cells. N-terminal Ig-like domain of CD31 is responsible for its homophilic binding, which plays an important role in cell-cell interactions. CD31 is a multifunctional molecule with diverse roles in modulation of integrin-mediated cell adhesion, transendothelial migration, angiogenesis, apoptosis, negative regulation of immunoreceptor signaling, autoimmunity, macrophage phagocytosis, IgE-mediated anaphylaxis and thrombosis. It is one of key regulatory molecules in vascular system.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

References (0)

Have you cited this product in a publication?
Let us know so we can reference it here.
Cite this product

Bioinformatics

Protein Aliases: CD31; CD31 antigen; EndoCAM; GPIIA'; PECA1; PECAM-1; Platelet endothelial cell adhesion molecule; platelet/endothelial cell adhesion molecule 1

View more View less

Gene Aliases: CD31; CD31/EndoCAM; endoCAM; GPIIA'; PECA1; PECAM-1; PECAM1

View more View less

UniProt ID: (Human) P16284

View more View less

Entrez Gene ID: (Human) 5175

View more View less

Function(s)
protein binding
Process(es)
platelet degranulation phagocytosis cell adhesion signal transduction cell recognition extracellular matrix organization leukocyte migration diapedesis glomerular endothelium development
It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0