Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Primary Antibodies ›
  • PDSS2 Antibodies

OriGene

PDSS2 Monoclonal Antibody (UMAB177), UltraMAB™

View all (32) PDSS2 antibodies

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Protocols
Questions & Answers
Datasheet
Protocols
Questions & Answers

Cite PDSS2 Monoclonal Antibody (UMAB177), UltraMAB™

  • Antibody Testing Data (10)
PDSS2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
Group 53 Created with Sketch.
PDSS2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
Group 53 Created with Sketch.

FIGURE: 1 / 10

{{ $ctrl.currentElement.advancedVerification.fullName }}
{{ $ctrl.currentElement.advancedVerification.text }}

PDSS2 Antibody (UM500108CF) in IHC (P)

Immunohistochemical staining of paraffin-embedded Adenocarcinoma of Human colon tissue using anti-PDSS2 mouse monoclonal antibody. (UM500108, heat-induced epitope retrieval by 1 mM EDTA in 10mM Tris, pH8.0, 120°C for 3min) {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
Published figure supplied by benchsci-logo
PMID: {{$ctrl.currentElement.benchSciPubmedId}}
View Product

{{$ctrl.videoDesc}}

{{$ctrl.videoLongDesc}}

PDSS2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
PDSS2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
PDSS2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
PDSS2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
PDSS2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
PDSS2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
PDSS2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
PDSS2 Antibody in Western Blot (WB)
PDSS2 Antibody in Western Blot (WB)
PDSS2 Antibody in Peptide array (ARRAY)
PDSS2 Monoclonal Antibody (UMAB177), UltraMAB™

Product Details

UM500108CF

Applications
Tested Dilution
Publications

Western Blot (WB)

Assay-dependent
-

Immunohistochemistry (Paraffin) (IHC (P))

1:100-1:200
-

ChIP assay (ChIP)

Assay-Dependent
-

Peptide Array (Array)

Assay-Dependent
-
Product Specifications

Species Reactivity

Human

Host/Isotype

Mouse / IgG1

Class

Monoclonal

Type

Antibody

Clone

UMAB177

Immunogen

Full length human recombinant protein of human PDSS2 produced in HEK293T cell.
View immunogen

Conjugate

Unconjugated Unconjugated Unconjugated

Form

Lyophilized

Concentration

1 mg/mL

Purification

Affinity chromatography

Storage buffer

PBS with 8% trehalose

Contains

no preservative

Storage conditions

-20°C, Avoid Freeze/Thaw Cycles

Shipping conditions

Ambient (domestic); Wet ice (international)

Product Specific Information

For reconstitution, we recommend adding 100 µL distilled water to a final antibody concentration of about 1 mg/mL. To use this carrier-free antibody for conjugation experiments, we strongly recommend performing another round of desalting. (Zeba Spin Desalting Columns, 7KMWCO, 0.5 mL, Product # 89882)

Target Information

The isoprenoid chain of ubiquinone (coenzyme Q) varies in length between species and is determined by trans-polyprenyl diphosphate synthase. Humans possess a heterotetrameric decaprenyl diphosphate synthase composed of DPS1 and DLP1 (PDSS2) that produces Q10 ubiquinone.The isoprenoid chain of ubiquinone (coenzyme Q) varies in length between species and is determined by trans-polyprenyl diphosphate synthase. Humans possess a heterotetrameric decaprenyl diphosphate synthase composed of DPS1 and DLP1 (PDSS2) that produces Q10 ubiquinone.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

References (0)

Have you cited this product in a publication?
Let us know so we can reference it here.
Cite this product

Bioinformatics

Protein Aliases: All trans-polyprenyl-diphosphate synthase PDSS2; All-trans-decaprenyl-diphosphate synthase subunit 2; Candidate tumor suppressor protein; Decaprenyl pyrophosphate synthase subunit 2; decaprenyl pyrophosphate synthetase subunit 2; Decaprenyl-diphosphate synthase subunit 2; RP11-59I9.3; Solanesyl-diphosphate synthase subunit 2; subunit 2 of decaprenyl diphosphate synthase

View more View less

Gene Aliases: bA59I9.3; C6orf210; COQ10D3; DLP1; hDLP1; PDSS2

View more View less

UniProt ID: (Human) Q86YH6

View more View less

Entrez Gene ID: (Human) 57107

View more View less

Function(s)
trans-hexaprenyltranstransferase activity protein heterodimerization activity trans-octaprenyltranstransferase activity
Process(es)
ubiquinone biosynthetic process isoprenoid biosynthetic process regulation of body fluid levels protein heterotetramerization
It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-9dd846b7f-s9zn4:80/100.66.128.142:80.
git-commit: 17e8f6c92ed407c129a469238c057b56fdf8abbb
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0