Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Primary Antibodies ›
  • PDLIM2 Antibodies

OriGene

PDLIM2 Monoclonal Antibody (OTI9A11), TrueMAB™

View all (28) PDLIM2 antibodies

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Protocols
Questions & Answers
Datasheet
Protocols
Questions & Answers

Cite PDLIM2 Monoclonal Antibody (OTI9A11), TrueMAB™

  • Antibody Testing Data (5)
PDLIM2 Antibody in Immunocytochemistry (ICC/IF)
Group 53 Created with Sketch.
PDLIM2 Antibody in Immunocytochemistry (ICC/IF)
Group 53 Created with Sketch.

FIGURE: 1 / 5

{{ $ctrl.currentElement.advancedVerification.fullName }}
{{ $ctrl.currentElement.advancedVerification.text }}

PDLIM2 Antibody (TA502751) in ICC/IF

Anti-PDLIM2 mouse monoclonal antibody (TA502751) immunofluorescent staining of COS7 cells transiently transfected by pCMV6-ENTRY PDLIM2(RC210022). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
Published figure supplied by benchsci-logo
PMID: {{$ctrl.currentElement.benchSciPubmedId}}
View Product

{{$ctrl.videoDesc}}

{{$ctrl.videoLongDesc}}

PDLIM2 Antibody in Immunocytochemistry (ICC/IF)
PDLIM2 Antibody in Western Blot (WB)
PDLIM2 Antibody in Flow Cytometry (Flow)
PDLIM2 Antibody in Flow Cytometry (Flow)
PDLIM2 Antibody in Flow Cytometry (Flow)
PDLIM2 Monoclonal Antibody (OTI9A11), TrueMAB™

Product Details

TA502751

Applications
Tested Dilution
Publications

Western Blot (WB)

1:2,000
-

Immunocytochemistry (ICC/IF)

1:100
-

Flow Cytometry (Flow)

1:100
-
Product Specifications

Species Reactivity

Human

Host/Isotype

Mouse / IgG1

Class

Monoclonal

Type

Antibody

Clone

OTI9A11

Immunogen

Full length human recombinant protein of human PDLIM2 produced in HEK293T cell.
View immunogen

Conjugate

Unconjugated Unconjugated Unconjugated

Form

Liquid

Concentration

0.49 mg/mL

Purification

Affinity chromatography

Storage buffer

PBS with 1% BSA, 50% glycerol

Contains

0.02% sodium azide

Storage conditions

-20°C, Avoid Freeze/Thaw Cycles

Shipping conditions

Ambient (domestic); Wet ice (international)

Target Information

Probable adapter protein located at the actin cytoskeleton that promotes cell attachment. Necessary for the migratory capacity of epithelial cells. Overexpression enhances cell adhesion to collagen and fibronectin and suppresses anchorage independent growth. May contribute to tumor cell migratory capacity.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

References (0)

Have you cited this product in a publication?
Let us know so we can reference it here.
Cite this product

Bioinformatics

Protein Aliases: PDZ and LIM domain 2 (mystique); PDZ and LIM domain protein 2; PDZ-LIM protein mystique

View more View less

Gene Aliases: MYSTIQUE; PDLIM2; PP6345; SLIM

View more View less

UniProt ID: (Human) Q96JY6

View more View less

Entrez Gene ID: (Human) 64236

View more View less

Function(s)
protein binding zinc ion binding filamin binding myosin heavy chain binding muscle alpha-actinin binding
It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-55c564d944-rwwh4:80/100.66.128.142:80.
git-commit: 3092084cb0e494f0e07f07633350183de7fa10a8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0