Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Primary Antibodies ›
  • PAN2 Antibodies

Invitrogen

PAN2 Polyclonal Antibody

View all (5) PAN2 antibodies

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Protocols
Questions & Answers
Datasheet
Protocols
Questions & Answers

Cite PAN2 Polyclonal Antibody

  • Antibody Testing Data (1)
PAN2 Antibody in Western Blot (WB)
Group 53 Created with Sketch.
PAN2 Antibody in Western Blot (WB)
Group 53 Created with Sketch.

FIGURE: 1 / 1

{{ $ctrl.currentElement.advancedVerification.fullName }}
{{ $ctrl.currentElement.advancedVerification.text }}

PAN2 Antibody (PA5-78303) in WB

PAN2 Polyclonal Antibody detects PAN2 protein by western blot analysis. A. 30 µg Jurkat whole cell lysate/extract. B. 30 µg Raji whole cell lysate/extract.5 % SDS-PAGE. PAN2 Polyclonal Antibody (Product # PA5-78303) dilution: 1:500. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
Published figure supplied by benchsci-logo
PMID: {{$ctrl.currentElement.benchSciPubmedId}}
View Product

{{$ctrl.videoDesc}}

{{$ctrl.videoLongDesc}}

PAN2 Antibody in Western Blot (WB)
PAN2 Polyclonal Antibody

Product Details

PA5-78303

Applications
Tested Dilution
Publications

Western Blot (WB)

1:500-1:3,000
-
Product Specifications

Species Reactivity

Human

Host/Isotype

Rabbit / IgG

Class

Polyclonal

Type

Antibody

Immunogen

Recombinant protein encompassing a sequence within the N-terminus region of human PAN2. The exact sequence is proprietary.
View immunogen

Conjugate

Unconjugated Unconjugated Unconjugated

Form

Liquid

Concentration

0.41 mg/mL

Purification

Antigen affinity chromatography

Storage buffer

PBS, pH 7, with 20% glycerol

Contains

0.01% thimerosal

Storage conditions

Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.

Shipping conditions

Ambient (domestic); Wet ice (international)

RRID

AB_2736319

Product Specific Information

Positive Control: Jurkat, Raji

Predicted Reactivity: Mouse (97%), Rat (97%), Xenopus laevis (81%), Chicken (87%), Bovine (97%)

Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Target Information

This gene encodes a coregulator for the alpha and beta estrogen receptors and the orphan nuclear receptor NR1D2. The protein localizes to the nucleus, and is thought to have both coactivator and corepressor functions. Its interaction with nuclear receptors is independent of the AF2 domain on the receptors, which is known to regulate interaction with other coreceptors. Two alternatively spliced transcript variants for this gene have been described. However, the full length nature of one of the variants has not been determined.

⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

References (0)

Have you cited this product in a publication?
Let us know so we can reference it here.
Cite this product

Bioinformatics

Protein Aliases: Inactive ubiquitin carboxyl-terminal hydrolase 52; PAB-dependent poly(A)-specific ribonuclease subunit 2; PAB1P-dependent poly(A)-nuclease; PAB1P-dependent poly(A)-specific ribonuclease; PABP-dependent poly(A) nuclease 2; PAN deadenylation complex catalytic subunit 2; PAN deadenylation complex subunit 2; PAN2 poly(A) specific ribonuclease subunit homolog; PAN2 polyA specific ribonuclease subunit; PAN2-PAN3 deadenylation complex catalytic subunit PAN2; Poly(A)-nuclease deadenylation complex subunit 2; ubiquitin specific peptidase 52; ubiquitin specific protease 52

View more View less

Gene Aliases: KIAA0710; PAN2; USP52

View more View less

UniProt ID: (Human) Q504Q3

View more View less

Entrez Gene ID: (Human) 9924

View more View less

Function(s)
3'-5'-exoribonuclease activity nucleic acid binding poly(A)-specific ribonuclease activity protein binding
Process(es)
nuclear-transcribed mRNA poly(A) tail shortening mRNA processing positive regulation of cytoplasmic mRNA processing body assembly RNA phosphodiester bond hydrolysis, exonucleolytic
It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-657bd577ff-sg76p:80/100.66.128.142:80.
git-commit: cdba60da2f3289684898c31dc50804c008488b19
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0