Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Primary Antibodies ›
  • P2Y1 Antibodies

Proteintech

P2RY1 Monoclonal Antibody (1A1F3)

2 References
View all (24) P2Y1 antibodies

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Protocols
Questions & Answers
Datasheet
Protocols
Questions & Answers

Cite P2RY1 Monoclonal Antibody (1A1F3)

  • Antibody Testing Data (5)
P2RY1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
Group 53 Created with Sketch.
P2RY1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
Group 53 Created with Sketch.

FIGURE: 1 / 5

{{ $ctrl.currentElement.advancedVerification.fullName }}
{{ $ctrl.currentElement.advancedVerification.text }}

P2RY1 Antibody (67654-1-IG) in IHC (P)

Immunohistochemical analysis of paraffin-embedded mouse brain tissue slide using 67654-1-Ig (P2RY1 antibody) at dilution of 1:1000 (under 40x lens). Heat mediated antigen retrieval with Tris-EDTA buffer (pH 9.0). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
Published figure supplied by benchsci-logo
PMID: {{$ctrl.currentElement.benchSciPubmedId}}
View Product

{{$ctrl.videoDesc}}

{{$ctrl.videoLongDesc}}

P2RY1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
P2RY1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
P2RY1 Antibody in Immunohistochemistry (PFA fixed) (IHC (PFA))
P2RY1 Antibody in Immunohistochemistry (PFA fixed) (IHC (PFA))
P2RY1 Antibody in Western Blot (WB)

Product Details

67654-1-IG

Applications
Tested Dilution
Publications

Western Blot (WB)

1:10,000
View 1 publication 1 publication

Immunohistochemistry (IHC)

-
View 1 publication 1 publication

Immunohistochemistry (Paraffin) (IHC (P))

1:1,000
-

Immunohistochemistry (PFA fixed) (IHC (PFA))

1:1,000
-
Product Specifications

Species Reactivity

Human, Mouse, Pig, Rabbit, Rat

Host/Isotype

Mouse / IgG1

Class

Monoclonal

Type

Antibody

Clone

1A1F3

Immunogen

P2RY1 Fusion Protein Ag12762

Conjugate

Unconjugated Unconjugated Unconjugated
  • CoraLite Plus 488
  • CoraLite Plus 647
  • Request custom conjugation

Form

Liquid

Concentration

500 µg/mL

Purification

Protein G

Storage buffer

PBS, pH 7.3, with 50% glycerol

Contains

0.02% sodium azide

Storage conditions

-20°C

Shipping conditions

Wet ice

Product Specific Information

Aliquoting is unnecessary for -20°C storage.

Target Information

P2Y1 is a Purinergic Receptor that causes a change in platelet shape and potentially leads to platelet aggregation by binding ADP, which results in the mobilization of intracellular calcium ions via activation of phospholipase C. This receptor also mediates the release of the relaxing factor nitric oxide and may be involved in the modulation of neuronal signal transmission. P2Y1 has been reported to be expressed in tissues throughout the body, particularly brain. ESTs have been isolated from human embryo, lung, and placenta libraries.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

Bioinformatics

Protein Aliases: ADP receptor; ATP receptor; P2 purinoceptor subtype Y1; P2 purinoreceptor subclass 2Y; P2Y ATP receptor 1; P2Y purinoceptor 1; p2y-1-r; P2Y1; P2Y1 receptor; p2y1r; platelet ADP receptor; Purinergic receptor; purinergic receptor P2Y, G-protein coupled, 1; purinergic receptor P2Y1; Purinoceptor p2y1; Purinoceptor p2yr1

View more View less

Gene Aliases: P2RY1; P2y; P2Y1; P2Y1R

View more View less

UniProt ID: (Human) P47900, (Rat) P49651, (Mouse) P49650

View more View less

Entrez Gene ID: (Pig) 503666, (Human) 5028, (Rabbit) 100353792, (Rat) 25265, (Mouse) 18441

View more View less

Function(s)
receptor activity protein binding ATP binding A1 adenosine receptor binding ADP binding G-protein coupled purinergic nucleotide receptor activity ATP-activated adenosine receptor activity ADP-activated adenosine receptor activity protein heterodimerization activity scaffold protein binding signal transducer activity G-protein coupled receptor activity
Process(es)
positive regulation of protein phosphorylation adenosine receptor signaling pathway G-protein coupled receptor signaling pathway adenylate cyclase-inhibiting G-protein coupled receptor signaling pathway positive regulation of cytosolic calcium ion concentration aging glial cell migration response to mechanical stimulus regulation of receptor activity negative regulation of norepinephrine secretion sensory perception of pain signal transduction involved in regulation of gene expression platelet activation positive regulation of inositol trisphosphate biosynthetic process G-protein coupled purinergic nucleotide receptor signaling pathway wound healing regulation of vasodilation eating behavior positive regulation of ion transport positive regulation of transcription from RNA polymerase II promoter positive regulation of hormone secretion negative regulation of binding positive regulation of cytosolic calcium ion concentration involved in phospholipase C-activating G-protein coupled signaling pathway positive regulation of penile erection positive regulation of ERK1 and ERK2 cascade response to growth factor cellular response to organic cyclic compound protein localization to plasma membrane relaxation of muscle cell surface receptor signaling pathway phospholipase C-activating G-protein coupled receptor signaling pathway signal transduction
It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0