Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Primary Antibodies ›
  • OGDH Antibodies

Proteintech

OGDH Monoclonal Antibody (1H7A2), CoraLite® Plus 488

View all (11) OGDH antibodies

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Protocols
Questions & Answers
Datasheet
Protocols
Questions & Answers

Cite OGDH Monoclonal Antibody (1H7A2), CoraLite® Plus 488

  • Antibody Testing Data (1)
OGDH Antibody in Immunocytochemistry (ICC/IF)
Group 53 Created with Sketch.
OGDH Antibody in Immunocytochemistry (ICC/IF)
Group 53 Created with Sketch.

FIGURE: 1 / 1

{{ $ctrl.currentElement.advancedVerification.fullName }}
{{ $ctrl.currentElement.advancedVerification.text }}

OGDH Antibody (CL488-66285) in ICC/IF

Immunofluorescent analysis of (4% PFA) fixed HepG2 cells using CL488-66285 (OGDH antibody) at dilution of 1:100 {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
Published figure supplied by benchsci-logo
PMID: {{$ctrl.currentElement.benchSciPubmedId}}
View Product

{{$ctrl.videoDesc}}

{{$ctrl.videoLongDesc}}

OGDH Antibody in Immunocytochemistry (ICC/IF)

Product Details

CL488-66285

Applications
Tested Dilution
Publications

Immunocytochemistry (ICC/IF)

1:50-1:500
-
Product Specifications

Species Reactivity

Human, Rat

Host/Isotype

Mouse / IgG2b

Class

Monoclonal

Type

Antibody

Clone

1H7A2

Immunogen

Recombinant Protein

Conjugate

CoraLite® Plus 488 CoraLite® Plus 488 CoraLite® Plus 488
  • Unconjugated
  • CoraLite 594
  • Request custom conjugation

Excitation/Emission Max

493/522 nm

Form

Liquid

Concentration

2 mg/mL

Purification

Protein A

Storage buffer

PBS, pH 7.3, with 0.5% BSA, 50% glycerol

Contains

0.05% ProClin 300

Storage conditions

-20°C, store in dark

Shipping conditions

Wet ice

Target Information

This gene encodes one subunit of the 2-oxoglutarate dehydrogenase complex. This complex catalyzes the overall conversion of 2-oxoglutarate (alpha-ketoglutarate) to succinyl-CoA and CO(2) during the Krebs cycle. The protein is located in the mitocondrial matrix and uses thiamine pyrophosphate as a cofactor. A congential deficiency in 2-oxoglutarate dehydrogenase activity is believed to lead to hypotonia, metabolic acidosis, and hyperlactatemia.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

References (0)

Have you cited this product in a publication?
Let us know so we can reference it here.
Cite this product

Bioinformatics

Protein Aliases: 2-oxoglutarate dehydrogenase complex component E1; 2-oxoglutarate dehydrogenase E1 component, mitochondrial; 2-oxoglutarate dehydrogenase, mitochondrial; Alpha-ketoglutarate dehydrogenase; Alpha-KGDH-E1; E1o; OGDC-E1; oxoglutarate (alpha-ketoglutarate) dehydrogenase (lipoamide); oxoglutarate decarboxylase; oxoglutarate dehydrogenase (lipoamide); oxoglutarate dehydrogenase (succinyl-transferring); testicular tissue protein Li 131; Thiamine diphosphate (ThDP)-dependent 2-oxoglutarate dehydrogenase

View more View less

Gene Aliases: AKGDH; E1k; OGDC; OGDH; Ogdhl

View more View less

UniProt ID: (Human) Q02218, (Rat) Q5XI78

View more View less

Entrez Gene ID: (Human) 4967, (Rat) 360975

View more View less

Function(s)
oxoglutarate dehydrogenase (succinyl-transferring) activity thiamine pyrophosphate binding heat shock protein binding oxoglutarate dehydrogenase (NAD+) activity metal ion binding chaperone binding
Process(es)
generation of precursor metabolites and energy glycolytic process tricarboxylic acid cycle 2-oxoglutarate metabolic process succinyl-CoA metabolic process lysine catabolic process NADH metabolic process cerebellar cortex development striatum development hippocampus development thalamus development pyramidal neuron development tangential migration from the subventricular zone to the olfactory bulb glyoxylate metabolic process olfactory bulb mitral cell layer development oxidation-reduction process
It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-c7d89468c-sbmwt:80/100.66.128.142:80.
git-commit: 4db89d68e0ee702f860d53b64def1eb94897fade
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0