Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Primary Antibodies ›
  • Nucleolin Antibodies

NeoBiotechnologies

Nucleolin (Marker of Human Cells) Monoclonal Antibody (SPM614)

View all (50) Nucleolin antibodies

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Protocols
Questions & Answers
Datasheet
Protocols
Questions & Answers

Cite Nucleolin (Marker of Human Cells) Monoclonal Antibody (SPM614)

  • Antibody Testing Data (5)
Nucleolin (Marker of Human Cells) Antibody in Immunocytochemistry (ICC/IF)
Group 53 Created with Sketch.
Nucleolin (Marker of Human Cells) Antibody in Immunocytochemistry (ICC/IF)
Group 53 Created with Sketch.

FIGURE: 1 / 5

{{ $ctrl.currentElement.advancedVerification.fullName }}
{{ $ctrl.currentElement.advancedVerification.text }}

Nucleolin (Marker of Human Cells) Antibody (4691-MSM1X-P1ABX) in ICC/IF

Immunofluorescence analysis of PFA-fixed K562 cells stained with Nucleolin Mouse Monoclonal Antibody (SPM614) followed by goat anti-mouse IgG-CF488 (green). Membrane stained with phalloidin-CF640. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
Published figure supplied by benchsci-logo
PMID: {{$ctrl.currentElement.benchSciPubmedId}}
View Product

{{$ctrl.videoDesc}}

{{$ctrl.videoLongDesc}}

Nucleolin (Marker of Human Cells) Antibody in Immunocytochemistry (ICC/IF)
Nucleolin (Marker of Human Cells) Antibody in Immunocytochemistry (ICC/IF)
Nucleolin (Marker of Human Cells) Antibody in Immunocytochemistry (ICC/IF)
Nucleolin (Marker of Human Cells) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
Nucleolin (Marker of Human Cells) Antibody in Flow Cytometry (Flow)
Nucleolin (Marker of Human Cells) Monoclonal Antibody (SPM614)

Product Details

4691-MSM1X-P1ABX

Applications
Tested Dilution
Publications

Western Blot (WB)

1-2 µg/mL
-

Immunohistochemistry (Paraffin) (IHC (P))

Assay-dependent
-

Immunohistochemistry (PFA fixed) (IHC (PFA))

1-2 µg/mL
-

Immunocytochemistry (ICC/IF)

1-4 µg/mL
-

Flow Cytometry (Flow)

0.5-1 µg/10^6 cells
-
Product Specifications

Species Reactivity

Human

Host/Isotype

Mouse / IgG1, kappa

Class

Monoclonal

Type

Antibody

Clone

SPM614

Immunogen

Recombinant full-length human NCL protein
View immunogen

Conjugate

Unconjugated Unconjugated Unconjugated

Form

Liquid

Concentration

1 mg/mL

Purification

Protein A/G

Storage buffer

PBS, pH 7.4

Contains

no preservative

Storage conditions

-20°C, Avoid Freeze/Thaw Cycles

Shipping conditions

Wet ice

Product Specific Information

Does not react with Mouse, Rat or Cow

Target Information

Nucleolin (NCL), a eukaryotic nucleolar phosphoprotein, is involved in the synthesis and maturation of ribosomes. It is located mainly in dense fibrillar regions of the nucleolus. Human NCL gene consists of 14 exons with 13 introns and spans approximately 11kb. The intron 11 of the NCL gene encodes a small nucleolar RNA, termed U20.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

References (0)

Have you cited this product in a publication?
Let us know so we can reference it here.
Cite this product

Bioinformatics

Protein Aliases: FLJ45706; FLJ59041; nucl antibody; Nucleolin; Protein C23

View more View less

Gene Aliases: C23; NCL

View more View less

UniProt ID: (Human) P19338

View more View less

Entrez Gene ID: (Human) 4691

View more View less

Function(s)
nucleotide binding single-stranded DNA binding RNA binding calcium ion binding protein binding protein C-terminus binding selenocysteine insertion sequence binding rRNA primary transcript binding telomeric DNA binding histone binding identical protein binding laminin binding poly(A) RNA binding ErbB-4 class receptor binding
Process(es)
angiogenesis endocytosis spermatogenesis positive regulation of tumor necrosis factor production negative regulation of apoptotic process positive regulation of transcription from RNA polymerase II promoter cellular response to lipopolysaccharide liver regeneration positive regulation of transcription of nuclear large rRNA transcript from RNA polymerase I promoter regulation of rRNA processing positive regulation of interleukin-6 secretion
It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0