Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Primary Antibodies ›
  • Nidogen Antibodies

NeoBiotechnologies

Nidogen/Entactin Monoclonal Antibody (ELM1)

View all (16) Nidogen antibodies

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Protocols
Questions & Answers
Datasheet
Protocols
Questions & Answers

Cite Nidogen/Entactin Monoclonal Antibody (ELM1)

  • Antibody Testing Data (1)
Nidogen/Entactin Antibody in SDS-PAGE (SDS-PAGE)
Group 53 Created with Sketch.
Nidogen/Entactin Antibody in SDS-PAGE (SDS-PAGE)
Group 53 Created with Sketch.

FIGURE: 1 / 1

{{ $ctrl.currentElement.advancedVerification.fullName }}
{{ $ctrl.currentElement.advancedVerification.text }}

Nidogen/Entactin Antibody (MS18073-RTM1-P0) in SDS-PAGE

SDS-PAGE analysis Purified Nidogen Mouse Monoclonal Antibody (ELM1). Confirmation of Purity and Integrity of Antibody. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
Published figure supplied by benchsci-logo
PMID: {{$ctrl.currentElement.benchSciPubmedId}}
View Product

{{$ctrl.videoDesc}}

{{$ctrl.videoLongDesc}}

Nidogen/Entactin Antibody in SDS-PAGE (SDS-PAGE)
Nidogen/Entactin Monoclonal Antibody (ELM1)

Product Details

MS18073-RTM1-P0

Applications
Tested Dilution
Publications

ELISA (ELISA)

2-4 µg/mL
-
Product Specifications

Species Reactivity

Guinea pig, Mouse

Host/Isotype

Rat / IgG2a

Class

Monoclonal

Type

Antibody

Clone

ELM1

Immunogen

Murine EHS laminin preparation
View immunogen

Conjugate

Unconjugated Unconjugated Unconjugated

Form

Liquid

Concentration

200 µg/mL

Purification

Protein A/G

Storage buffer

PBS, pH 7.4, with 0.05% BSA

Contains

0.05% sodium azide

Storage conditions

4°C, do not freeze

Shipping conditions

Ambient (domestic); Wet ice (international)

Product Specific Information

Does not react with human

Target Information

This gene encodes a member of the nidogen family of basement membrane glycoproteins. The protein interacts with several other components of basement membranes, and may play a role in cell interactions with the extracellular matrix.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

References (0)

Have you cited this product in a publication?
Let us know so we can reference it here.
Cite this product

Bioinformatics

Protein Aliases: Entactin; entactin 1; NID 1; NID-1; Nidogen (enactin); Nidogen precursor (Entactin); Nidogen-1; Nidogen-1;

View more View less

Gene Aliases: A630025O17; Ent; entactin; entactin-1; Nid; Nid1; nidogen-1

View more View less

UniProt ID: (Mouse) P10493

View more View less

Entrez Gene ID: (Mouse) 18073

View more View less

Function(s)
calcium ion binding protein binding collagen binding laminin binding laminin-1 binding proteoglycan binding extracellular matrix binding
Process(es)
cell adhesion cell-matrix adhesion positive regulation of cell-substrate adhesion extracellular matrix organization glomerular basement membrane development
It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0