Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Primary Antibodies ›
  • GPI Antibodies

Osenses

Neuroleukin Polyclonal Antibody

2 References
View all (44) GPI antibodies

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Protocols
Questions & Answers
Datasheet
Protocols
Questions & Answers

Cite Neuroleukin Polyclonal Antibody

Neuroleukin Polyclonal Antibody

Product Details

OSG00070W-100UL

Applications
Tested Dilution
Publications

Western Blot (WB)

1:300-1:2,000
-

Immunohistochemistry (IHC)

1:300-1:2,000
-

Miscellaneous PubMed (MISC)

-
View 2 publications 2 publications
Product Specifications

Species Reactivity

Human, Mouse, Rat

Host/Isotype

Rabbit / Ig

Class

Polyclonal

Type

Antibody

Immunogen

A synthetic peptide from c-terminal region of mouse Neuroleukin (GPI) conjugated to an immunogenic carrier protein was used as the antigen
View immunogen

Conjugate

Unconjugated Unconjugated Unconjugated

Form

Lyophilized

Concentration

Conc. Not Determined

Storage buffer

whole serum

Contains

no preservative

Storage conditions

Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. Glycerol (1:1) may be added for added stability.

Shipping conditions

Ambient (domestic); Wet ice (international)

Product Specific Information

Reconstitute in 100 µL of sterile water. Centrifuge to remove any insoluble material.

Specificity of this antibody: Neuroleukin.

Target Information

This gene belongs to the GPI family whose members encode multifunctional phosphoglucose isomerase proteins involved in energy pathways. The protein encoded by this gene is a dimeric enzyme that catalyzes the reversible isomerization of glucose-6-phosphate and fructose-6-phosphate. The protein functions in different capacities inside and outside the cell. In the cytoplasm, the gene product is involved in glycolysis and gluconeogenesis, while outside the cell it functions as a neurotrophic factor for spinal and sensory neurons. Defects in this gene are the cause of nonspherocytic hemolytic anemia and a severe enzyme deficiency can be associated with hydrops fetalis, immediate neonatal death and neurological impairment.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

Bioinformatics

Protein Aliases: AMF; Autocrine motility factor; glucose phosphate isomerase; glucose phosphate isomerase 1 complex; glucose phosphate isomerase 1, regulatory; glucose phosphate isomerase 1, structural; glucose phosphate isomerase 1, temporal; Glucose-6-phosphate isomerase; glucose-6-phosphate isomerase {ECO:0000250|UniProtKB:P06745}; GPI; GPI {ECO:0000250|UniProtKB:P06745}; gpi {ECO:0000312|RGD:2727}; hexose monophosphate isomerase; hexosephosphate isomerase; maturation factor; Neuroleukin; neuroleukin {ECO:0000250|UniProtKB:P06745}; NLK; NLK {ECO:0000250|UniProtKB:P06745}; oxoisomerase; PGI; PGI {ECO:0000250|UniProtKB:P06745}; PHI; PHI {ECO:0000250|UniProtKB:P06745}; Phosphoglucose isomerase; phosphoglucose isomerase {ECO:0000250|UniProtKB:P06745}; phosphohexomutase; Phosphohexose isomerase; phosphohexose isomerase {ECO:0000250|UniProtKB:P06745}; phosphosaccharomutase; SA-36; sperm antigen; Sperm antigen 36; sperm antigen-36

View more View less

Gene Aliases: AMF; GNPI; GPI; Gpi-1; Gpi-1r; Gpi-1s; Gpi-1t; Gpi1; Gpi1-r; Gpi1-s; Gpi1-t; Gpi1s; MF; NK; NK/GPI; NLK; Org; PGI; PHI; SA-36; SA36

View more View less

UniProt ID: (Human) P06744, (Rat) Q6P6V0, (Mouse) P06745

View more View less

Entrez Gene ID: (Human) 2821, (Rat) 292804, (Mouse) 14751

View more View less

Function(s)
glucose-6-phosphate isomerase activity cytokine activity growth factor activity intramolecular transferase activity ubiquitin protein ligase binding monosaccharide binding protein binding isomerase activity
Process(es)
angiogenesis in utero embryonic development mesoderm formation carbohydrate metabolic process gluconeogenesis glycolytic process methylglyoxal biosynthetic process erythrocyte homeostasis glucose homeostasis negative regulation of cysteine-type endopeptidase activity involved in apoptotic process negative regulation of neuron apoptotic process aldehyde catabolic process glucose 6-phosphate metabolic process glycolytic process through glucose-6-phosphate humoral immune response hemostasis learning or memory canonical glycolysis
It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-9dd846b7f-s9zn4:80/100.66.128.142:80.
git-commit: 17e8f6c92ed407c129a469238c057b56fdf8abbb
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0