Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Primary Antibodies ›
  • NUDT15 Antibodies

OriGene

NUDT15 Monoclonal Antibody (OTI7H2), TrueMAB™

View all (12) NUDT15 antibodies

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Protocols
Questions & Answers
Datasheet
Protocols
Questions & Answers

Cite NUDT15 Monoclonal Antibody (OTI7H2), TrueMAB™

  • Antibody Testing Data (1)
NUDT15 Antibody in Western Blot (WB)
Group 53 Created with Sketch.
NUDT15 Antibody in Western Blot (WB)
Group 53 Created with Sketch.

FIGURE: 1 / 1

{{ $ctrl.currentElement.advancedVerification.fullName }}
{{ $ctrl.currentElement.advancedVerification.text }}

NUDT15 Antibody (CF811515) in WB

HEK293T cells were transfected with the pCMV6-ENTRY control (Left lane) or pCMV6-ENTRY NUDT15 (RC210477, Right lane) cDNA for 48 hrs and lysed. Equivalent amounts of cell lysates (5 µg per lane) were separated by SDS-PAGE and immunoblotted with anti-NUDT1. Positive lysates LY402670 (100 µg) and LC402670 (20 µg) can be purchased separately from OriGene. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
Published figure supplied by benchsci-logo
PMID: {{$ctrl.currentElement.benchSciPubmedId}}
View Product

{{$ctrl.videoDesc}}

{{$ctrl.videoLongDesc}}

NUDT15 Antibody in Western Blot (WB)
NUDT15 Monoclonal Antibody (OTI7H2), TrueMAB™

Product Details

CF811515

Applications
Tested Dilution
Publications

Western Blot (WB)

1:2,000
-
Product Specifications

Species Reactivity

Human, Mouse, Rat

Host/Isotype

Mouse / IgG1

Class

Monoclonal

Type

Antibody

Clone

OTI7H2

Immunogen

Full length human recombinant protein of human NUDT15 produced in HEK293T cell.
View immunogen

Conjugate

Unconjugated Unconjugated Unconjugated

Form

Lyophilized

Concentration

1 mg/mL

Purification

Affinity chromatography

Storage buffer

PBS, pH 7.3, with 8% trehalose

Contains

no preservative

Storage conditions

-20°C, Avoid Freeze/Thaw Cycles

Shipping conditions

Ambient (domestic); Wet ice (international)

Product Specific Information

For reconstitution, we recommend adding 100 µL distilled water to a final antibody concentration of about 1 mg/mL. To use this carrier-free antibody for conjugation experiments, we strongly recommend performing another round of desalting. (Zeba Spin Desalting Columns, 7KMWCO, 0.5 mL, Product # 89882)

Target Information

Mediates the hydrolysis of some nucleoside diphosphate derivatives. Can degrade 8-oxo-dGTP in vitro, suggesting that it may remove an oxidatively damaged form of guanine (7,8-dihydro-8-oxoguanine) from DNA and the nucleotide pool, thereby preventing misincorporation of 8-oxo-dGTP into DNA thus preventing A:T to C:G transversions. Its substrate specificity in vivo however remains unclear. May have a role in DNA synthesis and cell cycle progression throught the interaction with PCNA.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

References (0)

Have you cited this product in a publication?
Let us know so we can reference it here.
Cite this product

Bioinformatics

Protein Aliases: 7,8-dihydro-8-oxoguanine-triphosphatase NUDT15; 8-oxo-dGTPase NUDT15; mMTH2; MTH2; MutT homolog 2; Nucleoside diphosphate-linked moiety X motif 15; Nucleoside diphosphate-linked to another moiety X hydrolase 15; Nucleotide triphosphate diphosphatase NUDT15; nudix (nucleoside diphosphate linked moiety X)-type motif 15; Nudix hydrolase 15; Nudix motif 15; nudix-type motif 15; probable 7,8-dihydro-8-oxoguanine triphosphatase NUDT15; probable 8-oxo-dGTP diphosphatase NUDT15; RP11-90M2.1

View more View less

Gene Aliases: 6530403O17; A730068G11Rik; MTH2; NUDT15; NUDT15D

View more View less

UniProt ID: (Human) Q9NV35, (Mouse) Q8BG93

View more View less

Entrez Gene ID: (Human) 55270, (Rat) 290365, (Mouse) 214254

View more View less

Function(s)
8-oxo-7,8-dihydroguanosine triphosphate pyrophosphatase activity nucleoside-diphosphatase activity 8-oxo-7,8-dihydrodeoxyguanosine triphosphate pyrophosphatase activity metal ion binding hydrolase activity
Process(es)
dGTP catabolic process nucleobase-containing small molecule catabolic process response to reactive oxygen species purine deoxyribonucleoside triphosphate catabolic process
It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-9dd846b7f-s9zn4:80/100.66.128.142:80.
git-commit: 17e8f6c92ed407c129a469238c057b56fdf8abbb
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0