Abnova
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Product may be used with Western Blot (Cell lysate).
Immunogen sequence: PLERGAPNKE ETPATESPDT GLYYHRYLQE VIDVLETDGH FREKLQAANA EDIKSGKLSR ELDFVSHHVR TKLDELERQE VSRPRMLLKA KMDAEQDPNV QVDHLNLLKQ FEHLDPQNQH TFEARDLELL IQTATRDLAQ YDAAHHEEFK RYEMLKEHER RRYLESLGEE QRKEAERKLE EQQRRHREHP KVNVPGSQAQ LKEVWEELDG LDPNRFNPKT FFILHDINSD GVLDEQELEA LFTKELEKVY DPKNEEDDMR EMEEERLRMR EHVMKNVDTN QDRLVTLEEF LASTQRKEFG DTGEGWETVE MHPAYTEEEL RRFEEELAAR EAELNAKAQR LSQETEALGR SQGRLEAQKR ELQQAVLHME QRKQQQQQQQ GHKAPAAHPE GQLKFHPDTD DVPVPAPAGD QKEVDTSEKK LLERLPEVEV PQHL
Nucleobindin 1 encodes a member of a small calcium-binding EF-hand protein family. The encoded protein is thought to have a key role in Golgi calcium homeostasis and Ca(2+)-regulated signal transduction events.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CALNUC; Nucleobindin-1
Gene Aliases: CALNUC; NUC; NUCB1
UniProt ID: (Human) Q02818
Entrez Gene ID: (Human) 4924
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support