Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Primary Antibodies ›
  • NT5D1 Antibodies

OriGene

NT5DC1 Monoclonal Antibody (OTI2A1), TrueMAB™

View all (25) NT5D1 antibodies

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Protocols
Questions & Answers
Datasheet
Protocols
Questions & Answers

Cite NT5DC1 Monoclonal Antibody (OTI2A1), TrueMAB™

  • Antibody Testing Data (11)
NT5DC1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
Group 53 Created with Sketch.
NT5DC1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
Group 53 Created with Sketch.

FIGURE: 1 / 11

{{ $ctrl.currentElement.advancedVerification.fullName }}
{{ $ctrl.currentElement.advancedVerification.text }}

NT5DC1 Antibody (CF501591) in IHC (P)

Immunohistochemical staining of paraffin-embedded human lymphoma tissue using anti-NT5DC1 mouse monoclonal antibody. (Heat-induced epitope retrieval by 10mM citric buffer, pH6.0, 100°C for 10min, TA501591) {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
Published figure supplied by benchsci-logo
PMID: {{$ctrl.currentElement.benchSciPubmedId}}
View Product

{{$ctrl.videoDesc}}

{{$ctrl.videoLongDesc}}

NT5DC1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
NT5DC1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
NT5DC1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
NT5DC1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
NT5DC1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
NT5DC1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
NT5DC1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
NT5DC1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
NT5DC1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
NT5DC1 Antibody in Western Blot (WB)
NT5DC1 Antibody in Flow Cytometry (Flow)
NT5DC1 Monoclonal Antibody (OTI2A1), TrueMAB™

Product Details

CF501591

Applications
Tested Dilution
Publications

Western Blot (WB)

1:2,000
-

Immunohistochemistry (Paraffin) (IHC (P))

1:150
-

Flow Cytometry (Flow)

1:100
-
Product Specifications

Species Reactivity

Human

Host/Isotype

Mouse / IgG1

Class

Monoclonal

Type

Antibody

Clone

OTI2A1

Immunogen

Full length human recombinant protein of human NT5DC1 produced in HEK293T cell.
View immunogen

Conjugate

Unconjugated Unconjugated Unconjugated

Form

Lyophilized

Concentration

1 mg/mL

Purification

Affinity chromatography

Storage buffer

PBS, pH 7.3, with 8% trehalose

Contains

no preservative

Storage conditions

-20°C, Avoid Freeze/Thaw Cycles

Shipping conditions

Ambient (domestic); Wet ice (international)

Product Specific Information

For reconstitution, we recommend adding 100 µL distilled water to a final antibody concentration of about 1 mg/mL. To use this carrier-free antibody for conjugation experiments, we strongly recommend performing another round of desalting. (Zeba Spin Desalting Columns, 7KMWCO, 0.5 mL, Product # 89882)

Target Information

While the exact function of the protein encoded by this gene is not known, it belongs to the 5'(3')-deoxyribonucleotidase family.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

References (0)

Have you cited this product in a publication?
Let us know so we can reference it here.
Cite this product

Bioinformatics

Protein Aliases: 5'-nucleotidase domain-containing protein 1; 5'-nucleotidase, cytosolic II-like 1 protein

View more View less

Gene Aliases: C6orf200; LP2642; NT5C2L1; NT5DC1

View more View less

UniProt ID: (Human) Q5TFE4

View more View less

Entrez Gene ID: (Human) 221294

View more View less

Function(s)
5'-nucleotidase activity metal ion binding
Process(es)
dephosphorylation
It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-9dd846b7f-s9zn4:80/100.66.128.142:80.
git-commit: 17e8f6c92ed407c129a469238c057b56fdf8abbb
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0