Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Primary Antibodies ›
  • NOTCH2 Antibodies

Invitrogen

NOTCH2 VHH-8His-Cys-tag Recombinant Alpaca Monoclonal Antibody (SAA1287)

View all (26) NOTCH2 antibodies

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Protocols
Questions & Answers
Datasheet
Protocols
Questions & Answers

Cite NOTCH2 VHH-8His-Cys-tag Recombinant Alpaca Monoclonal Antibody (SAA1287)

Product Details

MA5-54648

Applications
Tested Dilution
Publications

ELISA (ELISA)

Assay-dependent
-
Product Specifications

Species Reactivity

Human

Host/Isotype

Alpaca / VHH

Expression System

CHO cells

Class

Recombinant Monoclonal

Type

Antibody

Clone

SAA1287

Immunogen

Recombinant Human NOTCH2.

Conjugate

Unconjugated Unconjugated Unconjugated

Form

Liquid

Concentration

1 mg/mL

Purification

Metal-chelate chromatography

Storage buffer

PBS

Contains

no preservative

Storage conditions

Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.

RRID

AB_3666896

Product Specific Information

Compatible with Direct ELISA

Target Information

Neurogenic locus notch homolog protein 2 (NOTCH2) is a highly conserved Notch signaling pathway protein. NOTCH2 is a type 1 transmembrane protein with an extracellular domain consisting of multiple epidermal growth factor-like (EGF) repeats, and an intracellular domain consisting of multiple, different domain types. NOTCH2 functions as a receptor for membrane-bound ligands Jagged1, Jagged2 and Delta1 to regulate cell-fate determination. Defects in NOTCH2 are the cause of Alagille syndrome type 2 (ALGS2). Alagille syndrome is an autosomal dominant multisystem disorder defined clinically by hepatic bile duct paucity and cholestasis in association with cardiac, skeletal, and ophthalmologic manifestations. There are characteristic facial features and less frequent clinical involvement of the renal and vascular systems.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

References (0)

Have you cited this product in a publication?
Let us know so we can reference it here.
Cite this product

Bioinformatics

Protein Aliases: Neurogenic locus notch homolog protein 2; Notch 2; Notch homolog 2

View more View less

Gene Aliases: AGS2; HJCYS; hN2; NOTCH2

View more View less

UniProt ID: (Human) Q04721

View more View less

Entrez Gene ID: (Human) 4853

View more View less

Function(s)
receptor activity calcium ion binding protein binding transcription factor activity, ligand-activated RNA polymerase II transcription factor binding
Process(es)
cell fate determination marginal zone B cell differentiation pulmonary valve morphogenesis regulation of transcription, DNA-templated transcription initiation from RNA polymerase II promoter apoptotic process cell cycle arrest Notch signaling pathway multicellular organism development nervous system development negative regulation of cell proliferation organ morphogenesis cell growth stem cell population maintenance hemopoiesis intracellular receptor signaling pathway tissue regeneration negative regulation of apoptotic process positive regulation of Ras protein signal transduction bone remodeling regulation of developmental process atrial septum morphogenesis Notch signaling involved in heart development
It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0