Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Primary Antibodies ›
  • NME1 Antibodies

OriGene

NME1 Monoclonal Antibody (OTI3F2), TrueMAB™

View all (121) NME1 antibodies

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Protocols
Questions & Answers
Datasheet
Protocols
Questions & Answers

Cite NME1 Monoclonal Antibody (OTI3F2), TrueMAB™

  • Antibody Testing Data (10)
NME1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
Group 53 Created with Sketch.
NME1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
Group 53 Created with Sketch.

FIGURE: 1 / 10

{{ $ctrl.currentElement.advancedVerification.fullName }}
{{ $ctrl.currentElement.advancedVerification.text }}

NME1 Antibody (TA801258) in IHC (P)

Immunohistochemical staining of paraffin-embedded human lymphoma tissue using anti-NME1 mouse monoclonal antibody. (Heat-induced epitope retrieval by 10mM citric buffer, pH6.0, 120°C for 3min, TA801258) {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
Published figure supplied by benchsci-logo
PMID: {{$ctrl.currentElement.benchSciPubmedId}}
View Product

{{$ctrl.videoDesc}}

{{$ctrl.videoLongDesc}}

NME1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
NME1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
NME1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
NME1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
NME1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
NME1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
NME1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
NME1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
NME1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
NME1 Antibody in Western Blot (WB)
NME1 Monoclonal Antibody (OTI3F2), TrueMAB™

Product Details

TA801258

Applications
Tested Dilution
Publications

Western Blot (WB)

1:2,000
-

Immunohistochemistry (Paraffin) (IHC (P))

1:150
-
Product Specifications

Species Reactivity

Human

Host/Isotype

Mouse / IgG2a

Class

Monoclonal

Type

Antibody

Clone

OTI3F2

Immunogen

Full length human recombinant protein of human NME1 produced in E.coli.
View immunogen

Conjugate

Unconjugated Unconjugated Unconjugated

Form

Liquid

Concentration

1 mg/mL

Purification

Affinity chromatography

Storage buffer

PBS with 1% BSA, 50% glycerol

Contains

0.02% sodium azide

Storage conditions

-20°C, Avoid Freeze/Thaw Cycles

Shipping conditions

Ambient (domestic); Wet ice (international)

Target Information

NME1 was identified because of its reduced mRNA transcript levels in highly metastatic cells. Nucleoside diphosphate kinase (NDK) exists as a hexamer composed of 'A' (encoded by this gene) and 'B' (encoded by NME2) isoforms. Mutations in the gene have been identified in aggressive neuroblastomas. Two transcript variants encoding different isoforms have been found for this gene. Co-transcription of this gene and the neighboring downstream gene (NME2) generates naturally-occurring transcripts (NME1-NME2), which encodes a fusion protein comprised of sequence sharing identity with each individual gene product.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

References (0)

Have you cited this product in a publication?
Let us know so we can reference it here.
Cite this product

Bioinformatics

Protein Aliases: GAAD; Granzyme A activated DNase (GAAD); Granzyme A-activated DNase; GZMA activated DNase; included; Metastasis inhibition factor nm23; NDK A; NDP kinase A; NM23-H1; NM23H1B; NME1-NME2 spliced read-through transcript; non-metastatic cells 1, protein (NM23A) expressed in; Nucleoside diphosphate kinase A; Tumor metastatic process-associated protein

View more View less

Gene Aliases: AWD; GAAD; NB; NBS; NDKA; NDPK-A; NDPKA; NM23; NM23-H1; NME1

View more View less

UniProt ID: (Human) P15531

View more View less

Entrez Gene ID: (Human) 4830

View more View less

Function(s)
magnesium ion binding RNA polymerase II regulatory region sequence-specific DNA binding single-stranded DNA binding deoxyribonuclease activity nucleoside diphosphate kinase activity protein binding ATP binding GTP binding intermediate filament binding protein kinase binding identical protein binding gamma-tubulin binding ribosomal small subunit binding poly(A) RNA binding
Process(es)
negative regulation of myeloid leukocyte differentiation purine nucleotide metabolic process nucleoside diphosphate phosphorylation GTP biosynthetic process pyrimidine nucleotide metabolic process UTP biosynthetic process CTP biosynthetic process DNA metabolic process endocytosis lactation negative regulation of cell proliferation nucleoside triphosphate biosynthetic process negative regulation of gene expression positive regulation of neuron projection development response to amine nucleobase-containing small molecule interconversion hippocampus development response to testosterone cellular response to drug regulation of apoptotic process positive regulation of DNA binding positive regulation of epithelial cell proliferation response to cAMP cellular response to glucose stimulus cellular response to fatty acid
It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0