Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Primary Antibodies ›
  • Nidogen 2 Antibodies

Abnova

NID2 Monoclonal Antibody (4G8)

View all (9) Nidogen 2 antibodies

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Protocols
Questions & Answers
Datasheet
Protocols
Questions & Answers

Cite NID2 Monoclonal Antibody (4G8)

  • Antibody Testing Data (1)
NID2 Antibody in Western Blot (WB)
Group 53 Created with Sketch.
NID2 Antibody in Western Blot (WB)
Group 53 Created with Sketch.

FIGURE: 1 / 1

{{ $ctrl.currentElement.advancedVerification.fullName }}
{{ $ctrl.currentElement.advancedVerification.text }}

NID2 Antibody (H00022795-M01) in WB

Western Blot analysis of NID2 expression in transfected 293T cell line by NID2 monoclonal antibody (M01), clone 4G8. Lane 1: NID2 transfected lysate (Predicted MW: 107.1 KDa). Lane 2: Non-transfected lysate. . {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
Published figure supplied by benchsci-logo
PMID: {{$ctrl.currentElement.benchSciPubmedId}}
View Product

{{$ctrl.videoDesc}}

{{$ctrl.videoLongDesc}}

NID2 Antibody in Western Blot (WB)
NID2 Monoclonal Antibody (4G8)

Product Details

H00022795-M01

Applications
Tested Dilution
Publications

Western Blot (WB)

1-5 µg/mL
-

ELISA (ELISA)

10 µg/mL
-
Product Specifications

Species Reactivity

Human

Host/Isotype

Mouse / IgG2b, kappa

Class

Monoclonal

Type

Antibody

Clone

4G8

Immunogen

NID2 (NP_031387.2, 1276 a.a. approximately 1375 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
View immunogen

Conjugate

Unconjugated Unconjugated Unconjugated

Form

Liquid

Concentration

1 mg/mL

Purification

Protein A

Storage buffer

PBS, pH 7.4

Contains

no preservative

Storage conditions

-20°C, Avoid Freeze/Thaw Cycles

Shipping conditions

Wet ice

Product Specific Information

Product may be used with Western Blot (Transfected lysate).

Immunogen sequence: PNGLTFDPFS KLLCWADAGT KKLECTLPDG TGRRVIQNNL KYPFSIVSYA DHFYHTDWRR DGVVSVNKHS GQFTDEYLPE QRSHLYGITA VYPYCPTGRK

Target Information

Basement membranes, which are composed of type IV collagens, laminins, perlecan, and nidogen, are thin pericellular protein matrices that control a large number of cellular activities, including adhesion, migration, differentiation, gene expression, and apoptosis. Basement membranes, which are composed of type IV collagens, perlecan, and nidogen, are thin pericellular protein matrices that control a large number of cellular activities, including adhesion, migration, differentiation, gene expression, and apoptosis.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

References (0)

Have you cited this product in a publication?
Let us know so we can reference it here.
Cite this product

Bioinformatics

Protein Aliases: NID-2; Nidogen; nidogen 2 (osteonidogen); Nidogen-2; Osteonidogen

View more View less

Gene Aliases: NID-2; NID2

View more View less

UniProt ID: (Human) Q14112

View more View less

Entrez Gene ID: (Human) 22795

View more View less

Function(s)
calcium ion binding protein binding collagen binding
Process(es)
cell adhesion cell-matrix adhesion extracellular matrix organization basement membrane organization
It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-55c564d944-rwwh4:80/100.66.128.142:80.
git-commit: 3092084cb0e494f0e07f07633350183de7fa10a8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0