Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Primary Antibodies ›
  • NDUFV2 Antibodies

Proteintech

NDUFV2 Monoclonal Antibody (3B4A5)

View all (11) NDUFV2 antibodies

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Protocols
Questions & Answers
Datasheet
Protocols
Questions & Answers

Cite NDUFV2 Monoclonal Antibody (3B4A5)

  • Antibody Testing Data (4)
NDUFV2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
Group 53 Created with Sketch.
NDUFV2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
Group 53 Created with Sketch.

FIGURE: 1 / 4

{{ $ctrl.currentElement.advancedVerification.fullName }}
{{ $ctrl.currentElement.advancedVerification.text }}

NDUFV2 Antibody (68150-1-IG) in IHC (P)

Immunohistochemical analysis of paraffin-embedded mouse heart tissue slide using 68150-1-Ig (NDUFV2 antibody) at dilution of 1:0 (under 40x lens). Heat mediated antigen retrieval with Tris-EDTA buffer (pH 9.0). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
Published figure supplied by benchsci-logo
PMID: {{$ctrl.currentElement.benchSciPubmedId}}
View Product

{{$ctrl.videoDesc}}

{{$ctrl.videoLongDesc}}

NDUFV2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
NDUFV2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
NDUFV2 Antibody in Western Blot (WB)
NDUFV2 Antibody in Western Blot (WB)
NDUFV2 Monoclonal Antibody (3B4A5)

Product Details

68150-1-IG

Applications
Tested Dilution
Publications

Western Blot (WB)

1:5,000-1:50,000
-

Immunohistochemistry (Paraffin) (IHC (P))

1:500-1:2,000
-

ELISA (ELISA)

Assay-dependent
-
Product Specifications

Species Reactivity

Chicken, Human, Mouse, Pig, Rabbit, Rat

Host/Isotype

Mouse / IgG1

Class

Monoclonal

Type

Antibody

Clone

3B4A5

Immunogen

Recombinant protein
View immunogen

Conjugate

Unconjugated Unconjugated Unconjugated

Form

Liquid

Purification

Protein G

Storage buffer

PBS with 50% glycerol

Contains

0.02% sodium azide

Storage conditions

-20°C

Shipping conditions

Wet ice

Product Specific Information

Immunogen sequence: MFFSAALRAR AAGLTAHWGR HVRNLHKTVM QNGAGGALFV HRDTPENNPD TPFDFTPENY KRIEAIVKNY PEGHKAAAVL PVLDLAQRQN GWLPISAMNK VAEVLQVPPM RVYEVATFYT MYNRKPVGKY HIQVCTTTPC MLRNSDSILE AIQKKLGIKV GETTPDKLFT LIEVECLGAC VNAPMVQIND NYYEDLTAKD IEEIIDELKA GKIPKPGPRS GRFSCEPAGG LTSLTEPPKG PGFGVQAGL

Target Information

Core subunit of the mitochondrial membrane respiratory chain NADH dehydrogenase (Complex I) that is believed to belong to the minimal assembly required for catalysis. Complex I functions in the transfer of electrons from NADH to the respiratory chain. The immediate electron acceptor for the enzyme is believed to be ubiquinone.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

References (0)

Have you cited this product in a publication?
Let us know so we can reference it here.
Cite this product

Bioinformatics

Protein Aliases: 24-kDa subunit of mitochondrial NADH dehydrogenase; complex I 24kDa subunit; complex I, mitochondrial respitory chain, 24 kD subunit; NADH dehydrogenase (ubiquinone) flavoprotein 2, 24kDa; NADH dehydrogenase [ubiquinone] flavoprotein 2, mitochondrial; NADH dehydrogenase ubiquinone flavoprotein 2; NADH dehydrogenase ubiquinone flavoprotein 2, mitochondrial; NADH-ubiquinone oxidoreductase 24 kDa subunit; NADH-ubiquinone oxidoreductase flavoprotein 2; NDUFV2; nuclear-encoded mitochondrial NADH-ubiquinone reductase 24Kd subunit

View more View less

Gene Aliases: 2900010C23Rik; CI-24k; NDUFV2

View more View less

UniProt ID: (Human) P19404, (Mouse) Q9D6J6, (Rat) P19234

View more View less

Entrez Gene ID: (Rabbit) 100358684, (Human) 4729, (Pig) 100037991, (Mouse) 72900, (Rat) 81728

View more View less

Function(s)
NADH dehydrogenase (ubiquinone) activity electron carrier activity metal ion binding 2 iron, 2 sulfur cluster binding NADH dehydrogenase activity oxidoreductase activity iron-sulfur cluster binding molecular_function
Process(es)
mitochondrial electron transport, NADH to ubiquinone nervous system development mitochondrial respiratory chain complex I assembly cardiac muscle tissue development transport oxidation-reduction process biological_process
It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0