Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: VLIQPSNSA AHVQRFCQVL GLDPDQLFML QPREPDVSSP TRLPQPCSMR HLVSHYLDIA SVPRRSFFEL LACLSLHELE REKLLEFSSA QGQEELFEYC NRPRRTILEV LCDFPHTAAA IPPDYLLDLI PVIRPRAFSI ASSLLTHPSR LQILVAVVQF QTRLKEPRRG LCSSWLASLD PGQGPVRVPL WVRPGSLAFP ETPDTPVIMV GPGTGVAPFR AAIQERVAQG QTGNFLFFGC RWRDQDFYWE AEWQELEKRD CLTLIPAFSR EQEQKIYVQH RLRELGSLVW ELLDRQGAYF YLAGNAKSMP ADVSEALMSI FQEEGGLCSP DAAAYLARLQ QTRRFQTETW A (248-597 aa encoded by BC015735)
This gene encodes an NADPH-dependent diflavin reductase that contains both flavin mononucleotide (FMN) and flavin adenine dinucleotide (FAD) binding domains. The encoded protein is an enzyme that catalyzes the transfers electrons from NADPH through FAD and FMN cofactors to potential redox partners. Alternative splicing results in multiple transcript variants.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: NADPH-dependent diflavin oxidoreductase 1; NADPH-dependent FMN and FAD-containing oxidoreductase; NDOR1; Novel reductase 1
Gene Aliases: 4930447P04Rik; bA350O14.9; Ndor; NDOR1; NR1
UniProt ID: (Human) Q9UHB4, (Mouse) A2AI05
Entrez Gene ID: (Human) 27158, (Mouse) 78797, (Rat) 311799
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support