Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Secondary Antibodies ›
  • Mouse anti-Rat IgG2b Secondary Antibodies

Invitrogen

Mouse anti-Rat IgG2b Secondary Antibody, Biotin, eBioscience™

View all (12) Mouse anti-Rat IgG2b antibodies

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Create Citation
Questions & Answers
Datasheet
Create Citation
Questions & Answers

Cite Mouse anti-Rat IgG2b Secondary Antibody, Biotin, eBioscience™

  • Antibody Testing Data (1)
Rat IgG2b Secondary Antibody in Immunohistochemistry (Paraffin) (IHC (P))
Group 53 Created with Sketch.
Rat IgG2b Secondary Antibody in Immunohistochemistry (Paraffin) (IHC (P))
Group 53 Created with Sketch.

FIGURE: 1 / 1

{{ $ctrl.currentElement.advancedVerification.fullName }}
{{ $ctrl.currentElement.advancedVerification.text }}

Rat IgG2b Secondary Antibody (13-4815-82) in IHC (P)

Immunohistochemistry of formalin-fixed paraffin embedded human tonsil tissue using 20 µg/mL of Rat IgG2b K Isotype Control Purified (left) or 20 µg/mL of Anti-Mouse Granzyme B Purified (right) followed by 10 µg/mL of Anti-Rat IgG2b Biotin, Streptavidin HRP and DAB visualization. Nuclei are counterstained with hematoxylin. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
Published figure supplied by benchsci-logo
PMID: {{$ctrl.currentElement.benchSciPubmedId}}
View Product

{{$ctrl.videoDesc}}

{{$ctrl.videoLongDesc}}

Rat IgG2b Secondary Antibody in Immunohistochemistry (Paraffin) (IHC (P))

Product Details

13-4815-82

Applications
Tested Dilution

Immunohistochemistry (Paraffin) (IHC (P))

10 µg/mL

Immunohistochemistry (Frozen) (IHC (F))

10 µg/mL

Immunocytochemistry (ICC/IF)

Assay-Dependent
Product Specifications

Species Reactivity

Rat

Host/Isotype

Mouse / IgG1, kappa

Class

Monoclonal

Type

Secondary Antibody

Clone

R2B-7C3

Conjugate

Biotin Biotin Biotin
  • eFluor 570
  • eFluor 660
  • FITC
  • PE 
  • Request custom conjugation

Form

Liquid

Concentration

0.5 mg/mL

Purification

Affinity chromatography

Storage buffer

PBS, pH 7.2

Contains

0.09% sodium azide

Storage conditions

4°C, store in dark, DO NOT FREEZE!

Shipping conditions

Ambient (domestic); Wet ice (international)

RRID

AB_2572792

Target

IgG2b

Antibody Form

Whole Antibody

Product Specific Information

Description: The monoclonal antibody R2B-7C3 recognizes Rat IgG2b antibodies and can be used as a second step reagent in flow cytometry and microscopy. The monoclonal does not recognize other rat isotypes nor does it crossreact to mouse IgG2b or any mouse isotype antibodies.

Applications Reported: This R2B-7C3 antibody has been reported for use in immunohistochemical staining of frozen tissue sections, immunohistochemical staining of formalin-fixed paraffin embedded tissue sections, microscopy, and immunocytochemistry.

Applications Tested: This R2B-7C3 antibody has been tested by immunohistochemistry of formalin-fixed paraffin embedded tissue sections that have been stained first with a rat IgG2b anti-human antibody. This can be used at less than or equal to 10 µg/mL. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.

Filtration: 0.2 µm post-manufacturing filtered.

Target Information

Anti-Rat secondary antibodies are affinity-purified antibodies with well-characterized specificity for rat immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

View product citations for antibody 13-4815-82 on CiteAb

References (0)

Have you cited this product in a publication?
Let us know so we can reference it here.
Cite this product
It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0