Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Primary Antibodies ›
  • Metallothionein Antibodies

Invitrogen

Metallothionein Monoclonal Antibody (8D8), PerCP

View all (13) Metallothionein antibodies

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Protocols
Questions & Answers
Datasheet
Protocols
Questions & Answers

Cite Metallothionein Monoclonal Antibody (8D8), PerCP

  • Antibody Testing Data (1)
Metallothionein Antibody in Western Blot (WB)
Group 53 Created with Sketch.
Metallothionein Antibody in Western Blot (WB)
Group 53 Created with Sketch.

FIGURE: 1 / 1

{{ $ctrl.currentElement.advancedVerification.fullName }}
{{ $ctrl.currentElement.advancedVerification.text }}

Metallothionein Antibody (MA5-45904) in WB

Western Blot analysis of Pseudomonas aeruginosa Metallothionein (PmtA) GST tagged showing detection of 36 kDa Metallothionein protein. Lane 1: Molecular Weight Ladder (MW). Lane 2: GST. Lane 3: Purified Pseudomonas aeruginosa Metallothionein (PmtA) GST tagged. Load: 1 µg. Blocking: 5% Skim Milk powder in TBST. Samples were incubated with Metallothionein monoclonal antibody (Product # MA5-45904) at 1:1,000 for 2 hours at RT, followed by Goat Anti-Mouse IgG:HRP at 1:5,000 for 1 hour at RT. Color Development: ECL solution for 5 min in RT. Pre... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
Published figure supplied by benchsci-logo
PMID: {{$ctrl.currentElement.benchSciPubmedId}}
View Product

{{$ctrl.videoDesc}}

{{$ctrl.videoLongDesc}}

Metallothionein Antibody in Western Blot (WB)

Product Details

MA5-45904

Applications
Tested Dilution
Publications

Western Blot (WB)

1:1,000
-

ELISA (ELISA)

1:800
-
Product Specifications

Species Reactivity

Bacteria

Host/Isotype

Mouse / IgG1

Class

Monoclonal

Type

Antibody

Clone

8D8

Immunogen

Recombinant full length Pseudomonas aeruginosa Metallothionein protein

Conjugate

PerCP PerCP PerCP
  • APC 
  • FITC
  • PE 
  • Request custom conjugation

Excitation/Emission Max

482/675 nm View spectra spectra

Form

Liquid

Concentration

1 mg/mL

Purification

Protein G

Storage buffer

95.64mM phosphate/2.48mM MES, pH 7.4, with 0.5M EDTA

Contains

no preservative

Storage conditions

4°C

Shipping conditions

Ambient (domestic); Wet ice (international)

RRID

AB_2932358

Product Specific Information

A 1:1,000 dilution of MA5-45904 was sufficient for detection of Metallothionein in 1 µg of purified Pseudomonas aeruginosa Metallothionein (PmtA)-GST tagged protein by ECL immunoblot analysis using goat anti-mouse IgG:HRP as the secondary antibody.|Detects 36 kDa band corresponding to Pseudomonas aeruginosa Metallothionein.

Target Information

Metallothioneins (MTs) are a family of low molecular weight, heavy metal-binding proteins characterized by a high cysteine content and lack of aromatic amino acids. MTs bind 7 to 12 heavy metal atoms per molecule of protein. They are ubiquitous in the animal and plant kingdoms and are also found in prokaryotes. In mammals, the cysteine residues are absolutely conserved and serve to coordinate heavy metal atoms such as zinc, cadmium, and copper via mercaptide linkages.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

References (0)

Have you cited this product in a publication?
Let us know so we can reference it here.
Cite this product

Bioinformatics

Protein Aliases: CES 1; CES1; Metallothionein; Metallothionein 2; Metallothionein 2A; Metallothionein IA; Metallothionein II; Metallothionein2; MGC32848; MT 1A; MT 2; MT 2A; MT IA; MT II; MT2; MT2A; Probable pmtA

View more View less

It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-9c87ff496-4djhr:80/100.66.128.142:80.
git-commit: 146a63324c51a096ecccde4707f5fe4015d9ef66
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36_secrets_rm