Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Primary Antibodies ›
  • MTH1 Antibodies

Proteintech

MTH1 Monoclonal Antibody (2D7G4), CoraLite® Plus 488

View all (14) MTH1 antibodies

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Protocols
Questions & Answers
Datasheet
Protocols
Questions & Answers

Cite MTH1 Monoclonal Antibody (2D7G4), CoraLite® Plus 488

  • Antibody Testing Data (2)
MTH1 Antibody in Immunocytochemistry (ICC/IF)
Group 53 Created with Sketch.
MTH1 Antibody in Immunocytochemistry (ICC/IF)
Group 53 Created with Sketch.

FIGURE: 1 / 2

{{ $ctrl.currentElement.advancedVerification.fullName }}
{{ $ctrl.currentElement.advancedVerification.text }}

MTH1 Antibody (CL488-67443) in ICC/IF

Immunofluorescent analysis of (4% PFA) fixed HepG2 cells using CoraLite® Plus 488 MTH1 antibody (CL488-67443, Clone: 2D7G4 ) at dilution of 1:200. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
Published figure supplied by benchsci-logo
PMID: {{$ctrl.currentElement.benchSciPubmedId}}
View Product

{{$ctrl.videoDesc}}

{{$ctrl.videoLongDesc}}

MTH1 Antibody in Immunocytochemistry (ICC/IF)
MTH1 Antibody in Flow Cytometry (Flow)
MTH1 Monoclonal Antibody (2D7G4), CoraLite® Plus 488

Product Details

CL488-67443

Applications
Tested Dilution
Publications

Immunocytochemistry (ICC/IF)

1:50-1:500
-

Flow Cytometry (Flow)

0.4 µg/1x10^6 cell suspension
-
Product Specifications

Species Reactivity

Human

Host/Isotype

Mouse / IgG1

Class

Monoclonal

Type

Antibody

Clone

2D7G4

Immunogen

Recombinant protein
View immunogen

Conjugate

CoraLite® Plus 488 CoraLite® Plus 488 CoraLite® Plus 488
  • Unconjugated
  • CoraLite 594
  • Request custom conjugation

Excitation/Emission Max

493/522 nm

Form

Liquid

Purification

Protein G

Storage buffer

PBS with 0.5% BSA, 50% glycerol

Contains

0.05% ProClin 300

Storage conditions

-20°C, store in dark

Shipping conditions

Wet ice

Product Specific Information

Immunogen sequence: MSGISPQQMG EPEGSWSGKN PGTMGASRLY TLVLVLQPQR VLLGMKKRGF GAGRWNGFGG KVQEGETIED GARRELQEES GLTVDALHKV GQIVFEFVGE PELMDVHVFC TDSIQGTPVE SDEMRPCWFQ LDQIPFKDMW PDDSYWFPLL LQKKKFHGYF KFQGQDTILD YTLREVDTV

Target Information

Oxygen radicals damage chromosomal DNA causing cell death and inducing mutations. Among the various classes of DNA damage caused by oxygen radicals, an oxidized form of guanine base (8-oxoguanine) appears to be important as it can pair with cytosine and adenine and G:C to T:A transversion mutation occurs. A significant amount of 8-OxoG is formed in the chromosomal DNA of mammalian cells, with most damaged nucleotides excised from the DNA and excreted in the urine. Along with 8-oxoG being present in oxidatively damaged DNA, 8-oxo-deoxyguanosine (8-oxo-dGTP) is formed in the nucleotide pool during normal cellular metabolism and following oxidative stress. The 8-oxo-dGTP nucleotide can be incorporated in DNA during polymerization and can result in a mispairing unless repaired. MTH converts 8-oxo-dGTP in the nucleotide pool to the monophosphate and prevents the misincorporation of 8-oxo-dGTP into DNA (Figure courtesy of Dr. Mark Kelley). MTH also recognizes 8-oxo-rGTP, which could incorporate into RNA during gene transcription leading to missense or nonsense protein production.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

References (0)

Have you cited this product in a publication?
Let us know so we can reference it here.
Cite this product

Bioinformatics

Protein Aliases: 2-hydroxy-dATP diphosphatase; 7,8-dihydro-8-oxoguanine triphosphatase; 8-oxo-7,8-dihydrodeoxyguanosine triphosphatase; 8-oxo-7,8-dihydroguanosine triphosphatase; 8-oxo-dGTPase; Methylated purine nucleoside triphosphate hydrolase; mutT human homolog 1; Nucleoside diphosphate-linked moiety X motif 1; nucleoside diphosphate-linked moiety X-type motif 1; nudix (nucleoside diphosphate linked moiety X)-type motif 1; Nudix motif 1; Oxidized purine nucleoside triphosphate hydrolase

View more View less

Gene Aliases: MTH1; NUDT1

View more View less

UniProt ID: (Human) P36639

View more View less

Entrez Gene ID: (Human) 4521

View more View less

Function(s)
GTPase activity protein binding 8-oxo-7,8-dihydroguanosine triphosphate pyrophosphatase activity snoRNA binding 8-oxo-7,8-dihydrodeoxyguanosine triphosphate pyrophosphatase activity GTP diphosphatase activity 2-hydroxy-adenosine triphosphate pyrophosphatase activity 2-hydroxy-(deoxy)adenosine-triphosphate pyrophosphatase activity metal ion binding ATP diphosphatase activity m7G(5')pppN diphosphatase activity
Process(es)
purine nucleotide catabolic process dGTP catabolic process DNA repair response to oxidative stress nucleobase-containing small molecule catabolic process DNA protection dATP catabolic process
It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-9dd846b7f-s9zn4:80/100.66.128.142:80.
git-commit: 17e8f6c92ed407c129a469238c057b56fdf8abbb
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0