Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Primary Antibodies ›
  • MTH1 Antibodies

Invitrogen

MTH1 Recombinant Rabbit Monoclonal Antibody (0P6T10)

View all (14) MTH1 antibodies

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Protocols
Questions & Answers
Datasheet
Protocols
Questions & Answers

Cite MTH1 Recombinant Rabbit Monoclonal Antibody (0P6T10)

  • Antibody Testing Data (1)
MTH1 Antibody in Western Blot (WB)
Group 53 Created with Sketch.
MTH1 Antibody in Western Blot (WB)
Group 53 Created with Sketch.

FIGURE: 1 / 1

{{ $ctrl.currentElement.advancedVerification.fullName }}
{{ $ctrl.currentElement.advancedVerification.text }}

MTH1 Antibody (MA5-35421) in WB

Western blot analysis of MTH1 in extracts of various cell lines. Samples were incubated with MTH1 Monoclonal antibody (Product # MA5-35421) using a dilution of 1:1,000, followed by HRP Goat Anti-Rabbit IgG (H+L) at a dilution of 1:10,000. Lysates/proteins: 25 µg per lane. Blocking buffer: 3% nonfat dry milk in TBST. Detection: ECL Basic Kit. Exposure time: 10s. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
Published figure supplied by benchsci-logo
PMID: {{$ctrl.currentElement.benchSciPubmedId}}
View Product

{{$ctrl.videoDesc}}

{{$ctrl.videoLongDesc}}

MTH1 Antibody in Western Blot (WB)

Product Details

MA5-35421

Applications
Tested Dilution
Publications

Western Blot (WB)

1:500-1:2,000
-

ELISA (ELISA)

1 µg/mL
-
Product Specifications

Species Reactivity

Human

Host/Isotype

Rabbit / IgG

Expression System

HEK293 cells

Class

Recombinant Monoclonal

Type

Antibody

Clone

0P6T10

Immunogen

Recombinant fusion protein containing a sequence corresponding to amino acids 50-200 of human MTH1 (P36639)

Conjugate

Unconjugated Unconjugated Unconjugated

Form

Liquid

Concentration

0.63 mg/mL

Purification

Affinity Chromatography

Storage buffer

PBS, pH 7.3, with 50% glycerol, 0.05% BSA

Contains

0.02% sodium azide

Storage conditions

-20°C, Avoid Freeze/Thaw Cycles

Shipping conditions

Ambient (domestic); Wet ice (international)

RRID

AB_2849322

Product Specific Information

Immunogen sequence: RRELQEESGL TVDALHKVGQ IVFEFVGEPE LMDVHVFCTD SIQGTPVESD EMRPCWFQLD QIPFKDMWPD DSYWFPLLLQ KKKFHGYFKF QGQDTILDYT LREVDTV

Target Information

Oxygen radicals damage chromosomal DNA causing cell death and inducing mutations. Among the various classes of DNA damage caused by oxygen radicals, an oxidized form of guanine base (8-oxoguanine) appears to be important as it can pair with cytosine and adenine and G:C to T:A transversion mutation occurs. A significant amount of 8-OxoG is formed in the chromosomal DNA of mammalian cells, with most damaged nucleotides excised from the DNA and excreted in the urine. Along with 8-oxoG being present in oxidatively damaged DNA, 8-oxo-deoxyguanosine (8-oxo-dGTP) is formed in the nucleotide pool during normal cellular metabolism and following oxidative stress. The 8-oxo-dGTP nucleotide can be incorporated in DNA during polymerization and can result in a mispairing unless repaired. MTH converts 8-oxo-dGTP in the nucleotide pool to the monophosphate and prevents the misincorporation of 8-oxo-dGTP into DNA (Figure courtesy of Dr. Mark Kelley). MTH also recognizes 8-oxo-rGTP, which could incorporate into RNA during gene transcription leading to missense or nonsense protein production.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

References (0)

Have you cited this product in a publication?
Let us know so we can reference it here.
Cite this product

Bioinformatics

Protein Aliases: 2-hydroxy-dATP diphosphatase; 7,8-dihydro-8-oxoguanine triphosphatase; 8-oxo-7,8-dihydrodeoxyguanosine triphosphatase; 8-oxo-7,8-dihydroguanosine triphosphatase; 8-oxo-dGTPase; Methylated purine nucleoside triphosphate hydrolase; mutT human homolog 1; Nucleoside diphosphate-linked moiety X motif 1; nucleoside diphosphate-linked moiety X-type motif 1; nudix (nucleoside diphosphate linked moiety X)-type motif 1; Nudix motif 1; Oxidized purine nucleoside triphosphate hydrolase

View more View less

Gene Aliases: MTH1; NUDT1

View more View less

UniProt ID: (Human) P36639

View more View less

Entrez Gene ID: (Human) 4521

View more View less

Function(s)
GTPase activity protein binding 8-oxo-7,8-dihydroguanosine triphosphate pyrophosphatase activity snoRNA binding 8-oxo-7,8-dihydrodeoxyguanosine triphosphate pyrophosphatase activity GTP diphosphatase activity 2-hydroxy-adenosine triphosphate pyrophosphatase activity 2-hydroxy-(deoxy)adenosine-triphosphate pyrophosphatase activity metal ion binding ATP diphosphatase activity m7G(5')pppN diphosphatase activity
Process(es)
purine nucleotide catabolic process dGTP catabolic process DNA repair response to oxidative stress nucleobase-containing small molecule catabolic process DNA protection dATP catabolic process
It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-9dd846b7f-s9zn4:80/100.66.128.142:80.
git-commit: 17e8f6c92ed407c129a469238c057b56fdf8abbb
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0