Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Primary Antibodies ›
  • MIXL1 Antibodies

Invitrogen

MIXL1 Polyclonal Antibody

View all (11) MIXL1 antibodies

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Protocols
Questions & Answers
Datasheet
Protocols
Questions & Answers

Cite MIXL1 Polyclonal Antibody

  • Antibody Testing Data (4)
MIXL1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
Group 53 Created with Sketch.
MIXL1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
Group 53 Created with Sketch.

FIGURE: 1 / 4

{{ $ctrl.currentElement.advancedVerification.fullName }}
{{ $ctrl.currentElement.advancedVerification.text }}

MIXL1 Antibody (PA5-114762) in IHC (P)

Immunohistochemistry analysis of MIXL1 in mouse heart tissue. Samples were treated with formaldehyde and treated with citrate buffer for antigen retrieval, blocked, and incubated (1.5 hours at 22°C) with polyclonal antibody (Product # PA5-114762) at a dilution of 1:100. Secondary staining was applied with HRP conjugated anti-Rabbit. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
Published figure supplied by benchsci-logo
PMID: {{$ctrl.currentElement.benchSciPubmedId}}
View Product

{{$ctrl.videoDesc}}

{{$ctrl.videoLongDesc}}

MIXL1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
MIXL1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
MIXL1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
MIXL1 Antibody in Western Blot (WB)
MIXL1 Polyclonal Antibody

Product Details

PA5-114762

Applications
Tested Dilution
Publications

Western Blot (WB)

1:500-1:2,000
-

Immunohistochemistry (Paraffin) (IHC (P))

1:50-1:200
-
Product Specifications

Species Reactivity

Human, Mouse, Rat

Host/Isotype

Rabbit / IgG

Class

Polyclonal

Type

Antibody

Immunogen

A synthesized peptide derived from human MIXL1(Accession Q9H2W2), corresponding to amino acid residues L115-G165.
View immunogen

Conjugate

Unconjugated Unconjugated Unconjugated

Form

Liquid

Concentration

1 mg/mL

Purification

Affinity chromatography

Storage buffer

PBS with 50% glycerol

Contains

0.02% sodium azide

Storage conditions

-20°C

Shipping conditions

Wet ice

RRID

AB_2899398

Product Specific Information

Antibody detects endogenous levels of total MIXL1.

Target Information

MIXL1 is a novel human Mix-like homeobox gene. In normal hematopoiesis, its expression appears to be restricted to immature B- and T-lymphoid cells.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

References (0)

Have you cited this product in a publication?
Let us know so we can reference it here.
Cite this product

Bioinformatics

Protein Aliases: hMix; Homeobox protein MIXL1; Homeodomain protein MIX; Mix-like homeobox protein 1; Mix.1 homeobox-like protein; MIX1 homeobox-like protein 1; mMix

View more View less

Gene Aliases: MILD1; MIX; MIXL; MIXL1; Mm1; Mml

View more View less

UniProt ID: (Human) Q9H2W2, (Mouse) Q9WUI0

View more View less

Entrez Gene ID: (Human) 83881, (Rat) 289311, (Mouse) 27217

View more View less

Function(s)
RNA polymerase II core promoter proximal region sequence-specific DNA binding RNA polymerase II distal enhancer sequence-specific DNA binding core promoter proximal region sequence-specific DNA binding transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding RNA polymerase II transcription factor binding RNA polymerase II repressing transcription factor binding transcriptional activator activity, RNA polymerase II transcription factor binding transcriptional activator activity, RNA polymerase II distal enhancer sequence-specific binding protein homodimerization activity sequence-specific DNA binding DNA binding
Process(es)
endoderm formation hematopoietic progenitor cell differentiation regulation of transcription, DNA-templated transcription from RNA polymerase II promoter gastrulation endoderm development heart development hemopoiesis endodermal cell differentiation cell migration involved in gastrulation positive regulation of transcription from RNA polymerase II promoter digestive tract development negative regulation of hematopoietic progenitor cell differentiation positive regulation of mesoderm development transcription, DNA-templated multicellular organism development cell differentiation
It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0