Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Primary Antibodies ›
  • MICAL1 Antibodies

OriGene

MICAL1 Monoclonal Antibody (OTI1C2), TrueMAB™

View all (34) MICAL1 antibodies

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Protocols
Questions & Answers
Datasheet
Protocols
Questions & Answers

Cite MICAL1 Monoclonal Antibody (OTI1C2), TrueMAB™

  • Antibody Testing Data (11)
MICAL1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
Group 53 Created with Sketch.
MICAL1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
Group 53 Created with Sketch.

FIGURE: 1 / 11

{{ $ctrl.currentElement.advancedVerification.fullName }}
{{ $ctrl.currentElement.advancedVerification.text }}

MICAL1 Antibody (CF501841) in IHC (P)

Immunohistochemical staining of paraffin-embedded human lymphoma tissue using anti-MICAL1 mouse monoclonal antibody (TA501841) at 1:150 dilution. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
Published figure supplied by benchsci-logo
PMID: {{$ctrl.currentElement.benchSciPubmedId}}
View Product

{{$ctrl.videoDesc}}

{{$ctrl.videoLongDesc}}

MICAL1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
MICAL1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
MICAL1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
MICAL1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
MICAL1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
MICAL1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
MICAL1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
MICAL1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
MICAL1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
MICAL1 Antibody in Western Blot (WB)
MICAL1 Antibody in Flow Cytometry (Flow)
MICAL1 Monoclonal Antibody (OTI1C2), TrueMAB™

Product Details

CF501841

Applications
Tested Dilution
Publications

Western Blot (WB)

1:2,000
-

Immunohistochemistry (Paraffin) (IHC (P))

1:150
-

Flow Cytometry (Flow)

1:100
-
Product Specifications

Species Reactivity

Human

Host/Isotype

Mouse / IgG1

Class

Monoclonal

Type

Antibody

Clone

OTI1C2

Immunogen

Full length human recombinant protein of human MICAL1 produced in HEK293T cell.
View immunogen

Conjugate

Unconjugated Unconjugated Unconjugated

Form

Lyophilized

Concentration

1 mg/mL

Purification

Affinity chromatography

Storage buffer

PBS, pH 7.3, with 8% trehalose

Contains

no preservative

Storage conditions

-20°C, Avoid Freeze/Thaw Cycles

Shipping conditions

Ambient (domestic); Wet ice (international)

Product Specific Information

For reconstitution, we recommend adding 100 µL distilled water to a final antibody concentration of about 1 mg/mL. To use this carrier-free antibody for conjugation experiments, we strongly recommend performing another round of desalting. (Zeba Spin Desalting Columns, 7KMWCO, 0.5 mL, Product # 89882)

Target Information

May be a cytoskeletal regulator that connects NEDD9 to intermediate filaments.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

References (0)

Have you cited this product in a publication?
Let us know so we can reference it here.
Cite this product

Bioinformatics

Protein Aliases: [F-actin]-monooxygenase MICAL1; MICAL-1; Molecule interacting with CasL protein 1; NEDD9-interacting protein with calponin homology and LIM domains

View more View less

Gene Aliases: MICAL; MICAL-1; MICAL1; NICAL

View more View less

UniProt ID: (Human) Q8TDZ2

View more View less

Entrez Gene ID: (Human) 64780

View more View less

Function(s)
actin binding protein binding zinc ion binding oxidoreductase activity, acting on paired donors, with incorporation or reduction of molecular oxygen, NAD(P)H as one donor, and incorporation of one atom of oxygen SH3 domain binding Rab GTPase binding FAD binding
Process(es)
negative regulation of protein phosphorylation cytoskeleton organization signal transduction sulfur oxidation actin filament depolymerization negative regulation of cysteine-type endopeptidase activity involved in apoptotic process oxidation-reduction process
It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-9dd846b7f-s9zn4:80/100.66.128.142:80.
git-commit: 17e8f6c92ed407c129a469238c057b56fdf8abbb
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0