Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Primary Antibodies ›
  • MIB1 Antibodies

Invitrogen

MIB1 Monoclonal Antibody (6A9C9)

View all (15) MIB1 antibodies

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Protocols
Questions & Answers
Datasheet
Protocols
Questions & Answers

Cite MIB1 Monoclonal Antibody (6A9C9)

  • Antibody Testing Data (6)
MIB1 Antibody in Immunocytochemistry (ICC/IF)
Group 53 Created with Sketch.
MIB1 Antibody in Immunocytochemistry (ICC/IF)
Group 53 Created with Sketch.

FIGURE: 1 / 6

{{ $ctrl.currentElement.advancedVerification.fullName }}
{{ $ctrl.currentElement.advancedVerification.text }}

MIB1 Antibody (MA5-31797) in ICC/IF

Immunocytochemistry analysis of MIB1 in Hela cells (green). Sample was incubated with MIB1 monoclonal antibody (Product # MA5-31797) using a dilution of 1:200-1:1000 followed by DRAQ5 fluorescent DNA dye (blue), and Alexa Fluor- 555 phalloidin (red labeled actin filaments) . {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
Published figure supplied by benchsci-logo
PMID: {{$ctrl.currentElement.benchSciPubmedId}}
View Product

{{$ctrl.videoDesc}}

{{$ctrl.videoLongDesc}}

MIB1 Antibody in Immunocytochemistry (ICC/IF)
MIB1 Antibody in Western Blot (WB)
MIB1 Antibody in Western Blot (WB)
MIB1 Antibody in Western Blot (WB)
MIB1 Antibody in Flow Cytometry (Flow)
MIB1 Antibody in ELISA (ELISA)
MIB1 Monoclonal Antibody (6A9C9)

Product Details

MA5-31797

Applications
Tested Dilution
Publications

Western Blot (WB)

1:500-1:2,000
-

Immunocytochemistry (ICC/IF)

1:200-1:1,000
-

Flow Cytometry (Flow)

1:200-1:400
-

ELISA (ELISA)

1:10,000
-
Product Specifications

Species Reactivity

Human

Host/Isotype

Mouse / IgG1

Class

Monoclonal

Type

Antibody

Clone

6A9C9

Immunogen

Purified recombinant fragment of human MIB1 (AA: 6-221) expressed in E. coli.
View immunogen

Conjugate

Unconjugated Unconjugated Unconjugated

Form

Liquid

Concentration

1 mg/mL

Purification

Protein G

Storage buffer

PBS

Contains

0.05% sodium azide

Storage conditions

Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.

Shipping conditions

Ambient (domestic); Wet ice (international)

RRID

AB_2787420

Product Specific Information

MA5-31797 has been tested in indirect ELISA.

Target Information

MIB is an E3 ubiquitin-protein ligase that mediates ubiquitination of Delta receptors, which act as ligands of Notch proteins. This protein positively regulates the Delta-mediated Notch signaling by ubiquitinating the intracellular domain of Delta, leading to endocytosis of Delta receptors. MIB probably mediates ubiquitination and subsequent proteasomal degradation of DAPK1, thereby antagonizing anti-apoptotic effects of DAPK1 to promote TNF-induced apoptosis.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

References (0)

Have you cited this product in a publication?
Let us know so we can reference it here.
Cite this product

Bioinformatics

Protein Aliases: DAPK-interacting protein 1; DIP-1; E3 ubiquitin-protein ligase MIB1; Mind bomb homolog 1; RING-type E3 ubiquitin transferase MIB1; ubiquitin ligase mind bomb; Zinc finger ZZ type with ankyrin repeat domain protein 2

View more View less

Gene Aliases: DIP-1; DIP1; KIAA1323; LVNC7; MIB; MIB1; ZZANK2; ZZZ6

View more View less

UniProt ID: (Human) Q86YT6

View more View less

Entrez Gene ID: (Human) 57534

View more View less

Function(s)
ubiquitin-protein transferase activity protein binding zinc ion binding ligase activity
Process(es)
blood vessel development in utero embryonic development somitogenesis neural tube formation heart looping endocytosis Notch signaling pathway protein ubiquitination negative regulation of neuron differentiation positive regulation of endocytosis
It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-55c564d944-rwwh4:80/100.66.128.142:80.
git-commit: 3092084cb0e494f0e07f07633350183de7fa10a8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0