Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Primary Antibodies ›
  • MIB1 Antibodies

Invitrogen

MIB1 Polyclonal Antibody

View all (15) MIB1 antibodies

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Protocols
Questions & Answers
Datasheet
Protocols
Questions & Answers

Cite MIB1 Polyclonal Antibody

  • Antibody Testing Data (3)
MIB1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
Group 53 Created with Sketch.
MIB1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
Group 53 Created with Sketch.

FIGURE: 1 / 3

{{ $ctrl.currentElement.advancedVerification.fullName }}
{{ $ctrl.currentElement.advancedVerification.text }}

MIB1 Antibody (PA5-68450) in IHC (P)

Immunohistochemical analysis of MIB1 in paraffin embedded human kidney tissue using MIB1 Polyclonal Antibody (Product # PA5-68450) at a 1:200 dilution. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
Published figure supplied by benchsci-logo
PMID: {{$ctrl.currentElement.benchSciPubmedId}}
View Product

{{$ctrl.videoDesc}}

{{$ctrl.videoLongDesc}}

MIB1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
MIB1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
MIB1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
MIB1 Polyclonal Antibody

Product Details

PA5-68450

Applications
Tested Dilution
Publications

Western Blot (WB)

1:10,000
-

Immunohistochemistry (Paraffin) (IHC (P))

1:100-1:300
-
Product Specifications

Species Reactivity

Human, Mouse, Rat

Host/Isotype

Rabbit / IgG

Class

Polyclonal

Type

Antibody

Immunogen

Synthesized peptide derived from human MIB1 [aa 901-950].
View immunogen

Conjugate

Unconjugated Unconjugated Unconjugated

Form

Liquid

Concentration

1 mg/mL

Purification

affinity chromatography

Storage buffer

PBS, pH 7.4, with 50% glycerol, 0.5% BSA

Contains

0.02% sodium azide

Storage conditions

-20°C

Shipping conditions

Ambient (domestic); Wet ice (international)

RRID

AB_2688417

Product Specific Information

Predicted to react with Mouse and Rat samples.

Target Information

MIB is an E3 ubiquitin-protein ligase that mediates ubiquitination of Delta receptors, which act as ligands of Notch proteins. This protein positively regulates the Delta-mediated Notch signaling by ubiquitinating the intracellular domain of Delta, leading to endocytosis of Delta receptors. MIB probably mediates ubiquitination and subsequent proteasomal degradation of DAPK1, thereby antagonizing anti-apoptotic effects of DAPK1 to promote TNF-induced apoptosis.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

References (0)

Have you cited this product in a publication?
Let us know so we can reference it here.
Cite this product

Bioinformatics

Protein Aliases: DAPK-interacting protein 1; DAPK-interacting protein-1; DIP-1; E3 ubiquitin-protein ligase MIB1; Mind bomb homolog 1; mind bomb-1; mindbomb homolog 1; RING-type E3 ubiquitin transferase MIB1; skeletrophin; ubiquitin ligase mind bomb; Zinc finger ZZ type with ankyrin repeat domain protein 2

View more View less

Gene Aliases: DIP-1; DIP1; E430019M12Rik; KIAA1323; LVNC7; MIB; MIB1; mKIAA1323; ZZANK2; ZZZ6

View more View less

UniProt ID: (Human) Q86YT6, (Mouse) Q80SY4

View more View less

Entrez Gene ID: (Human) 57534, (Mouse) 225164, (Rat) 307594

View more View less

Function(s)
ubiquitin-protein transferase activity protein binding zinc ion binding ligase activity UDP-N-acetylmuramoylalanyl-D-glutamyl-2,6-diaminopimelate-D-alanyl-D-alanine ligase activity ribosomal S6-glutamic acid ligase activity coenzyme F420-0 gamma-glutamyl ligase activity coenzyme F420-2 alpha-glutamyl ligase activity metal ion binding protein-glycine ligase activity protein-glycine ligase activity, initiating protein-glycine ligase activity, elongating tubulin-glycine ligase activity protein-glutamic acid ligase activity tubulin-glutamic acid ligase activity molecular_function
Process(es)
blood vessel development in utero embryonic development somitogenesis neural tube formation heart looping endocytosis Notch signaling pathway heart development protein ubiquitination negative regulation of neuron differentiation positive regulation of endocytosis biological_process
It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-7887d5bf67-gxz2v:80/100.66.128.142:80.
git-commit: 1c5b77651f08bdd3ffd11596e5fe4ce144025943
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0