Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Primary Antibodies ›
  • LXR beta Antibodies

NeoBiotechnologies

Liver X Receptor beta (LXRB) (Orphan Receptor) Monoclonal Antibody (LXRB/2731)

View all (18) LXR beta antibodies

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Protocols
Questions & Answers
Datasheet
Protocols
Questions & Answers

Cite Liver X Receptor beta (LXRB) (Orphan Receptor) Monoclonal Antibody (LXRB/2731)

  • Antibody Testing Data (2)
Liver X Receptor beta (LXRB) (Orphan Receptor) Antibody in Peptide array (ARRAY)
Group 53 Created with Sketch.
Liver X Receptor beta (LXRB) (Orphan Receptor) Antibody in Peptide array (ARRAY)
Group 53 Created with Sketch.

FIGURE: 1 / 2

{{ $ctrl.currentElement.advancedVerification.fullName }}
{{ $ctrl.currentElement.advancedVerification.text }}

Liver X Receptor beta (LXRB) (Orphan Receptor) Antibody (7376-MSM1-P0) in ARRAY

Analysis of Protein Array containing >19,000 full-length human proteins using LX Receptor beta (NR1H2) Mouse Monoclonal Antibody (LXRB/2731) Z- and S- Score: The Z-score represents the strength of a signal that a monoclonal antibody (Monoclonal Antibody) (in combination with a fluorescently-tagged anti-IgG secondary antibody) produces when binding to a particular protein on the HuProtTM array. Z-scores are ... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
Published figure supplied by benchsci-logo
PMID: {{$ctrl.currentElement.benchSciPubmedId}}
View Product

{{$ctrl.videoDesc}}

{{$ctrl.videoLongDesc}}

Liver X Receptor beta (LXRB) (Orphan Receptor) Antibody in Peptide array (ARRAY)
Liver X Receptor beta (LXRB) (Orphan Receptor) Antibody in SDS-PAGE (SDS-PAGE)
Liver X Receptor beta (LXRB) (Orphan Receptor) Monoclonal Antibody (LXRB/2731)

Product Details

7376-MSM1-P0

Applications
Tested Dilution
Publications

ELISA (ELISA)

Assay-dependent
-

Peptide Array (Array)

Assay-dependent
-
Product Specifications

Species Reactivity

Human

Host/Isotype

Mouse / IgG2c, kappa

Class

Monoclonal

Type

Antibody

Clone

LXRB/2731

Immunogen

Recombinant full-length human NR1H2 (LXRB) protein
View immunogen

Conjugate

Unconjugated Unconjugated Unconjugated

Form

Liquid

Concentration

200 µg/mL

Purification

Protein A/G

Storage buffer

PBS, pH 7.4, with 0.05% BSA

Contains

0.05% sodium azide

Storage conditions

4°C, do not freeze

Shipping conditions

Wet ice

Target Information

The liver X receptors, LXRA and LXRB, form a subfamily of the nuclear receptor superfamily and are key regulators of macrophage function, controlling transcriptional programs involved in lipid homeostasis and inflammation. The inducible LXRA is highly expressed in liver, adrenal gland, intestine, adipose tissue, macrophages, lung, and kidney, whereas LXRB is ubiquitously expressed. Ligand-activated LXRs form obligate heterodimers with retinoid X receptors and regulate expression of target genes containing LXR response elements.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

References (0)

Have you cited this product in a publication?
Let us know so we can reference it here.
Cite this product

Bioinformatics

Protein Aliases: liver X nuclear receptor beta; Liver X receptor beta; LX receptor beta; nuclear orphan receptor LXR-beta; Nuclear receptor NER; Nuclear receptor subfamily 1 group H member 2; nuclear receptor subfamily 1, group H, member 2; Oxysterols receptor LXR-beta; steroid hormone-nuclear receptor NER; Ubiquitously-expressed nuclear receptor

View more View less

Gene Aliases: LXR-b; LXRB; NER; NER-I; NR1H2; RIP15; UNR

View more View less

UniProt ID: (Human) P55055

View more View less

Entrez Gene ID: (Human) 7376

View more View less

Function(s)
RNA polymerase II core promoter proximal region sequence-specific DNA binding transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding RNA polymerase II transcription factor activity, sequence-specific transcription regulatory region DNA binding DNA binding steroid hormone receptor activity RNA polymerase II transcription factor activity, ligand-activated sequence-specific DNA binding protein binding zinc ion binding apolipoprotein A-I receptor binding retinoid X receptor binding ATPase binding
Process(es)
negative regulation of transcription from RNA polymerase II promoter transcription initiation from RNA polymerase II promoter negative regulation of macrophage derived foam cell differentiation positive regulation of triglyceride biosynthetic process positive regulation of cholesterol efflux positive regulation of lipid storage negative regulation of cholesterol storage positive regulation of cellular protein metabolic process negative regulation of lipid transport positive regulation of cholesterol transport cholesterol homeostasis steroid hormone mediated signaling pathway cellular lipid metabolic process positive regulation of fatty acid biosynthetic process negative regulation of proteolysis negative regulation of transcription, DNA-templated positive regulation of transcription, DNA-templated positive regulation of transcription from RNA polymerase II promoter retinoic acid receptor signaling pathway negative regulation of pinocytosis positive regulation of lipoprotein lipase activity negative regulation of interferon-gamma-mediated signaling pathway positive regulation of high-density lipoprotein particle assembly positive regulation of pancreatic juice secretion positive regulation of secretion of lysosomal enzymes regulation of cholesterol homeostasis
It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-9dd846b7f-s9zn4:80/100.66.128.142:80.
git-commit: 17e8f6c92ed407c129a469238c057b56fdf8abbb
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0