Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Primary Antibodies ›
  • Lamin A Antibodies

Invitrogen

Lamin A Monoclonal Antibody (JOL4), FITC

View all (17) Lamin A antibodies

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Protocols
Questions & Answers
Datasheet
Protocols
Questions & Answers

Cite Lamin A Monoclonal Antibody (JOL4), FITC

Product Details

MA5-46604

Applications
Tested Dilution
Publications

Immunohistochemistry (Frozen) (IHC (F))

Assay-dependent
-

Immunocytochemistry (ICC/IF)

Assay-dependent
-
Product Specifications

Species Reactivity

Human, Mouse

Host/Isotype

Mouse / IgG

Class

Monoclonal

Type

Antibody

Clone

JOL4

Immunogen

Recombinant human lamin A

Conjugate

FITC FITC FITC

Excitation/Emission Max

498/517 nm View spectra spectra

Form

Liquid

Concentration

1 mg/mL

Purification

Protein G

Storage buffer

PBS

Contains

0.09% sodium azide

Storage conditions

Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. Store in the dark.

Shipping conditions

Ambient (domestic); Wet ice (international)

RRID

AB_2937676

Target Information

Nuclear lamins form a network of intermediate-type filaments at the nucleoplasmic site of the nuclear membrane. Two main subtypes of nuclear lamins can be distinguished, i.e. A-type lamins and B-type lamins. The A-type lamins comprise a set of three proteins arising from the same gene by alternative splicing, i.e. lamin A, lamin C and lamin Adel 10, while the B-type lamins include two proteins arising from two distinct genes, i.e. lamin B1 and lamin B2. Recent evidence has revealed that mutations in A-type lamins give rise to a range of rare but dominant genetic disorders, including Emery-Dreifuss muscular dystrophy, dilated cardiomyopathy with conduction-system disease and Dunnigan-type familial partial lipodystrophy. In addition, the expression of A-type lamins coincides with cell differentiation and as A-type lamins specifically interact with chromatin, a role in the regulation of differential gene expression has been suggested for A-type lamins.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

References (0)

Have you cited this product in a publication?
Let us know so we can reference it here.
Cite this product

Bioinformatics

Protein Aliases: 70 kDa lamin; lamin A/C-like 1; lamin C; lamin-A/C; OTTHUMP00000015843; OTTHUMP00000015846; OTTHUMP00000015847; Prelamin-A/C; renal carcinoma antigen NY-REN-32

View more View less

Gene Aliases: CDCD1; CDDC; CMD1A; CMT2B1; Dhe; EMD2; FPL; FPLD; FPLD2; HGPS; IDC; LDP1; LFP; LGMD1B; LMN1; LMNA; LMNC; LMNL1; PRO1

View more View less

UniProt ID: (Human) P02545, (Mouse) P48678

View more View less

Entrez Gene ID: (Human) 4000, (Mouse) 16905

View more View less

Function(s)
structural molecule activity protein binding protein phosphatase 1 binding
Process(es)
nucleus organization nuclear envelope organization muscle organ development establishment of cell polarity regulation of cell migration establishment or maintenance of microtubule cytoskeleton polarity protein localization to nucleus sterol regulatory element binding protein import into nucleus ventricular cardiac muscle cell development negative regulation of release of cytochrome c from mitochondria positive regulation of cell aging regulation of protein localization to nucleus negative regulation of extrinsic apoptotic signaling pathway mitotic nuclear envelope disassembly mitotic nuclear envelope reassembly spermatogenesis response to mechanical stimulus IRE1-mediated unfolded protein response positive regulation of osteoblast differentiation cellular response to hypoxia negative regulation of adipose tissue development
It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-9dd846b7f-s9zn4:80/100.66.128.142:80.
git-commit: 17e8f6c92ed407c129a469238c057b56fdf8abbb
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0