Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Primary Antibodies ›
  • LIPG Antibodies

OriGene

LIPG Monoclonal Antibody (OTI3C7), TrueMAB™

View all (75) LIPG antibodies

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Protocols
Questions & Answers
Datasheet
Protocols
Questions & Answers

Cite LIPG Monoclonal Antibody (OTI3C7), TrueMAB™

  • Antibody Testing Data (8)
LIPG Antibody in Immunocytochemistry (ICC/IF)
Group 53 Created with Sketch.
LIPG Antibody in Immunocytochemistry (ICC/IF)
Group 53 Created with Sketch.

FIGURE: 1 / 8

{{ $ctrl.currentElement.advancedVerification.fullName }}
{{ $ctrl.currentElement.advancedVerification.text }}

LIPG Antibody (TA501017) in ICC/IF

Anti-LIPG mouse monoclonal antibody (TA501017) immunofluorescent staining of COS7 cells transiently transfected by pCMV6-ENTRY LIPG(RC209248). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
Published figure supplied by benchsci-logo
PMID: {{$ctrl.currentElement.benchSciPubmedId}}
View Product

{{$ctrl.videoDesc}}

{{$ctrl.videoLongDesc}}

LIPG Antibody in Immunocytochemistry (ICC/IF)
LIPG Antibody in Immunohistochemistry (Paraffin) (IHC (P))
LIPG Antibody in Immunohistochemistry (Paraffin) (IHC (P))
LIPG Antibody in Immunohistochemistry (Paraffin) (IHC (P))
LIPG Antibody in Immunohistochemistry (Paraffin) (IHC (P))
LIPG Antibody in Western Blot (WB)
LIPG Antibody in Flow Cytometry (Flow)
LIPG Antibody in Immunoprecipitation (IP)
LIPG Monoclonal Antibody (OTI3C7), TrueMAB™

Product Details

TA501017

Applications
Tested Dilution
Publications

Western Blot (WB)

1:2,000
-

Immunohistochemistry (Paraffin) (IHC (P))

1:50
-

Immunocytochemistry (ICC/IF)

1:100
-

Flow Cytometry (Flow)

1:100
-

Immunoprecipitation (IP)

2 µg/500 µL
-
Product Specifications

Species Reactivity

Human

Host/Isotype

Mouse / IgG2a

Class

Monoclonal

Type

Antibody

Clone

OTI3C7

Immunogen

Full length human recombinant protein of human LIPG produced in HEK293T cell.
View immunogen

Conjugate

Unconjugated Unconjugated Unconjugated

Form

Liquid

Concentration

0.9 mg/mL

Purification

Affinity chromatography

Storage buffer

PBS with 1% BSA, 50% glycerol

Contains

0.02% sodium azide

Storage conditions

-20°C, Avoid Freeze/Thaw Cycles

Shipping conditions

Ambient (domestic); Wet ice (international)

Target Information

LIPG (Lipase G, Endohelial type) is an endothelial lipase which is a novel member of the lipoprotein lipase gene family. LIPG is synthesized by endothelial cells and functions at the site of synthesis. LIPG most likely plays a physiological role in modulating lipoprotein metabolism. Overexpressed LIPG has been shown to reduce HDL cholesterol levels, whereas an LIPG deficiency appears to be effective in elevating HDL cholesterol levels in animals. LIPG has substantial phospholipase activity and may be involved in lipoprotein metabolism and vascular biology. Also, LIPG is designated a member of the TG lipase family by its sequence and characteristic lid region which provides substrate specificity for enzymes of the TG lipase family. LIPG is a carboxyl esterase that hydrolyzes high density lipoproteins, emulsifies triglycerides, is essential for the efficient digestion of dietary fats and can bind to heparin.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

References (0)

Have you cited this product in a publication?
Let us know so we can reference it here.
Cite this product

Bioinformatics

Protein Aliases: EDL; Endothelial cell-derived lipase; Endothelial lipase; lipase, endothelial; lipoprotein lipase H; Phospholipase A1; PL

View more View less

Gene Aliases: EDL; EL; LIPG; PRO719; UNQ387/PRO719

View more View less

UniProt ID: (Human) Q9Y5X9

View more View less

Entrez Gene ID: (Human) 9388

View more View less

Function(s)
lipoprotein lipase activity phospholipase activity heparin binding phosphatidylcholine 1-acylhydrolase activity
Process(es)
lipid metabolic process response to nutrient cell proliferation phospholipid catabolic process positive regulation of high-density lipoprotein particle clearance positive regulation of cholesterol transport high-density lipoprotein particle remodeling cholesterol homeostasis reverse cholesterol transport lipid digestion regulation of lipoprotein metabolic process phospholipid homeostasis
It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-9dd846b7f-s9zn4:80/100.66.128.142:80.
git-commit: 17e8f6c92ed407c129a469238c057b56fdf8abbb
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0