Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Primary Antibodies ›
  • LHX6 Antibodies

Abnova

LHX6 Monoclonal Antibody (3E8)

View all (10) LHX6 antibodies

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Protocols
Questions & Answers
Datasheet
Protocols
Questions & Answers

Cite LHX6 Monoclonal Antibody (3E8)

  • Antibody Testing Data (2)
LHX6 Antibody in Western Blot (WB)
Group 53 Created with Sketch.
LHX6 Antibody in Western Blot (WB)
Group 53 Created with Sketch.

FIGURE: 1 / 2

{{ $ctrl.currentElement.advancedVerification.fullName }}
{{ $ctrl.currentElement.advancedVerification.text }}

LHX6 Antibody (H00026468-M05) in WB

LHX6 monoclonal antibody (M05), clone 3E8. Western Blot analysis of LHX6 expression in NIH/3T3 (Cat # L018V1). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
Published figure supplied by benchsci-logo
PMID: {{$ctrl.currentElement.benchSciPubmedId}}
View Product

{{$ctrl.videoDesc}}

{{$ctrl.videoLongDesc}}

LHX6 Antibody in Western Blot (WB)
LHX6 Antibody in Western Blot (WB)
LHX6 Monoclonal Antibody (3E8)

Product Details

H00026468-M05

Applications
Tested Dilution
Publications

Western Blot (WB)

1-5 µg/mL
-

ELISA (ELISA)

10 µg/mL
-
Product Specifications

Species Reactivity

Human, Mouse

Host/Isotype

Mouse / IgG2b, kappa

Class

Monoclonal

Type

Antibody

Clone

3E8

Immunogen

LHX6 (NP_055183, 274 a.a. approximately 363 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
View immunogen

Conjugate

Unconjugated Unconjugated Unconjugated

Form

Liquid

Concentration

See Label

Purification

Affinity chromatography

Storage buffer

PBS, pH 7.4

Contains

no preservative

Storage conditions

-20°C, Avoid Freeze/Thaw Cycles

Shipping conditions

Ambient (domestic); Wet ice (international)

Product Specific Information

Sequence of this protein is as follows: HKKHTPQHPV PPSGAPPSRL PSALSDDIHY TPFSSPERAR MVTLHGYIES QVQCGQVHCR LPYTAPPVHL KADMDGPLSN RGEKVILFQY

Target Information

LHX6 (LIM/homeobox protein Lhx6) is a probably transcription factor required for the expression of a subset of genes involved in interneurons migration and development. It functions in the specification of cortical interneuron subtypes and in the migration of GABAergic interneuron prescursors from the subpallium to the cerebral cortex.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

References (0)

Have you cited this product in a publication?
Let us know so we can reference it here.
Cite this product

Bioinformatics

Protein Aliases: LIM homeobox protein 6; LIM homeodomain protein 6.1; LIM-homeodomain protein 6.1; LIM/homeobox protein Lhx6; LIM/homeobox protein Lhx6.1; MGC119542; MGC119544; MGC119545; RP11-498E2.6

View more View less

Gene Aliases: LHX6; LHX6.1

View more View less

UniProt ID: (Human) Q9UPM6, (Mouse) Q9R1R0

View more View less

Entrez Gene ID: (Human) 26468, (Mouse) 16874

View more View less

Function(s)
transcription factor activity, sequence-specific DNA binding protein binding zinc ion binding sequence-specific DNA binding DNA binding metal ion binding
Process(es)
transcription, DNA-templated regulation of transcription, DNA-templated multicellular organism development nervous system development cerebral cortex radially oriented cell migration cerebral cortex tangential migration cerebral cortex GABAergic interneuron migration forebrain neuron fate commitment forebrain neuron development cerebral cortex neuron differentiation cell differentiation cell maturation
It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0