Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Primary Antibodies ›
  • LGR5 Antibodies

OriGene

LGR5 Monoclonal Antibody (OTI2A2), TrueMAB™

View all (84) LGR5 antibodies

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Protocols
Questions & Answers
Datasheet
Protocols
Questions & Answers

Cite LGR5 Monoclonal Antibody (OTI2A2), TrueMAB™

  • Antibody Testing Data (18)
LGR5 Antibody in Immunocytochemistry (ICC/IF)
Group 53 Created with Sketch.
LGR5 Antibody in Immunocytochemistry (ICC/IF)
Group 53 Created with Sketch.

FIGURE: 1 / 18

{{ $ctrl.currentElement.advancedVerification.fullName }}
{{ $ctrl.currentElement.advancedVerification.text }}

LGR5 Antibody (TA503316) in ICC/IF

Figure from citation: [Nature] cells expressing the stem cell marker Lgr5 was detected by immunostaining of hCECs in 2D culture. View Citation
Pubmed id: 27398792
Memo: Human; IF
{{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
Published figure supplied by benchsci-logo
PMID: {{$ctrl.currentElement.benchSciPubmedId}}
View Product

{{$ctrl.videoDesc}}

{{$ctrl.videoLongDesc}}

LGR5 Antibody in Immunocytochemistry (ICC/IF)
LGR5 Antibody in Immunocytochemistry (ICC/IF)
LGR5 Antibody in Immunocytochemistry (ICC/IF)
LGR5 Antibody in Immunocytochemistry (ICC/IF)
LGR5 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
LGR5 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
LGR5 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
LGR5 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
LGR5 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
LGR5 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
LGR5 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
LGR5 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
LGR5 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
LGR5 Antibody in Western Blot (WB)
LGR5 Antibody in Flow Cytometry (Flow)
LGR5 Antibody in Flow Cytometry (Flow)
LGR5 Antibody in Flow Cytometry (Flow)
LGR5 Antibody in Immunoprecipitation (IP)
LGR5 Monoclonal Antibody (OTI2A2), TrueMAB™

Product Details

TA503316

Applications
Tested Dilution
Publications

Western Blot (WB)

1:2,000
-

Immunohistochemistry (Paraffin) (IHC (P))

1:150
-

Immunocytochemistry (ICC/IF)

1:100
-

Flow Cytometry (Flow)

1:100
-

Immunoprecipitation (IP)

2 µg/500 µL
-
Product Specifications

Species Reactivity

Human, Mouse

Host/Isotype

Mouse / IgG1

Class

Monoclonal

Type

Antibody

Clone

OTI2A2

Immunogen

Human recombinant protein fragment corresponding to amino acids 250-550 of human LGR5 produced in HEK293T Cell.
View immunogen

Conjugate

Unconjugated Unconjugated Unconjugated
  • Biotin 
  • DyLight 488
  • DyLight 550
  • DyLight 633
  • HRP
  • PE 
  • Request custom conjugation

Form

Liquid

Concentration

1 mg/mL

Purification

Affinity chromatography

Storage buffer

PBS with 1% BSA, 50% glycerol

Contains

0.02% sodium azide

Storage conditions

-20°C, Avoid Freeze/Thaw Cycles

Shipping conditions

Ambient (domestic); Wet ice (international)

Target Information

G protein-coupled receptor GPR49 has been reported to be expressed in brain, skeletal muscle, placenta, and spinal cord. ESTs have been isolated from normal human brain (amygdala), embryo, and placenta libraries and from human germ cell, uterus, and brain libraries. G-protein Coupled Receptors (GPCRs) comprise one of the largest families of signaling molecules with more than a thousand members currently predicted to exist. All GPCRs share a structural motif consisting of seven membrane-spanning helices, and exist in both active and inactive forms. An array of activating ligands participate in the conformation of GPCRs which leads to signaling via G-proteins and downstream effectors. Ongoing studies have also shown the vast series of reactions which participate in the negative regulation of GPCRs. This "turn-off" activity has tremendous implications for the physiological action of the cell, and continues to drive pharmacological research for new drug candidates. Two blockbuster drugs which have been developed as GPCR-targeted pharmaceuticals are Zyprexa (Eli Lilly) and Claritin (Schering-Plough) which have multi-billion dollar shares of the mental health and allergy markets, respectively.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

References (0)

Have you cited this product in a publication?
Let us know so we can reference it here.
Cite this product

Bioinformatics

Protein Aliases: follicle expressed hormone; G protein-coupled receptor 49; G-protein coupled receptor 49; G-protein coupled receptor 67; G-protein coupled receptor HG38; leucine-rich repeat containing G protein-coupled receptor 5; leucine-rich repeat-containing G protein-coupled receptor 5; Leucine-rich repeat-containing G-protein coupled receptor 5; orphan G protein-coupled receptor HG38; Orphan G-protein coupled receptor FEX

View more View less

Gene Aliases: FEX; GPR49; GPR67; GRP49; HG38; LGR5

View more View less

UniProt ID: (Human) O75473, (Mouse) Q9Z1P4

View more View less

Entrez Gene ID: (Human) 8549, (Mouse) 14160

View more View less

Function(s)
transmembrane signaling receptor activity G-protein coupled receptor activity protein binding signal transducer activity protein-hormone receptor activity
Process(es)
hair follicle development signal transduction G-protein coupled receptor signaling pathway oocyte differentiation regulation of cell proliferation inner ear development positive regulation of canonical Wnt signaling pathway epithelial cell proliferation involved in renal tubule morphogenesis
It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-6d4c5dbbff-7nlnn:80/100.66.128.142:80.
git-commit: 3992c1c7134a3a3ff95f78ba9fe94218727d3638
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0