Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Primary Antibodies ›
  • CD171 (L1CAM) Antibodies

OriGene

L1CAM Monoclonal Antibody (OTI4F4), TrueMAB™

View all (90) CD171 (L1CAM) antibodies

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Protocols
Questions & Answers
Datasheet
Protocols
Questions & Answers

Cite L1CAM Monoclonal Antibody (OTI4F4), TrueMAB™

  • Antibody Testing Data (13)
L1CAM Antibody in Immunocytochemistry (ICC/IF)
Group 53 Created with Sketch.
L1CAM Antibody in Immunocytochemistry (ICC/IF)
Group 53 Created with Sketch.

FIGURE: 1 / 13

{{ $ctrl.currentElement.advancedVerification.fullName }}
{{ $ctrl.currentElement.advancedVerification.text }}

L1CAM Antibody (TA500800) in ICC/IF

Anti-L1CAM mouse monoclonal antibody (TA500800) immunofluorescent staining of COS7 cells transiently transfected by pCMV6-ENTRY L1CAM(RC211601). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
Published figure supplied by benchsci-logo
PMID: {{$ctrl.currentElement.benchSciPubmedId}}
View Product

{{$ctrl.videoDesc}}

{{$ctrl.videoLongDesc}}

L1CAM Antibody in Immunocytochemistry (ICC/IF)
L1CAM Antibody in Immunohistochemistry (Paraffin) (IHC (P))
L1CAM Antibody in Immunohistochemistry (Paraffin) (IHC (P))
L1CAM Antibody in Immunohistochemistry (Paraffin) (IHC (P))
L1CAM Antibody in Immunohistochemistry (Paraffin) (IHC (P))
L1CAM Antibody in Immunohistochemistry (Paraffin) (IHC (P))
L1CAM Antibody in Immunohistochemistry (Paraffin) (IHC (P))
L1CAM Antibody in Immunohistochemistry (Paraffin) (IHC (P))
L1CAM Antibody in Western Blot (WB)
L1CAM Antibody in Western Blot (WB)
L1CAM Antibody in Flow Cytometry (Flow)
L1CAM Antibody in Flow Cytometry (Flow)
L1CAM Antibody in Immunoprecipitation (IP)
L1CAM Monoclonal Antibody (OTI4F4), TrueMAB™

Product Details

TA500800

Applications
Tested Dilution
Publications

Western Blot (WB)

1:1,000-1:2,000
-

Immunohistochemistry (Paraffin) (IHC (P))

1:50
-

Immunocytochemistry (ICC/IF)

1:100
-

Flow Cytometry (Flow)

1:100
-

Immunoprecipitation (IP)

2-4 µg/mg
-
Product Specifications

Species Reactivity

Human

Host/Isotype

Mouse / IgG1

Class

Monoclonal

Type

Antibody

Clone

OTI4F4

Immunogen

Full length human recombinant protein of human L1CAM produced in HEK293T cell.
View immunogen

Conjugate

Unconjugated Unconjugated Unconjugated

Form

Liquid

Concentration

0.9 mg/mL

Purification

Affinity chromatography

Storage buffer

PBS with 1% BSA, 50% glycerol

Contains

0.02% sodium azide

Storage conditions

-20°C, Avoid Freeze/Thaw Cycles

Shipping conditions

Ambient (domestic); Wet ice (international)

Target Information

L1CAM/CD171 is an axonal glycoprotein belonging to the immunoglobulin supergene family. The ectodomain, consisting of several immunoglobulin-like domains and fibronectin-like repeats (type III), is linked via a single transmembrane sequence to a conserved cytoplasmic domain. This cell adhesion molecule plays an important role in nervous system development, including neuronal migration and differentiation. Mutations in the gene cause three X-linked neurological syndromes known by the acronym CRASH (corpus callosum hypoplasia, retardation, aphasia, spastic paraplegia and hydrocephalus). Alternative splicing of a neuron-specific exon is thought to be functionally relevant.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

References (0)

Have you cited this product in a publication?
Let us know so we can reference it here.
Cite this product

Bioinformatics

Protein Aliases: antigen identified by monoclonal antibody R1; CD 171; CD171; CD171 molecule; N-CAM-L1; Neural cell adhesion molecule L1; sCD171; sL1 CAM; sL1CAM; soluble CD 171; soluble CD171; soluble L1 CAM; soluble L1CAM

View more View less

Gene Aliases: CAML1; CD171; HSAS; HSAS1; L1CAM; MASA; MIC5; N-CAM-L1; N-CAML1; NCAM-L1; S10; SPG1

View more View less

UniProt ID: (Human) P32004

View more View less

Entrez Gene ID: (Human) 3897

View more View less

Process(es)
chemotaxis cell adhesion nervous system development axon guidance positive regulation of axon extension leukocyte migration
It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0