Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Primary Antibodies ›
  • Cytokeratin 8 Antibodies

OriGene

KRT8 Monoclonal Antibody (OTI2F5), TrueMAB™

View all (103) Cytokeratin 8 antibodies

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Protocols
Questions & Answers
Datasheet
Protocols
Questions & Answers

Cite KRT8 Monoclonal Antibody (OTI2F5), TrueMAB™

  • Antibody Testing Data (11)
KRT8 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
Group 53 Created with Sketch.
KRT8 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
Group 53 Created with Sketch.

FIGURE: 1 / 11

{{ $ctrl.currentElement.advancedVerification.fullName }}
{{ $ctrl.currentElement.advancedVerification.text }}

KRT8 Antibody (CF806987) in IHC (P)

Immunohistochemical staining of paraffin-embedded human pancreas tissue within the normal limits using anti-KRT8 mouse monoclonal antibody. (Heat-induced epitope retrieval by 1mM EDTA in 10mM Tris buffer (pH8.5) at 120°C for 3 min, TA806987)(1:2000). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
Published figure supplied by benchsci-logo
PMID: {{$ctrl.currentElement.benchSciPubmedId}}
View Product

{{$ctrl.videoDesc}}

{{$ctrl.videoLongDesc}}

KRT8 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
KRT8 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
KRT8 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
KRT8 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
KRT8 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
KRT8 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
KRT8 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
KRT8 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
KRT8 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
KRT8 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
KRT8 Antibody in Western Blot (WB)
KRT8 Monoclonal Antibody (OTI2F5), TrueMAB™

Product Details

CF806987

Applications
Tested Dilution
Publications

Immunohistochemistry (Paraffin) (IHC (P))

1:2,000
-
Product Specifications

Species Reactivity

Human

Host/Isotype

Mouse / IgG2b

Class

Monoclonal

Type

Antibody

Clone

OTI2F5

Immunogen

Human recombinant protein fragment corresponding to amino acids 91-381 of human KRT8 produced in E.coli.
View immunogen

Conjugate

Unconjugated Unconjugated Unconjugated

Form

Lyophilized

Concentration

1 mg/mL

Purification

Affinity chromatography

Storage buffer

PBS, pH 7.3, with 8% trehalose

Contains

no preservative

Storage conditions

-20°C, Avoid Freeze/Thaw Cycles

Shipping conditions

Ambient (domestic); Wet ice (international)

Product Specific Information

For reconstitution, we recommend adding 100 µL distilled water to a final antibody concentration of about 1 mg/mL. To use this carrier-free antibody for conjugation experiments, we strongly recommend performing another round of desalting. (Zeba Spin Desalting Columns, 7KMWCO, 0.5 mL, Product # 89882)

Target Information

Cytokeratin 8 is a member of the type II keratin family clustered on the long arm of chromosome 12. Type I and type II keratins heteropolymerize to form intermediate-sized filaments in the cytoplasm of epithelial cells. Cutokeratin 8 typically dimerizes with keratin 18 to form an intermediate filament in simple single-layered epithelial cells. This protein plays a role in maintaining cellular structural integrity and also functions in signal transduction and cellular differentiation. Mutations in this gene cause cryptogenic cirrhosis.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

References (0)

Have you cited this product in a publication?
Let us know so we can reference it here.
Cite this product

Bioinformatics

Protein Aliases: CK-8; Cytokeratin-8; K8; keratin 8, type II; Keratin, type II cytoskeletal 8; Keratin-8; Type-II keratin Kb8

View more View less

Gene Aliases: CARD2; CK-8; CK8; CYK8; K2C8; K8; KO; KRT8

View more View less

UniProt ID: (Human) P05787

View more View less

Entrez Gene ID: (Human) 3856

View more View less

Function(s)
structural molecule activity protein binding protein complex binding scaffold protein binding
Process(es)
viral process tumor necrosis factor-mediated signaling pathway sarcomere organization response to hydrostatic pressure response to other organism cell differentiation involved in embryonic placenta development extrinsic apoptotic signaling pathway hepatocyte apoptotic process
It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0