Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Primary Antibodies ›
  • KLF2 Antibodies

Bioss

KLF2 Polyclonal Antibody, FITC

View all (44) KLF2 antibodies

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Protocols
Questions & Answers
Datasheet
Protocols
Questions & Answers

Cite KLF2 Polyclonal Antibody, FITC

KLF2 Polyclonal Antibody, FITC

Product Details

BS-2772R-FITC

Applications
Tested Dilution
Publications

Western Blot (WB)

1:300-5,000
-

Immunohistochemistry (Paraffin) (IHC (P))

1:50-1:200
-

Immunohistochemistry (Frozen) (IHC (F))

1:50-1:200
-

Immunocytochemistry (ICC/IF)

1:50-1:200
-

Flow Cytometry (Flow)

1:20-1:100
-
Product Specifications

Species Reactivity

Human, Mouse, Rat

Host/Isotype

Rabbit / IgG

Class

Polyclonal

Type

Antibody

Immunogen

KLH conjugated synthetic peptide derived from human KLF2
View immunogen

Conjugate

FITC FITC FITC

Excitation/Emission Max

498/517 nm View spectra spectra

Form

Liquid

Concentration

1 mg/mL

Purification

Protein A

Storage buffer

TBS, pH 7.4, with 1% BSA, 50% glycerol

Contains

0.03% ProClin 300

Storage conditions

-20°C, store in dark

Shipping conditions

Ambient (domestic); Wet ice (international)

Target Information

KLF2 belongs to the krueppel C2H2-type zinc-finger protein family. It contains 3 C2H2-type zinc fingers. KLF2 binds to the CACCC box in the beta-globin gene promoter and activates transcription.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

How to use the Panel Builder

Watch the video to learn how to use the Invitrogen Flow Cytometry Panel Builder to build your next flow cytometry panel in 5 easy steps.

References (0)

Have you cited this product in a publication?
Let us know so we can reference it here.
Cite this product

Bioinformatics

Protein Aliases: Krueppel-like factor 2; Kruppel-like factor 2 (lung); Kruppel-like factor LKLF; Lung krueppel-like factor; lung Kruppel-like factor; lung Kruppel-like zinc finger transcription factor

View more View less

Gene Aliases: KLF2; LKLF

View more View less

UniProt ID: (Human) Q9Y5W3, (Mouse) Q60843, (Rat) Q9ET58

View more View less

Entrez Gene ID: (Human) 10365, (Mouse) 16598, (Rat) 306330

View more View less

Function(s)
transcription factor activity, sequence-specific DNA binding protein binding sequence-specific DNA binding metal ion binding nucleic acid binding DNA binding
Process(es)
cell morphogenesis in utero embryonic development transcription, DNA-templated regulation of transcription, DNA-templated negative regulation of interleukin-6 production erythrocyte homeostasis multicellular organism growth positive regulation of transcription from RNA polymerase II promoter in response to stress regulation of gene expression, epigenetic erythrocyte maturation positive regulation of transcription, DNA-templated positive regulation of transcription from RNA polymerase II promoter positive regulation of protein metabolic process Type I pneumocyte differentiation cellular response to laminar fluid shear stress response to laminar fluid shear stress cellular response to hydrogen peroxide cellular response to interleukin-1 cellular response to tumor necrosis factor cellular response to organic cyclic compound cellular response to cycloheximide cellular response to peptide positive regulation of nitric oxide biosynthetic process
It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-9dd846b7f-s9zn4:80/100.66.128.142:80.
git-commit: 17e8f6c92ed407c129a469238c057b56fdf8abbb
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0