Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Primary Antibodies ›
  • KIF11 Antibodies

Invitrogen

KIF11 Recombinant Rabbit Monoclonal Antibody (6G8V8)

View all (18) KIF11 antibodies

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Protocols
Questions & Answers
Datasheet
Protocols
Questions & Answers

Cite KIF11 Recombinant Rabbit Monoclonal Antibody (6G8V8)

  • Antibody Testing Data (1)
KIF11 Antibody in Western Blot (WB)
Group 53 Created with Sketch.
KIF11 Antibody in Western Blot (WB)
Group 53 Created with Sketch.

FIGURE: 1 / 1

{{ $ctrl.currentElement.advancedVerification.fullName }}
{{ $ctrl.currentElement.advancedVerification.text }}

KIF11 Antibody (MA5-35554) in WB

Western blot analysis of KIF11 in extracts of various cell lines. Samples were incubated with KIF11 Monoclonal antibody (Product # MA5-35554) using a dilution of 1:1,000, followed by HRP Goat Anti-Rabbit IgG (H+L) at a dilution of 1:10,000. Lysates/proteins: 25 µg per lane. Blocking buffer: 3% nonfat dry milk in TBST. Detection: ECL Basic Kit. Exposure time: 30s. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
Published figure supplied by benchsci-logo
PMID: {{$ctrl.currentElement.benchSciPubmedId}}
View Product

{{$ctrl.videoDesc}}

{{$ctrl.videoLongDesc}}

KIF11 Antibody in Western Blot (WB)
KIF11 Recombinant Rabbit Monoclonal Antibody (6G8V8)

Product Details

MA5-35554

Applications
Tested Dilution
Publications

Western Blot (WB)

1:1,000-1:4,000
-

ELISA (ELISA)

1 µg/mL
-
Product Specifications

Species Reactivity

Human

Host/Isotype

Rabbit / IgG

Expression System

HEK293 cells

Class

Recombinant Monoclonal

Type

Antibody

Clone

6G8V8

Immunogen

Recombinant fusion protein containing a sequence corresponding to amino acids 900-1056 of human EG5/KIF11 (P52732)
View immunogen

Conjugate

Unconjugated Unconjugated Unconjugated

Form

Liquid

Concentration

0.53 mg/mL

Purification

Affinity Chromatography

Storage buffer

PBS, pH 7.3, with 50% glycerol, 0.05% BSA

Contains

0.02% sodium azide

Storage conditions

-20°C, Avoid Freeze/Thaw Cycles

Shipping conditions

Ambient (domestic); Wet ice (international)

RRID

AB_2849454

Product Specific Information

Immunogen sequence: ETIKIGLTKL NCFLEQDLKL DIPTGTTPQR KSYLYPSTLV RTEPREHLLD QLKRKQPELL MMLNCSENNK EETIPDVDVE EAVLGQYTEE PLSQEPSVDA GVDCSSIGGV PFFQHKKSHG KDKENRGINT LERSKVEETT EHLVTKSRLP LRAQINL

Target Information

This gene encodes a motor protein that belongs to the kinesin-like protein family. Members of this protein family are known to be involved in various kinds of spindle dynamics. The function of this gene product includes chromosome positioning, centrosome separation and establishing a bipolar spindle during cell mitosis.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

References (0)

Have you cited this product in a publication?
Let us know so we can reference it here.
Cite this product

Bioinformatics

Protein Aliases: Kines; Kinesin-like protein 1; Kinesin-like protein KIF11; Kinesin-like spindle protein HKSP; Kinesin-related motor protein Eg5; Thyroid receptor-interacting protein 5; TR-interacting protein 5; TRIP-5

View more View less

Gene Aliases: EG5; HKSP; KIF11; KNSL1; MCLMR; TRIP5

View more View less

UniProt ID: (Human) P52732

View more View less

Entrez Gene ID: (Human) 3832

View more View less

Function(s)
microtubule motor activity ATP binding microtubule binding ATP-dependent microtubule motor activity, plus-end-directed protein kinase binding protein complex binding
Process(es)
retrograde vesicle-mediated transport, Golgi to ER microtubule-based movement spindle organization mitotic spindle organization chromosome segregation mitotic nuclear division mitotic centrosome separation antigen processing and presentation of exogenous peptide antigen via MHC class II regulation of mitotic centrosome separation cell division mitotic spindle assembly
It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-9dd846b7f-s9zn4:80/100.66.128.142:80.
git-commit: 17e8f6c92ed407c129a469238c057b56fdf8abbb
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0