Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Primary Antibodies ›
  • JAM-A (CD321) Antibodies

Invitrogen

JAM-A (CD321) Monoclonal Antibody (M.Ab.F11)

View all (36) JAM-A (CD321) antibodies

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Protocols
Questions & Answers
Datasheet
Protocols
Questions & Answers

Cite JAM-A (CD321) Monoclonal Antibody (M.Ab.F11)

JAM-A (CD321) Monoclonal Antibody (M.Ab.F11)

Product Details

MA5-49041

Applications
Tested Dilution
Publications

Western Blot (WB)

1:50
-

Immunohistochemistry (Paraffin) (IHC (P))

1:50
-

Immunocytochemistry (ICC/IF)

1:50
-

Flow Cytometry (Flow)

1:50
-

Immunoprecipitation (IP)

1:50
-

Functional Assay (Functional)

1:50
-

Inhibition Assays (IA)

1:50
-
Product Specifications

Species Reactivity

Human

Host/Isotype

Mouse / IgG1

Class

Monoclonal

Type

Antibody

Clone

M.Ab.F11

Immunogen

Human platelet membranes
View immunogen

Conjugate

Unconjugated Unconjugated Unconjugated

Form

Liquid

Concentration

100 µg/mL

Purification

Protein G

Storage buffer

PBS with 0.1% BSA

Contains

no preservative

Storage conditions

4°C

Shipping conditions

Ambient (domestic); Wet ice (international)

RRID

AB_3091066

Product Specific Information

The monoclonal antibody M.Ab.F11 recognizes junctional adhesion molecule-A (JAM-A) also known as the human platelet F11-Receptor (F11R) or JAM-1.

Target Information

CD321 (F11R, JAM-1, JAM-A) is a cell-surface adhesion molecule belonging to the Ig superfamily of receptors. CD321 is a 32-41 kD glycoprotein, possessing two extracellular Ig-like domains, a transmembrane region, and a short cytosolic tail. The protein assembles at the membrane as a homodimer, and can associate laterally with other membrane proteins including integrins. CD321 is expressed on a variety of cell types, including leukocytes, platelets, erythrocytes, stem cells, epithelial and endothelial cells, and plays a role in mediating cell-cell contact. CD321 can be found in close proximity to several components of epithelial tight junctions, including ZO-1 and occludin. CD321 is involved in the maintenance of epithelial and endothelial barriers, hematopoiesis, angiogenesis, and tumor proliferation, invasion, and metastasis.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

References (0)

Have you cited this product in a publication?
Let us know so we can reference it here.
Cite this product

Bioinformatics

Protein Aliases: CD321; F11 R; JAM-1; JAM-A; Junctional adhesion molecule 1; Junctional adhesion molecule A; PAM-1; Platelet adhesion molecule 1; Platelet F11 receptor

View more View less

Gene Aliases: CD321; F11R; JAM; JAM1; JAMA; JCAM; KAT; PAM-1; UNQ264/PRO301

View more View less

UniProt ID: (Human) Q9Y624

View more View less

Entrez Gene ID: (Human) 50848

View more View less

Function(s)
virus receptor activity protein binding PDZ domain binding cadherin binding involved in cell-cell adhesion
Process(es)
regulation of cytokine production inflammatory response transforming growth factor beta receptor signaling pathway response to radiation extracellular matrix organization actomyosin structure organization negative regulation of GTPase activity positive regulation of GTPase activity positive regulation of blood pressure viral entry into host cell intestinal absorption leukocyte migration bicellular tight junction assembly establishment of protein localization to plasma membrane establishment of endothelial intestinal barrier regulation of membrane permeability cell-cell adhesion regulation of actin cytoskeleton reorganization
It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0