Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Primary Antibodies ›
  • ISG20 Antibodies

Abnova

ISG20 Monoclonal Antibody (1B2-3C9)

View all (8) ISG20 antibodies

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Protocols
Questions & Answers
Datasheet
Protocols
Questions & Answers

Cite ISG20 Monoclonal Antibody (1B2-3C9)

  • Antibody Testing Data (1)
ISG20 Antibody in Western Blot (WB)
Group 53 Created with Sketch.
ISG20 Antibody in Western Blot (WB)
Group 53 Created with Sketch.

FIGURE: 1 / 1

{{ $ctrl.currentElement.advancedVerification.fullName }}
{{ $ctrl.currentElement.advancedVerification.text }}

ISG20 Antibody (H00003669-M01) in WB

Western Blot analysis of ISG20 expression in transfected 293T cell line by ISG20 monoclonal antibody (M01), clone 1B2-3C9. Lane 1: ISG20 transfected lysate(20.4 KDa). Lane 2: Non-transfected lysate. . {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
Published figure supplied by benchsci-logo
PMID: {{$ctrl.currentElement.benchSciPubmedId}}
View Product

{{$ctrl.videoDesc}}

{{$ctrl.videoLongDesc}}

ISG20 Antibody in Western Blot (WB)
ISG20 Monoclonal Antibody (1B2-3C9)

Product Details

H00003669-M01

Applications
Tested Dilution
Publications

Western Blot (WB)

1-5 µg/mL
-

Immunocytochemistry (ICC/IF)

10-100 µg/mL
-

ELISA (ELISA)

10 µg/mL
-
Product Specifications

Species Reactivity

Human

Host/Isotype

Mouse / IgG1, kappa

Class

Monoclonal

Type

Antibody

Clone

1B2-3C9

Immunogen

ISG20 (AAH07922.1, 1 a.a. approximately 181 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
View immunogen

Conjugate

Unconjugated Unconjugated Unconjugated

Form

Liquid

Concentration

1 mg/mL

Purification

Protein A

Storage buffer

PBS, pH 7.4

Contains

no preservative

Storage conditions

-20°C, Avoid Freeze/Thaw Cycles

Shipping conditions

Ambient (domestic); Wet ice (international)

Product Specific Information

Product may be used with Western Blot (Transfected lysate).

Immunogen sequence: MAGSREVVAM DCEMVGLGPH RESGLARCSL VNVHGAVLYD KFIRPEGEIT DYRTRVSGVT PQHMVGATPF AVARLEILQL LKGKLVVGHD LKHDFQALKE DMSGYTIYDT STDRLLWREA KLDHCRRVSL RVLSERLLHK SIQNSLLGHS SVEDARATME LYQISQRIRA RRGLPRLAVS D

Target Information

Interferon-induced antiviral exoribonuclease that acts on single-stranded RNA and also has minor activity towards single-stranded DNA. Exhibits antiviral activity against RNA viruses including hepatitis C virus (HCV), hepatitis A virus (HAV) and yellow fever virus (YFV) in an exonuclease-dependent manner. May also play additional roles in the maturation of snRNAs and rRNAs, and in ribosome biogenesis. [UniProt]

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

References (0)

Have you cited this product in a publication?
Let us know so we can reference it here.
Cite this product

Bioinformatics

Protein Aliases: Estrogen-regulated transcript 45 protein; interferon stimulated exonuclease gene 20kDa; Interferon-stimulated gene 20 kDa protein; Promyelocytic leukemia nuclear body-associated protein ISG20

View more View less

Gene Aliases: CD25; HEM45; ISG20

View more View less

UniProt ID: (Human) Q96AZ6

View more View less

Entrez Gene ID: (Human) 3669

View more View less

Function(s)
3'-5'-exoribonuclease activity exonuclease activity single-stranded DNA 3'-5' exodeoxyribonuclease activity exoribonuclease II activity U1 snRNA binding U2 snRNA binding U3 snoRNA binding metal ion binding
Process(es)
DNA catabolic process, exonucleolytic rRNA processing RNA catabolic process cell proliferation response to virus negative regulation of viral genome replication defense response to virus type I interferon signaling pathway RNA phosphodiester bond hydrolysis, exonucleolytic
It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0