Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Primary Antibodies ›
  • HSC70 Antibodies

Invitrogen

HSC70 Monoclonal Antibody (1F2-H5), FITC

View all (36) HSC70 antibodies

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Protocols
Questions & Answers
Datasheet
Protocols
Questions & Answers

Cite HSC70 Monoclonal Antibody (1F2-H5), FITC

  • Antibody Testing Data (4)
HSC70 Antibody in Immunocytochemistry (ICC/IF)
Group 53 Created with Sketch.
HSC70 Antibody in Immunocytochemistry (ICC/IF)
Group 53 Created with Sketch.

FIGURE: 1 / 4

{{ $ctrl.currentElement.advancedVerification.fullName }}
{{ $ctrl.currentElement.advancedVerification.text }}

HSC70 Antibody (MA5-45190) in ICC/IF

Immunocytochemistry/Immunofluorescence analysis using human heat shocked cervical cancer cells. Fixation involved 2% Formaldehyde for 20 min at RT. Samples were incubated with Hsc70 (Hsp73) monoclonal antibody (Product # MA5-45190) at 1:100 for 12 hours at 4°C, followed by R-PE Goat Anti-Mouse (yellow) at 1:200 for 2 hours at RT. Counterstain used was DAPI (blue) nuclear stain at 1:40,000 for 2 hours at RT. Localization: Cytoplasm. Melanosome. Localizes to nucleus upon heat shock. Magnification: 20x. (A) DAPI (blue) nuclear stain. (B) Anti... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
Published figure supplied by benchsci-logo
PMID: {{$ctrl.currentElement.benchSciPubmedId}}
View Product

{{$ctrl.videoDesc}}

{{$ctrl.videoLongDesc}}

HSC70 Antibody in Immunocytochemistry (ICC/IF)
HSC70 Antibody in Immunocytochemistry (ICC/IF)
HSC70 Antibody in Immunocytochemistry (ICC/IF)
HSC70 Antibody in Western Blot (WB)
HSC70 Monoclonal Antibody (1F2-H5), FITC

Product Details

MA5-45190

Applications
Tested Dilution
Publications

Western Blot (WB)

1:1,000
-

Immunohistochemistry (IHC)

Assay-dependent
-

Immunocytochemistry (ICC/IF)

1:100
-

ELISA (ELISA)

Assay-dependent
-

Immunoprecipitation (IP)

Assay-dependent
-

Functional Assay (Functional)

Assay-dependent
-

in situ PLA (PLA)

Assay-dependent
-

Immunomicroscopy (IM)

Assay-dependent
-

Antibody Microarray (AM)

Assay-dependent
-
Product Specifications

Species Reactivity

Human, Mouse, Rat

Host/Isotype

Mouse / IgG2a, kappa

Class

Monoclonal

Type

Antibody

Clone

1F2-H5

Immunogen

Full length human HSC70
View immunogen

Conjugate

FITC FITC FITC
  • APC 
  • PerCP 
  • Request custom conjugation

Excitation/Emission Max

498/517 nm View spectra spectra

Form

Liquid

Concentration

1 mg/mL

Purification

Protein G

Storage buffer

9.1mM sodium bicarbonate/PBS, pH 7.4, with 640.91mM DMSO, 136.36mM ethanolamine

Contains

no preservative

Storage conditions

4°C, store in dark

Shipping conditions

Ambient (domestic); Wet ice (international)

RRID

AB_2931644

Product Specific Information

1 µg/mL of MA5-45190 was sufficient for detection of HSC70 in 10 µg of HeLa lysate by colorimetric immunoblot analysis using Goat anti-mouse IgG:HRP as the secondary antibody.|Detects approximately 73kDa. Does not cross react with HSP70.

Target Information

The HSP70 family is a set of highly conserved proteins that are induced by a variety of biological stresses, including heat stress, in every organism in which the proteins have been examined. The human HSP70 family members include: HSP70, a protein which is strongly inducible in all organisms but which is also constitutively expressed in primate cells; HSP72, a 72 kDa protein that is induced exclusively under stress conditions; HSC70, or cognate protein, is a 72 kDa, constitutively expressed, protein which is involved in the uncoating of clathrin coated vesicles; GRP78, or BiP, is a glucose regulated 78 kDa protein localized in the endoplasmic reticulum; and p75, or HSP75, a 75 kDa protein that is found within the mitochondria. HSC70 (also known as HSC71, HSC73, HSP73, p72, prp73) is expressed constitutively and is slightly heat-inducible. HSC70 binds to the exposed loop of clathrin light chains to promote uncoating and can also bind the cytoskeleton which may facilitate cytoskeletal rearrangements. HSC70 has been shown to stimulate lysosomal degradation of intracellular proteins and to retard both aggregation and folding of mitochondrial precursor proteins in vitro.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

References (0)

Have you cited this product in a publication?
Let us know so we can reference it here.
Cite this product

Bioinformatics

Protein Aliases: AAG2; constitutive heat shock protein 70; epididymis luminal protein 33; epididymis secretory sperm binding protein Li 72p; FAM10A1; Heat shock 70 kDa protein 8; heat shock 70kd protein 10; heat shock 70kD protein 8; heat shock 70kDa protein 8; Heat shock cognate 71 kDa protein; heat shock cognate hsc73; heat shock cognate protein 54; Heat shock cognate protein 70; heat shock protein 8; heat shock protein A8; heat shock protein cognate 70; Heat shock protein family A member 8; HIP; LAP; LAP-1; Lipopolysaccharide-associated protein 1; LPS-associated protein 1; MGC131511; MGC29929; N-myristoyltransferase inhibitor protein 71; Protein FAM10A1; SNC6

View more View less

Gene Aliases: 2410008N15Rik; HEL-33; HEL-S-72p; HSC54; HSC70; HSC71; Hsc73; HSP71; HSP73; HSPA10; HSPA8; LAP-1; LAP1; NIP71

View more View less

UniProt ID: (Human) P11142, (Rat) P63018, (Mouse) P63017

View more View less

Entrez Gene ID: (Human) 3312, (Rat) 24468, (Mouse) 15481

View more View less

Function(s)
G-protein coupled receptor binding phosphatidylserine binding protein binding ATP binding ATPase activity enzyme binding MHC class II protein complex binding heat shock protein binding ubiquitin protein ligase binding ATPase activity, coupled poly(A) RNA binding unfolded protein binding C3HC4-type RING finger domain binding cadherin binding involved in cell-cell adhesion receptor binding ADP binding nucleotide binding
Process(es)
transcription, DNA-templated mRNA processing protein folding RNA splicing protein refolding negative regulation of transcription, DNA-templated positive regulation of mRNA splicing, via spliceosome chaperone mediated protein folding requiring cofactor regulation of cell cycle chaperone-mediated protein folding clathrin coat disassembly negative regulation of fibril organization mRNA splicing, via spliceosome protein targeting to lysosome response to unfolded protein neurotransmitter secretion cellular response to starvation viral process regulation of protein stability regulation of protein complex assembly regulation of mRNA stability ATP metabolic process regulation of protein complex stability chaperone-mediated autophagy late endosomal microautophagy protein targeting to lysosome involved in chaperone-mediated autophagy chaperone-mediated protein transport involved in chaperone-mediated autophagy cell-cell adhesion regulation of cellular response to heat regulation of protein import chaperone-mediated autophagy translocation complex disassembly regulation of transcription, DNA-templated
It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0