Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Primary Antibodies ›
  • HEYL Antibodies

OriGene

HEYL Monoclonal Antibody (OTI4D8), TrueMAB™

View all (19) HEYL antibodies

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Protocols
Questions & Answers
Datasheet
Protocols
Questions & Answers

Cite HEYL Monoclonal Antibody (OTI4D8), TrueMAB™

  • Antibody Testing Data (1)
HEYL Antibody in Western Blot (WB)
Group 53 Created with Sketch.
HEYL Antibody in Western Blot (WB)
Group 53 Created with Sketch.

FIGURE: 1 / 1

{{ $ctrl.currentElement.advancedVerification.fullName }}
{{ $ctrl.currentElement.advancedVerification.text }}

HEYL Antibody (TA812473) in WB

HEK293T cells were transfected with the pCMV6-ENTRY control (Left lane) or pCMV6-ENTRY HEYL (RC202851, Right lane) cDNA for 48 hrs and lysed. Equivalent amounts of cell lysates (5 µg per lane) were separated by SDS-PAGE and immunoblotted with anti-HEYL. (1. Positive lysates LY415208 (100 µg) and LC415208 (20 µg) can be purchased separately from OriGene. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
Published figure supplied by benchsci-logo
PMID: {{$ctrl.currentElement.benchSciPubmedId}}
View Product

{{$ctrl.videoDesc}}

{{$ctrl.videoLongDesc}}

HEYL Antibody in Western Blot (WB)
HEYL Monoclonal Antibody (OTI4D8), TrueMAB™

Product Details

TA812473

Applications
Tested Dilution
Publications

Western Blot (WB)

1:2,000
-
Product Specifications

Species Reactivity

Human

Host/Isotype

Mouse / IgG2b

Class

Monoclonal

Type

Antibody

Clone

OTI4D8

Immunogen

Full length human recombinant protein of human HEYL produced in E.coli.
View immunogen

Conjugate

Unconjugated Unconjugated Unconjugated

Form

Liquid

Concentration

1 mg/mL

Purification

Affinity chromatography

Storage buffer

PBS with 1% BSA, 50% glycerol

Contains

0.02% sodium azide

Storage conditions

-20°C, Avoid Freeze/Thaw Cycles

Shipping conditions

Ambient (domestic); Wet ice (international)

Target Information

This gene encodes a member of the hairy and enhancer of split-related (HESR) family of basic helix-loop-helix (bHLH)-type transcription factors. The sequence of the encoded protein contains a conserved bHLH and orange domain, but its YRPW motif has diverged from other HESR family members. It is thought to be an effector of Notch signaling and a regulator of cell fate decisions. Alternatively spliced transcript variants have been found, but their biological validity has not been determined.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

References (0)

Have you cited this product in a publication?
Let us know so we can reference it here.
Cite this product

Bioinformatics

Protein Aliases: bHLHb33; Class B basic helix-loop-helix protein 33; Hairy-related transcription factor 3; hairy/enhancer-of-split related with YRPW motif 3; Hairy/enhancer-of-split related with YRPW motif-like protein; hes-related family bHLH transcription factor with YRPW motif-like; HEY-like protein; hHeyL; hHRT3; HRT-3

View more View less

Gene Aliases: BHLHB33; HESR3; HEY3; HEYL; HRT3

View more View less

UniProt ID: (Human) Q9NQ87

View more View less

Entrez Gene ID: (Human) 26508

View more View less

Function(s)
RNA polymerase II core promoter sequence-specific DNA binding transcription factor activity, RNA polymerase II core promoter sequence-specific transcription factor activity, protein binding transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding RNA polymerase II transcription corepressor activity transcription factor activity, sequence-specific DNA binding transcription corepressor activity protein binding transcription factor binding microsatellite binding protein homodimerization activity protein heterodimerization activity AF-1 domain binding
Process(es)
negative regulation of transcription from RNA polymerase II promoter outflow tract morphogenesis atrioventricular valve morphogenesis pulmonary valve morphogenesis epithelial to mesenchymal transition involved in endocardial cushion formation endocardial cushion morphogenesis cardiac ventricle morphogenesis transcription from RNA polymerase II promoter Notch signaling pathway mesenchymal cell development glomerulus development skeletal muscle cell differentiation positive regulation of neuron differentiation negative regulation of transcription, DNA-templated positive regulation of transcription from RNA polymerase II promoter cardiac epithelial to mesenchymal transition ventricular septum morphogenesis negative regulation of androgen receptor signaling pathway cellular response to BMP stimulus proximal tubule development negative regulation of androgen receptor activity
It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-6d4c5dbbff-7nlnn:80/100.66.128.142:80.
git-commit: 3992c1c7134a3a3ff95f78ba9fe94218727d3638
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0