Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Primary Antibodies ›
  • GYS1 Antibodies

Proteintech

GYS1 Polyclonal Antibody

14 References
View all (25) GYS1 antibodies

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Protocols
Questions & Answers
Datasheet
Protocols
Questions & Answers

Cite GYS1 Polyclonal Antibody

  • Antibody Testing Data (12)
GYS1 Antibody in Immunocytochemistry (ICC/IF)
Group 53 Created with Sketch.
GYS1 Antibody in Immunocytochemistry (ICC/IF)
Group 53 Created with Sketch.

FIGURE: 1 / 12

{{ $ctrl.currentElement.advancedVerification.fullName }}
{{ $ctrl.currentElement.advancedVerification.text }}

GYS1 Antibody (10566-1-AP) in ICC/IF

Immunofluorescent analysis of ( -20°C Ethanol ) fixed HepG2 cells using 10566-1-AP (GYS1 antibody) at dilution of 1:50 and Alexa Fluor 488-conjugated AffiniPure Goat Anti-Rabbit IGG (H+L). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
Published figure supplied by benchsci-logo
PMID: {{$ctrl.currentElement.benchSciPubmedId}}
View Product

{{$ctrl.videoDesc}}

{{$ctrl.videoLongDesc}}

GYS1 Antibody in Immunocytochemistry (ICC/IF)
GYS1 Antibody in Immunocytochemistry (ICC/IF)
GYS1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
GYS1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
GYS1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
GYS1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
GYS1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
GYS1 Antibody in Western Blot (WB)
GYS1 Antibody in Western Blot (WB)
GYS1 Antibody in Western Blot (WB)
GYS1 Antibody in Flow Cytometry (Flow)
GYS1 Antibody in Immunoprecipitation (IP)
GYS1 Polyclonal Antibody

Product Details

10566-1-AP

Applications
Tested Dilution
Publications

Western Blot (WB)

1:1,000-1:4,000
View 9 publications 9 publications

Immunohistochemistry (IHC)

-
View 3 publications 3 publications

Immunohistochemistry (Paraffin) (IHC (P))

1:50-1:500
-

Immunocytochemistry (ICC/IF)

1:50-1:500
View 1 publication 1 publication

Flow Cytometry (Flow)

0.2 µg/1x10^6 cells
-

Immunoprecipitation (IP)

0.5-4.0 µg
-

Not applicable (N/A)

-
View 1 publication 1 publication
Product Specifications

Species Reactivity

Human, Mouse, Rat

Published species

Mouse, Rat

Host/Isotype

Rabbit / IgG

Class

Polyclonal

Type

Antibody

Immunogen

GYS1 Fusion Protein Ag0857 (430-737 aa encoded by BC007688)
View immunogen

Conjugate

Unconjugated Unconjugated Unconjugated

Form

Liquid

Concentration

0.36 mg/mL

Purification

Antigen affinity chromatography

Storage buffer

PBS, pH 7.3, with 50% glycerol

Contains

0.02% sodium azide

Storage conditions

-20°C

Shipping conditions

Wet ice (domestic); Dry ice (international)

Product Specific Information

Immunogen sequence: RAIFATQRQ SFPPVCTHNM LDDSSDPILT TIRRIGLFNS SADRVKVIFH PEFLSSTSPL LPVDYEEFVR GCHLGVFPSY YEPWGYTPAE CTVMGIPSIS TNLSGFGCFM EEHIADPSAY GIYILDRRFR SLDDSCSQLT SFLYSFCQQS RRQRIIQRNR TERLSDLLDW KYLGRYYMSA RHMALSKAFP EHFTYEPNEA DAAQGYRYPR PASVPPSPSL SRHSSPHQSE DEEDPRNGPL EEDGERYDED EEAAKDRRNI RAPEWPRRAS CTSSTSGSKR NSVDTATSSS LSTPSEPLSP TSSLGEERN (430-737 aa encoded by BC007688)

Target Information

Glycogen Synthase 1 (GYS1) catalyzes the addition of glucose monomers to the growing glycogen molecule through the formation of alpha-1,4-glycoside linkages. Mutations in this gene are associated with muscle glycogen storage disease. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

Bioinformatics

Protein Aliases: EC 2.4.1.11; Glycogen [starch] synthase, muscle; glycogen synthase; Glycogen synthase 1; glycogen synthase 1 (muscle); glycogen synthase 3, brain

View more View less

Gene Aliases: GSY; GYS; GYS1; Gys3; MGS

View more View less

UniProt ID: (Human) P13807, (Rat) A2RRU1, (Mouse) Q9Z1E4

View more View less

Entrez Gene ID: (Human) 2997, (Rat) 690987, (Mouse) 14936

View more View less

Function(s)
glycogen (starch) synthase activity protein binding glucose binding protein kinase binding glycogen synthase activity, transferring glucose-1-phosphate catalytic activity transferase activity transferase activity, transferring glycosyl groups
Process(es)
glycogen metabolic process glycogen biosynthetic process heart development metabolic process
It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-55c564d944-rwwh4:80/100.66.128.142:80.
git-commit: 3092084cb0e494f0e07f07633350183de7fa10a8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0