Disclaimer

Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

  • Primary Antibodies ›
  • HSPA9 Antibodies

Proteintech

GRP75 Monoclonal Antibody (2B12F2)

View all (33) HSPA9 antibodies

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

Datasheet
Protocols
Questions & Answers
Datasheet
Protocols
Questions & Answers

Cite GRP75 Monoclonal Antibody (2B12F2)

  • Antibody Testing Data (2)
GRP75 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
Group 53 Created with Sketch.
GRP75 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
Group 53 Created with Sketch.

FIGURE: 1 / 2

{{ $ctrl.currentElement.advancedVerification.fullName }}
{{ $ctrl.currentElement.advancedVerification.text }}

GRP75 Antibody (67563-1-IG) in IHC (P)

Immunohistochemical analysis of paraffin-embedded human breast cancer tissue slide using 67563-1-Ig (GRP75 antibody) at dilution of 1:1000 (under 10x lens). Heat mediated antigen retrieval with Tris-EDTA buffer (pH 9.0). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
Published figure supplied by benchsci-logo
PMID: {{$ctrl.currentElement.benchSciPubmedId}}
View Product

{{$ctrl.videoDesc}}

{{$ctrl.videoLongDesc}}

GRP75 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
GRP75 Antibody in Western Blot (WB)

Product Details

67563-1-IG

Applications
Tested Dilution
Publications

Western Blot (WB)

1:10,000
-

Immunohistochemistry (Paraffin) (IHC (P))

1:1,000
-
Product Specifications

Species Reactivity

Human, Mouse, Rat

Host/Isotype

Mouse / IgG2a

Class

Monoclonal

Type

Antibody

Clone

2B12F2

Immunogen

GRP75 Fusion Protein Ag7125

Conjugate

Unconjugated Unconjugated Unconjugated
  • CoraLite 594
  • Request custom conjugation

Form

Liquid

Concentration

1000 µg/mL

Purification

Protein A

Storage buffer

PBS, pH 7.3, with 50% glycerol

Contains

0.02% sodium azide

Storage conditions

-20°C

Shipping conditions

Wet ice

Product Specific Information

Aliquoting is unnecessary for -20°C storage.

Target Information

The HSP70 family is composed of four highly conserved proteins: HSP70, HSC70, GRP75 and GRP78. These proteins serve a variety of roles. GRP75 expression is restricted to the mitochondrial matrix and aids in the translocation and folding of nascent polypeptide chains of both nuclear and mitochondrial origin. GRP75 and GRP78 are unresponsive to heat stress and are induced by glucose deprivation.

For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

References (0)

Have you cited this product in a publication?
Let us know so we can reference it here.
Cite this product

Bioinformatics

Protein Aliases: 75 kDa glucose-regulated protein; C3H-specific antigen; catecholamine-regulated protein 40; epididymis secretory sperm binding protein Li 124m; GRP 75; GRP-75; Heat shock 70 kDa protein 9; heat shock 70kD protein 9B; heat shock 70kDa protein 9 (mortalin); heat shock 70kDa protein 9A; Heat shock protein; heat shock protein 9A; heat shock protein cognate 74; Heat shock protein family A member 9; heat shock protein, 74 kDa, A; heat shock protein, A; HSP; MGC4500; Mortalin; mortalin, perinuclear; mortalin-2; mortalin2; MOT; mtHSP70; p66 MOT; p66-mortalin; PBP74; Peptide-binding protein 74; Stress-70 protein, mitochondrial

View more View less

Gene Aliases: 74kDa; CRP40; CSA; EVPLS; GRP-75; GRP75; HEL-S-124m; Hsc74; Hsp74; Hsp74a; HSPA9; Hspa9a; HSPA9B; Mortalin; MOT; Mot-2; MOT2; mt-HSP70; Mthsp70; MTHSP75; PBP74; SAAN; SIDBA4

View more View less

UniProt ID: (Human) P38646, (Rat) P48721, (Mouse) P38647

View more View less

Entrez Gene ID: (Human) 3313, (Rat) 291671, (Mouse) 15526

View more View less

Function(s)
protein binding ATP binding fibroblast growth factor binding heat shock protein binding ubiquitin protein ligase binding poly(A) RNA binding unfolded protein binding enzyme binding nucleotide binding
Process(es)
protein folding protein export from nucleus response to toxic substance cellular response to interleukin-1 erythrocyte differentiation negative regulation of apoptotic process negative regulation of erythrocyte differentiation
It has to be done as per old AB suggested Products section.
guarantee_icon

Performance Guarantee

If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

Learn more
help_icon

We're here to help

Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

Contact tech support

Your items have has been added!


Host server : magellan-search-6d4c5dbbff-7nlnn:80/100.66.128.142:80.
git-commit: 3992c1c7134a3a3ff95f78ba9fe94218727d3638
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0